Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1929626-1930135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNNT3
Entrez Gene ID
7140
Gene Name
troponin T3, fast skeletal type
Gene Aliases
TNTF
UniGene
Hs.73454
Gene Chromosome Location
Chr.11: 1919551-1938706 on Build GRCh38
Transcripts
NM_001042780.2 NM_001042781.2 NM_001042782.2 NM_001297646.1 NM_006757.3 XM_006718288.3 XM_006718290.3 XM_006718293.2 XM_006718294.3 XM_006718296.3 XM_006718299.2 XM_006718300.3 XM_006718302.3 XM_011520343.2 XM_017018205.1 XM_017018206.1 XM_017018207.1 XM_017018208.1 XM_017018209.1 XM_017018210.1 XM_017018211.1 XM_017018212.1 XM_017018213.1 XM_017018214.1 XM_017018215.1 XM_017018216.1 XM_017018217.1 XM_017018218.1 XM_017018219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7255675 c.117_119+56del59 p.? Chr.11: 1929826-1929884 on Build GRCh38
1353115 c.119G>T p.R40I Chr.11: 1929822-1929822 on Build GRCh38
1353116 c.113G>T p.R38I Chr.11: 1929822-1929822 on Build GRCh38
5826946 c.95G>T p.R32I Chr.11: 1929822-1929822 on Build GRCh38
7255674 c.123_125+56del59 p.? Chr.11: 1929826-1929884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989680290 Chr.11: 1929914-1929914 on Build GRCh38
rs779697850 Chr.11: 1929919-1929919 on Build GRCh38
rs749078640 Chr.11: 1929724-1929724 on Build GRCh38
rs537821594 Chr.11: 1929854-1929854 on Build GRCh38
rs993285995 Chr.11: 1929758-1929758 on Build GRCh38
rs1014084930 Chr.11: 1930039-1930039 on Build GRCh38
rs754605874 Chr.11: 1929790-1929790 on Build GRCh38
rs766127019 Chr.11: 1929776-1929776 on Build GRCh38
rs1000488078 Chr.11: 1930048-1930048 on Build GRCh38
rs561029272 Chr.11: 1930085-1930085 on Build GRCh38
rs998488410 Chr.11: 1930056-1930056 on Build GRCh38
rs2292473 Chr.11: 1929676-1929676 on Build GRCh38
rs375241620 Chr.11: 1929731-1929731 on Build GRCh38
rs376551043 Chr.11: 1929872-1929872 on Build GRCh38
rs977284089 Chr.11: 1930057-1930057 on Build GRCh38
rs757778789 Chr.11: 1929802-1929802 on Build GRCh38
rs941983409 Chr.11: 1929938-1929938 on Build GRCh38
rs1031919212 Chr.11: 1930054-1930054 on Build GRCh38
rs369979472 Chr.11: 1929728-1929728 on Build GRCh38
rs372200170 Chr.11: 1929767-1929767 on Build GRCh38
rs970961115 Chr.11: 1929670-1929670 on Build GRCh38
rs2668864 Chr.11: 1930064-1930064 on Build GRCh38
rs181586935 Chr.11: 1929878-1929878 on Build GRCh38
rs2292472 Chr.11: 1929693-1929693 on Build GRCh38
rs770168335 Chr.11: 1929819-1929819 on Build GRCh38
rs2292471 Chr.11: 1929695-1929695 on Build GRCh38
rs139903501 Chr.11: 1929884-1929884 on Build GRCh38
rs1005398551 Chr.11: 1929797-1929797 on Build GRCh38
rs978995795 Chr.11: 1929837-1929837 on Build GRCh38
rs976894894 Chr.11: 1929891-1929891 on Build GRCh38
rs934820378 Chr.11: 1929860-1929860 on Build GRCh38
rs200287895 Chr.11: 1929815-1929815 on Build GRCh38
rs1014387704 Chr.11: 1929835-1929835 on Build GRCh38
rs536872922 Chr.11: 1929796-1929796 on Build GRCh38
rs778065947 Chr.11: 1929989-1929989 on Build GRCh38
rs556240627 Chr.11: 1929855-1929855 on Build GRCh38
rs1034418448 Chr.11: 1929943-1929943 on Build GRCh38
rs1055640223 Chr.11: 1930009-1930009 on Build GRCh38
rs760112135 Chr.11: 1929660-1929661 on Build GRCh38
rs746602543 Chr.11: 1929965-1929965 on Build GRCh38
rs922664549 Chr.11: 1929974-1929974 on Build GRCh38
rs545491880 Chr.11: 1929770-1929770 on Build GRCh38
rs941236771 Chr.11: 1930000-1930000 on Build GRCh38
rs1045479363 Chr.11: 1930040-1930040 on Build GRCh38
rs765067938 Chr.11: 1929937-1929937 on Build GRCh38
rs548081704 Chr.11: 1929723-1929723 on Build GRCh38
rs111660483 Chr.11: 1929692-1929693 on Build GRCh38
rs775771155 Chr.11: 1929839-1929839 on Build GRCh38
rs551937802 Chr.11: 1930091-1930091 on Build GRCh38
rs922726561 Chr.11: 1930071-1930071 on Build GRCh38
rs878951719 Chr.11: 1929772-1929772 on Build GRCh38
rs924192506 Chr.11: 1929692-1929692 on Build GRCh38
rs1037564919 Chr.11: 1929958-1929959 on Build GRCh38
rs549796494 Chr.11: 1929682-1929682 on Build GRCh38
rs373551064 Chr.11: 1929820-1929820 on Build GRCh38
rs745945471 Chr.11: 1930111-1930111 on Build GRCh38
rs386749741 Chr.11: 1929693-1929695 on Build GRCh38
rs547818719 Chr.11: 1929789-1929789 on Build GRCh38
rs959767792 Chr.11: 1929769-1929769 on Build GRCh38
rs74049812 Chr.11: 1929980-1929980 on Build GRCh38
rs894332737 Chr.11: 1930016-1930016 on Build GRCh38
rs897652231 Chr.11: 1929756-1929756 on Build GRCh38
rs112606502 Chr.11: 1929694-1929694 on Build GRCh38
rs774903606 Chr.11: 1930110-1930110 on Build GRCh38
rs924702463 Chr.11: 1929668-1929668 on Build GRCh38
rs377385915 Chr.11: 1929904-1929904 on Build GRCh38
rs763270021 Chr.11: 1929854-1929860 on Build GRCh38
rs386749742 Chr.11: 1929854-1929855 on Build GRCh38
rs200739738 Chr.11: 1929804-1929804 on Build GRCh38
rs561929640 Chr.11: 1930047-1930047 on Build GRCh38
rs924795738 Chr.11: 1929853-1929853 on Build GRCh38
rs542144713 Chr.11: 1929990-1929990 on Build GRCh38
rs147883316 Chr.11: 1929859-1929859 on Build GRCh38
rs978900794 Chr.11: 1929665-1929665 on Build GRCh38
rs954243955 Chr.11: 1930080-1930080 on Build GRCh38
rs1031243041 Chr.11: 1929898-1929898 on Build GRCh38
rs766854638 Chr.11: 1929846-1929846 on Build GRCh38
rs1041053032 Chr.11: 1929732-1929732 on Build GRCh38
rs376576654 Chr.11: 1929794-1929794 on Build GRCh38

View Full Product Details