Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2650801-2651297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNQ1
Entrez Gene ID
3784
Gene Name
potassium voltage-gated channel subfamily Q member 1
Gene Aliases
ATFB1, ATFB3, JLNS1, KCNA8, KCNA9, KVLQT1, Kv1.9, Kv7.1, LQT, LQT1, RWS, SQT2, WRS
UniGene
Hs.95162
Gene Chromosome Location
Chr.11: 2444991-2849110 on Build GRCh38
Transcripts
NM_000218.2 NM_181798.1 NR_002728.3

Gene Symbol
KCNQ1OT1
Entrez Gene ID
10984
Gene Name
KCNQ1 opposite strand/antisense transcript 1 (non-protein coding)
Gene Aliases
KCNQ1-AS2, KCNQ10T1, Kncq1, KvDMR1, KvLQT1-AS, LIT1, NCRNA00012
UniGene
Hs.604823
Gene Chromosome Location
Chr.11: 2608328-2699998 on Build GRCh38
Transcripts
NM_000218.2 NM_181798.1 NR_002728.3

SNP Detail

SNP ID Chromosome Location
rs183092035 Chr.11: 2651047-2651047 on Build GRCh38
rs907106684 Chr.11: 2651263-2651263 on Build GRCh38
rs371526023 Chr.11: 2651116-2651116 on Build GRCh38
rs558318258 Chr.11: 2651235-2651235 on Build GRCh38
rs916442748 Chr.11: 2650881-2650881 on Build GRCh38
rs1048714296 Chr.11: 2650895-2650895 on Build GRCh38
rs931700859 Chr.11: 2650885-2650885 on Build GRCh38
rs902597855 Chr.11: 2650938-2650938 on Build GRCh38
rs887418086 Chr.11: 2650918-2650918 on Build GRCh38
rs77809463 Chr.11: 2650934-2650934 on Build GRCh38
rs560482895 Chr.11: 2651162-2651162 on Build GRCh38
rs199720985 Chr.11: 2650983-2650983 on Build GRCh38
rs944854489 Chr.11: 2650837-2650837 on Build GRCh38
rs1004768952 Chr.11: 2650932-2650932 on Build GRCh38
rs908910508 Chr.11: 2651189-2651189 on Build GRCh38
rs576671282 Chr.11: 2651259-2651259 on Build GRCh38
rs182524671 Chr.11: 2651204-2651204 on Build GRCh38
rs778100237 Chr.11: 2651275-2651275 on Build GRCh38
rs377650965 Chr.11: 2650965-2650968 on Build GRCh38
rs34514246 Chr.11: 2651202-2651203 on Build GRCh38
rs983494705 Chr.11: 2651153-2651153 on Build GRCh38
rs764700594 Chr.11: 2651026-2651026 on Build GRCh38
rs1053803643 Chr.11: 2650968-2650968 on Build GRCh38
rs900244972 Chr.11: 2650846-2650846 on Build GRCh38
rs767976684 Chr.11: 2651154-2651154 on Build GRCh38
rs922082155 Chr.11: 2651131-2651131 on Build GRCh38
rs981973713 Chr.11: 2651271-2651271 on Build GRCh38
rs79220673 Chr.11: 2651113-2651113 on Build GRCh38
rs773060305 Chr.11: 2650961-2650962 on Build GRCh38
rs1054866058 Chr.11: 2650872-2650872 on Build GRCh38
rs1012313979 Chr.11: 2651015-2651015 on Build GRCh38
rs916413087 Chr.11: 2651188-2651188 on Build GRCh38
rs966273544 Chr.11: 2651080-2651080 on Build GRCh38
rs170762 Chr.11: 2651007-2651007 on Build GRCh38
rs528787638 Chr.11: 2650855-2650855 on Build GRCh38
rs968028243 Chr.11: 2651031-2651031 on Build GRCh38
rs997275595 Chr.11: 2651075-2651075 on Build GRCh38
rs949489075 Chr.11: 2650922-2650922 on Build GRCh38
rs544529006 Chr.11: 2650944-2650944 on Build GRCh38
rs990730162 Chr.11: 2651176-2651176 on Build GRCh38
rs1007173745 Chr.11: 2651022-2651022 on Build GRCh38
rs1022318743 Chr.11: 2651029-2651029 on Build GRCh38
rs1035145438 Chr.11: 2650988-2650988 on Build GRCh38
rs143246270 Chr.11: 2650962-2650965 on Build GRCh38
rs950504643 Chr.11: 2651139-2651139 on Build GRCh38
rs940342818 Chr.11: 2651200-2651200 on Build GRCh38
rs1040164437 Chr.11: 2650839-2650839 on Build GRCh38
rs568986352 Chr.11: 2651067-2651067 on Build GRCh38

View Full Product Details