Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3217852-3218354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRGPRG
Entrez Gene ID
386746
Gene Name
MAS related GPR family member G
Gene Aliases
GPR169, MRGG
UniGene
Hs.730306
Gene Chromosome Location
Chr.11: 3217944-3218813 on Build GRCh38
Transcripts
NM_001164377.1 NR_027138.1

Gene Symbol
MRGPRG-AS1
Entrez Gene ID
283303
Gene Name
MRGPRG antisense RNA 1
Gene Aliases
C11orf36, HSD-40
UniGene
Hs.151154
Gene Chromosome Location
Chr.11: 3218332-3223131 on Build GRCh38
Transcripts
NM_001164377.1 NR_027138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7352934 c.785C>T p.P262L Chr.11: 3218029-3218029 on Build GRCh38
6402136 c.479A>T p.H160L Chr.11: 3218335-3218335 on Build GRCh38
5451132 c.642G>A p.S214S Chr.11: 3218172-3218172 on Build GRCh38
7352935 c.646T>A p.F216I Chr.11: 3218168-3218168 on Build GRCh38
5477552 c.843A>G p.G281G Chr.11: 3217971-3217971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781548506 Chr.11: 3218115-3218115 on Build GRCh38
rs745993411 Chr.11: 3218133-3218133 on Build GRCh38
rs549380371 Chr.11: 3218011-3218011 on Build GRCh38
rs972023467 Chr.11: 3218263-3218263 on Build GRCh38
rs75066302 Chr.11: 3217954-3217954 on Build GRCh38
rs1003762888 Chr.11: 3217935-3217935 on Build GRCh38
rs902883905 Chr.11: 3218145-3218145 on Build GRCh38
rs997703447 Chr.11: 3218186-3218188 on Build GRCh38
rs140657678 Chr.11: 3218075-3218075 on Build GRCh38
rs751192545 Chr.11: 3218001-3218001 on Build GRCh38
rs946998225 Chr.11: 3218129-3218129 on Build GRCh38
rs928534750 Chr.11: 3218048-3218048 on Build GRCh38
rs1052711949 Chr.11: 3218155-3218156 on Build GRCh38
rs953594559 Chr.11: 3218208-3218208 on Build GRCh38
rs964159690 Chr.11: 3218006-3218006 on Build GRCh38
rs749556885 Chr.11: 3217904-3217904 on Build GRCh38
rs761502759 Chr.11: 3218223-3218223 on Build GRCh38
rs760212108 Chr.11: 3218286-3218286 on Build GRCh38
rs567960842 Chr.11: 3218012-3218012 on Build GRCh38
rs1004154834 Chr.11: 3218207-3218207 on Build GRCh38
rs1016757298 Chr.11: 3218225-3218225 on Build GRCh38
rs562205457 Chr.11: 3217928-3217928 on Build GRCh38
rs928571104 Chr.11: 3218267-3218267 on Build GRCh38
rs980493158 Chr.11: 3218298-3218298 on Build GRCh38
rs774443415 Chr.11: 3218148-3218148 on Build GRCh38
rs959927931 Chr.11: 3218276-3218276 on Build GRCh38
rs145777944 Chr.11: 3218136-3218136 on Build GRCh38
rs900009503 Chr.11: 3218147-3218147 on Build GRCh38
rs527961677 Chr.11: 3218007-3218007 on Build GRCh38
rs779439513 Chr.11: 3218029-3218029 on Build GRCh38
rs980524208 Chr.11: 3218083-3218083 on Build GRCh38
rs891232625 Chr.11: 3218194-3218194 on Build GRCh38
rs938981509 Chr.11: 3218106-3218106 on Build GRCh38
rs973250468 Chr.11: 3218062-3218062 on Build GRCh38
rs1044749658 Chr.11: 3217932-3217932 on Build GRCh38
rs573891233 Chr.11: 3218299-3218299 on Build GRCh38
rs763908484 Chr.11: 3217989-3217989 on Build GRCh38
rs920342244 Chr.11: 3217886-3217886 on Build GRCh38
rs7942566 Chr.11: 3217885-3217885 on Build GRCh38
rs762934321 Chr.11: 3218191-3218191 on Build GRCh38
rs1052130153 Chr.11: 3218116-3218116 on Build GRCh38
rs368352907 Chr.11: 3218168-3218168 on Build GRCh38
rs75003193 Chr.11: 3218329-3218329 on Build GRCh38
rs771249640 Chr.11: 3218113-3218113 on Build GRCh38
rs559925638 Chr.11: 3217970-3217970 on Build GRCh38
rs1055525760 Chr.11: 3218084-3218084 on Build GRCh38
rs973730438 Chr.11: 3218256-3218256 on Build GRCh38
rs565968937 Chr.11: 3218211-3218211 on Build GRCh38
rs536671714 Chr.11: 3218216-3218216 on Build GRCh38
rs996860097 Chr.11: 3218152-3218152 on Build GRCh38
rs532869123 Chr.11: 3217934-3217934 on Build GRCh38
rs947202552 Chr.11: 3218322-3218322 on Build GRCh38
rs939118781 Chr.11: 3218333-3218333 on Build GRCh38
rs1027455923 Chr.11: 3218280-3218280 on Build GRCh38
rs1031252103 Chr.11: 3217895-3217895 on Build GRCh38
rs753166994 Chr.11: 3218051-3218051 on Build GRCh38
rs747196155 Chr.11: 3218103-3218103 on Build GRCh38
rs531900362 Chr.11: 3218066-3218066 on Build GRCh38
rs1029122236 Chr.11: 3218193-3218193 on Build GRCh38
rs966240339 Chr.11: 3217898-3217898 on Build GRCh38
rs753220186 Chr.11: 3218324-3218324 on Build GRCh38
rs377620678 Chr.11: 3218019-3218019 on Build GRCh38
rs904107684 Chr.11: 3217880-3217880 on Build GRCh38
rs1014773804 Chr.11: 3218031-3218031 on Build GRCh38
rs571765485 Chr.11: 3218107-3218107 on Build GRCh38
rs892710575 Chr.11: 3218128-3218128 on Build GRCh38
rs1029095207 Chr.11: 3217959-3217959 on Build GRCh38
rs767250318 Chr.11: 3218224-3218224 on Build GRCh38
rs915664933 Chr.11: 3218088-3218088 on Build GRCh38
rs117134877 Chr.11: 3218245-3218245 on Build GRCh38
rs1011062500 Chr.11: 3217985-3217985 on Build GRCh38
rs554193425 Chr.11: 3218139-3218139 on Build GRCh38

View Full Product Details