Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 35435713-35436218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAMR1
Entrez Gene ID
25891
Gene Name
peptidase domain containing associated with muscle regeneration 1
Gene Aliases
DKFZP586H2123, FP938, RAMP
UniGene
Hs.55044
Gene Chromosome Location
Chr.11: 35431827-35530300 on Build GRCh38
Transcripts
NM_001001991.2 NM_001282675.1 NM_001282676.1 NM_015430.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5372403 c.1236C>T p.P412P Chr.11: 35436051-35436051 on Build GRCh38
4032574 c.1349G>T p.G450V Chr.11: 35435938-35435938 on Build GRCh38
3739040 c.1226C>G p.P409R Chr.11: 35436061-35436061 on Build GRCh38
6989338 c.1221G>A p.K407K Chr.11: 35436066-35436066 on Build GRCh38
1260850 c.1310G>A p.R437H Chr.11: 35435977-35435977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747582660 Chr.11: 35436122-35436122 on Build GRCh38
rs745862258 Chr.11: 35435869-35435869 on Build GRCh38
rs781387405 Chr.11: 35435882-35435882 on Build GRCh38
rs866907728 Chr.11: 35436073-35436073 on Build GRCh38
rs772369288 Chr.11: 35436108-35436108 on Build GRCh38
rs759669866 Chr.11: 35435893-35435893 on Build GRCh38
rs369444570 Chr.11: 35435934-35435934 on Build GRCh38
rs750310632 Chr.11: 35435937-35435937 on Build GRCh38
rs761237310 Chr.11: 35436157-35436157 on Build GRCh38
rs748184583 Chr.11: 35435958-35435958 on Build GRCh38
rs763793747 Chr.11: 35435924-35435924 on Build GRCh38
rs182409377 Chr.11: 35435975-35435975 on Build GRCh38
rs550902790 Chr.11: 35436105-35436105 on Build GRCh38
rs765674640 Chr.11: 35436193-35436193 on Build GRCh38
rs772764778 Chr.11: 35436170-35436170 on Build GRCh38
rs779858370 Chr.11: 35435948-35435948 on Build GRCh38
rs769933994 Chr.11: 35435889-35435889 on Build GRCh38
rs760147353 Chr.11: 35435906-35435906 on Build GRCh38
rs774719698 Chr.11: 35435978-35435978 on Build GRCh38
rs908874880 Chr.11: 35435754-35435754 on Build GRCh38
rs144584162 Chr.11: 35436106-35436106 on Build GRCh38
rs764814705 Chr.11: 35436008-35436008 on Build GRCh38
rs549382933 Chr.11: 35435839-35435839 on Build GRCh38
rs926135181 Chr.11: 35435784-35435784 on Build GRCh38
rs777574092 Chr.11: 35435864-35435864 on Build GRCh38
rs373744569 Chr.11: 35435859-35435859 on Build GRCh38
rs750169012 Chr.11: 35436009-35436009 on Build GRCh38
rs978830382 Chr.11: 35435764-35435764 on Build GRCh38
rs774689346 Chr.11: 35435902-35435902 on Build GRCh38
rs771186153 Chr.11: 35435977-35435977 on Build GRCh38
rs147274037 Chr.11: 35436014-35436014 on Build GRCh38
rs748237749 Chr.11: 35435888-35435888 on Build GRCh38
rs767923770 Chr.11: 35435909-35435909 on Build GRCh38
rs994027510 Chr.11: 35435892-35435892 on Build GRCh38
rs760745792 Chr.11: 35435832-35435832 on Build GRCh38
rs752190283 Chr.11: 35435912-35435912 on Build GRCh38
rs754424067 Chr.11: 35435854-35435854 on Build GRCh38
rs749383182 Chr.11: 35436076-35436076 on Build GRCh38
rs896078916 Chr.11: 35435829-35435829 on Build GRCh38
rs776227782 Chr.11: 35436154-35436154 on Build GRCh38
rs143939242 Chr.11: 35435999-35435999 on Build GRCh38
rs751549041 Chr.11: 35436057-35436057 on Build GRCh38
rs777948907 Chr.11: 35435974-35435974 on Build GRCh38
rs773896155 Chr.11: 35435942-35435944 on Build GRCh38
rs368197789 Chr.11: 35435866-35435866 on Build GRCh38
rs949635366 Chr.11: 35435826-35435826 on Build GRCh38
rs149699546 Chr.11: 35435805-35435805 on Build GRCh38
rs756974642 Chr.11: 35435925-35435925 on Build GRCh38
rs779920293 Chr.11: 35436020-35436020 on Build GRCh38
rs776165955 Chr.11: 35435988-35435988 on Build GRCh38
rs767121585 Chr.11: 35435858-35435858 on Build GRCh38
rs371066175 Chr.11: 35435981-35435981 on Build GRCh38
rs377424823 Chr.11: 35436107-35436107 on Build GRCh38
rs997428628 Chr.11: 35435972-35435972 on Build GRCh38
rs983166943 Chr.11: 35435753-35435753 on Build GRCh38
rs140461507 Chr.11: 35436015-35436015 on Build GRCh38
rs142812833 Chr.11: 35436059-35436059 on Build GRCh38
rs138210503 Chr.11: 35435923-35435923 on Build GRCh38
rs768610282 Chr.11: 35435874-35435874 on Build GRCh38
rs191103623 Chr.11: 35435792-35435792 on Build GRCh38
rs1018671021 Chr.11: 35435953-35435953 on Build GRCh38
rs757143150 Chr.11: 35435865-35435865 on Build GRCh38
rs770854999 Chr.11: 35435836-35435836 on Build GRCh38
rs764907045 Chr.11: 35436165-35436165 on Build GRCh38
rs761162418 Chr.11: 35436148-35436149 on Build GRCh38
rs769883360 Chr.11: 35435960-35435960 on Build GRCh38
rs777895987 Chr.11: 35436074-35436074 on Build GRCh38
rs771374689 Chr.11: 35435898-35435898 on Build GRCh38
rs370153315 Chr.11: 35436143-35436143 on Build GRCh38
rs760237633 Chr.11: 35435992-35435992 on Build GRCh38
rs955318279 Chr.11: 35435907-35435907 on Build GRCh38
rs776106666 Chr.11: 35436115-35436115 on Build GRCh38
rs758294879 Chr.11: 35435938-35435938 on Build GRCh38
rs905528578 Chr.11: 35435840-35435840 on Build GRCh38
rs374368312 Chr.11: 35436000-35436000 on Build GRCh38
rs540469932 Chr.11: 35435798-35435798 on Build GRCh38
rs773452643 Chr.11: 35435891-35435891 on Build GRCh38
rs529350968 Chr.11: 35435831-35435831 on Build GRCh38
rs755770819 Chr.11: 35435862-35435862 on Build GRCh38

View Full Product Details