Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 36296093-36296604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR5L
Entrez Gene ID
79899
Gene Name
proline rich 5 like
Gene Aliases
PROTOR2
UniGene
Hs.19987
Gene Chromosome Location
Chr.11: 36296175-36465204 on Build GRCh38
Transcripts
NM_001160167.1

SNP Detail

SNP ID Chromosome Location
rs547395825 Chr.11: 36296420-36296420 on Build GRCh38
rs531961567 Chr.11: 36296343-36296343 on Build GRCh38
rs906260944 Chr.11: 36296210-36296210 on Build GRCh38
rs953841504 Chr.11: 36296398-36296398 on Build GRCh38
rs551620193 Chr.11: 36296247-36296247 on Build GRCh38
rs894766101 Chr.11: 36296251-36296251 on Build GRCh38
rs549177439 Chr.11: 36296340-36296340 on Build GRCh38
rs1025202121 Chr.11: 36296213-36296213 on Build GRCh38
rs117306079 Chr.11: 36296168-36296168 on Build GRCh38
rs752611666 Chr.11: 36296410-36296410 on Build GRCh38
rs921217865 Chr.11: 36296161-36296161 on Build GRCh38
rs1042151173 Chr.11: 36296208-36296208 on Build GRCh38
rs576297741 Chr.11: 36296151-36296151 on Build GRCh38
rs112955556 Chr.11: 36296497-36296497 on Build GRCh38
rs562632279 Chr.11: 36296140-36296140 on Build GRCh38
rs754474197 Chr.11: 36296395-36296395 on Build GRCh38
rs4543964 Chr.11: 36296118-36296118 on Build GRCh38
rs4534558 Chr.11: 36296184-36296184 on Build GRCh38
rs773814908 Chr.11: 36296436-36296437 on Build GRCh38
rs945011168 Chr.11: 36296202-36296202 on Build GRCh38
rs59569542 Chr.11: 36296505-36296505 on Build GRCh38
rs948842142 Chr.11: 36296567-36296567 on Build GRCh38
rs758435467 Chr.11: 36296438-36296438 on Build GRCh38
rs569426591 Chr.11: 36296341-36296341 on Build GRCh38
rs189185687 Chr.11: 36296543-36296543 on Build GRCh38
rs181879128 Chr.11: 36296574-36296574 on Build GRCh38
rs765259623 Chr.11: 36296362-36296362 on Build GRCh38
rs915111832 Chr.11: 36296434-36296434 on Build GRCh38
rs936813071 Chr.11: 36296253-36296253 on Build GRCh38
rs573743778 Chr.11: 36296117-36296117 on Build GRCh38
rs777584364 Chr.11: 36296167-36296167 on Build GRCh38
rs910966802 Chr.11: 36296321-36296321 on Build GRCh38
rs780199762 Chr.11: 36296542-36296542 on Build GRCh38
rs753362789 Chr.11: 36296345-36296345 on Build GRCh38
rs569694615 Chr.11: 36296480-36296480 on Build GRCh38
rs896796125 Chr.11: 36296346-36296346 on Build GRCh38
rs150737747 Chr.11: 36296474-36296474 on Build GRCh38
rs536343850 Chr.11: 36296557-36296557 on Build GRCh38
rs546551950 Chr.11: 36296220-36296220 on Build GRCh38
rs921104765 Chr.11: 36296363-36296363 on Build GRCh38
rs184859998 Chr.11: 36296173-36296173 on Build GRCh38
rs754588319 Chr.11: 36296349-36296349 on Build GRCh38
rs1003068687 Chr.11: 36296214-36296214 on Build GRCh38
rs989428790 Chr.11: 36296429-36296429 on Build GRCh38
rs992119902 Chr.11: 36296196-36296196 on Build GRCh38
rs1013184998 Chr.11: 36296255-36296255 on Build GRCh38
rs888287354 Chr.11: 36296456-36296456 on Build GRCh38
rs1034220902 Chr.11: 36296527-36296527 on Build GRCh38
rs552030519 Chr.11: 36296344-36296344 on Build GRCh38
rs72946196 Chr.11: 36296540-36296540 on Build GRCh38
rs966917915 Chr.11: 36296216-36296216 on Build GRCh38
rs755891564 Chr.11: 36296137-36296137 on Build GRCh38
rs141194682 Chr.11: 36296339-36296339 on Build GRCh38
rs370423604 Chr.11: 36296519-36296519 on Build GRCh38
rs560830649 Chr.11: 36296246-36296247 on Build GRCh38
rs1057319901 Chr.11: 36296245-36296245 on Build GRCh38
rs1027611721 Chr.11: 36296266-36296266 on Build GRCh38
rs777587541 Chr.11: 36296470-36296470 on Build GRCh38

View Full Product Details