Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 36400918-36401417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR5L
Entrez Gene ID
79899
Gene Name
proline rich 5 like
Gene Aliases
PROTOR2
UniGene
Hs.19987
Gene Chromosome Location
Chr.11: 36296175-36465204 on Build GRCh38
Transcripts
NM_001160167.1 NM_001160168.1 NM_001160169.1 NM_024841.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
366788 c.19C>A p.P7T Chr.11: 36401140-36401140 on Build GRCh38
311207 c.31G>T p.V11F Chr.11: 36401152-36401152 on Build GRCh38
6756922 c.126G>A p.R42R Chr.11: 36401247-36401247 on Build GRCh38
6756924 c.34G>A p.E12K Chr.11: 36401155-36401155 on Build GRCh38
926714 c.164G>T p.S55I Chr.11: 36401285-36401285 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914600771 Chr.11: 36401387-36401387 on Build GRCh38
rs781075146 Chr.11: 36401106-36401106 on Build GRCh38
rs141974589 Chr.11: 36401152-36401152 on Build GRCh38
rs763823669 Chr.11: 36401242-36401243 on Build GRCh38
rs750002750 Chr.11: 36401090-36401090 on Build GRCh38
rs570136912 Chr.11: 36401192-36401192 on Build GRCh38
rs373714586 Chr.11: 36401113-36401113 on Build GRCh38
rs753716576 Chr.11: 36401310-36401310 on Build GRCh38
rs777175298 Chr.11: 36401131-36401131 on Build GRCh38
rs200570972 Chr.11: 36401317-36401317 on Build GRCh38
rs552785140 Chr.11: 36401223-36401223 on Build GRCh38
rs532779202 Chr.11: 36401193-36401193 on Build GRCh38
rs537019006 Chr.11: 36401245-36401245 on Build GRCh38
rs182590817 Chr.11: 36401065-36401065 on Build GRCh38
rs779506672 Chr.11: 36401096-36401096 on Build GRCh38
rs762202439 Chr.11: 36401194-36401194 on Build GRCh38
rs778348921 Chr.11: 36400941-36400941 on Build GRCh38
rs150695913 Chr.11: 36401162-36401162 on Build GRCh38
rs541079651 Chr.11: 36401143-36401143 on Build GRCh38
rs148376692 Chr.11: 36400959-36400959 on Build GRCh38
rs376716246 Chr.11: 36401264-36401264 on Build GRCh38
rs765285957 Chr.11: 36401318-36401318 on Build GRCh38
rs763848236 Chr.11: 36401308-36401308 on Build GRCh38
rs756037225 Chr.11: 36401158-36401158 on Build GRCh38
rs757868880 Chr.11: 36401080-36401080 on Build GRCh38
rs774404892 Chr.11: 36401114-36401114 on Build GRCh38
rs975658823 Chr.11: 36401282-36401282 on Build GRCh38
rs762534222 Chr.11: 36401351-36401351 on Build GRCh38
rs372842527 Chr.11: 36401076-36401076 on Build GRCh38
rs767030698 Chr.11: 36401284-36401284 on Build GRCh38
rs773749653 Chr.11: 36401246-36401246 on Build GRCh38
rs113399672 Chr.11: 36401362-36401362 on Build GRCh38
rs754681841 Chr.11: 36401099-36401099 on Build GRCh38
rs958397660 Chr.11: 36401006-36401006 on Build GRCh38
rs144807940 Chr.11: 36401137-36401137 on Build GRCh38
rs749234323 Chr.11: 36401163-36401163 on Build GRCh38
rs758120062 Chr.11: 36401092-36401092 on Build GRCh38
rs962104537 Chr.11: 36401066-36401066 on Build GRCh38
rs765777630 Chr.11: 36401208-36401208 on Build GRCh38
rs561124348 Chr.11: 36401154-36401154 on Build GRCh38
rs576646468 Chr.11: 36401111-36401111 on Build GRCh38
rs545627506 Chr.11: 36401350-36401350 on Build GRCh38
rs755059207 Chr.11: 36401333-36401333 on Build GRCh38
rs59943655 Chr.11: 36400982-36400982 on Build GRCh38
rs752347347 Chr.11: 36401214-36401214 on Build GRCh38
rs780212600 Chr.11: 36401179-36401179 on Build GRCh38
rs906787671 Chr.11: 36401189-36401189 on Build GRCh38
rs567974104 Chr.11: 36401243-36401243 on Build GRCh38
rs574520307 Chr.11: 36400984-36400984 on Build GRCh38
rs970730314 Chr.11: 36401004-36401004 on Build GRCh38
rs535001252 Chr.11: 36401236-36401236 on Build GRCh38
rs773808726 Chr.11: 36401195-36401195 on Build GRCh38
rs376275153 Chr.11: 36401086-36401086 on Build GRCh38
rs554257119 Chr.11: 36400983-36400983 on Build GRCh38
rs78731804 Chr.11: 36401151-36401151 on Build GRCh38
rs955470634 Chr.11: 36401340-36401340 on Build GRCh38
rs750341104 Chr.11: 36401332-36401332 on Build GRCh38
rs1041921488 Chr.11: 36401136-36401136 on Build GRCh38
rs985377846 Chr.11: 36401058-36401058 on Build GRCh38
rs1003957084 Chr.11: 36401008-36401008 on Build GRCh38
rs762312605 Chr.11: 36401149-36401149 on Build GRCh38
rs9630171 Chr.11: 36401129-36401129 on Build GRCh38
rs775515443 Chr.11: 36401128-36401128 on Build GRCh38
rs140007778 Chr.11: 36401211-36401211 on Build GRCh38
rs1013016037 Chr.11: 36401175-36401175 on Build GRCh38
rs1031041979 Chr.11: 36401046-36401046 on Build GRCh38
rs543159489 Chr.11: 36401033-36401033 on Build GRCh38
rs772194112 Chr.11: 36401122-36401122 on Build GRCh38
rs566415633 Chr.11: 36401231-36401231 on Build GRCh38
rs376587095 Chr.11: 36401075-36401075 on Build GRCh38
rs768622754 Chr.11: 36401238-36401238 on Build GRCh38
rs763549439 Chr.11: 36401207-36401207 on Build GRCh38
rs201146236 Chr.11: 36401180-36401180 on Build GRCh38
rs768971599 Chr.11: 36401186-36401186 on Build GRCh38
rs753737289 Chr.11: 36401227-36401227 on Build GRCh38
rs375733955 Chr.11: 36401250-36401250 on Build GRCh38
rs1010966587 Chr.11: 36401239-36401239 on Build GRCh38
rs775078547 Chr.11: 36401299-36401299 on Build GRCh38
rs771993504 Chr.11: 36401166-36401166 on Build GRCh38
rs556661996 Chr.11: 36401307-36401307 on Build GRCh38
rs760878916 Chr.11: 36401074-36401074 on Build GRCh38
rs750971237 Chr.11: 36401140-36401140 on Build GRCh38
rs982745158 Chr.11: 36401347-36401347 on Build GRCh38
rs769605091 Chr.11: 36401221-36401221 on Build GRCh38
rs892498327 Chr.11: 36401212-36401212 on Build GRCh38
rs757260286 Chr.11: 36401230-36401230 on Build GRCh38
rs952223260 Chr.11: 36401045-36401045 on Build GRCh38
rs768861738 Chr.11: 36401130-36401130 on Build GRCh38
rs371553701 Chr.11: 36401268-36401268 on Build GRCh38
rs775915624 Chr.11: 36401072-36401072 on Build GRCh38
rs902916002 Chr.11: 36400961-36400961 on Build GRCh38
rs1029529257 Chr.11: 36401309-36401309 on Build GRCh38
rs330261 Chr.11: 36401242-36401242 on Build GRCh38
rs369294600 Chr.11: 36401098-36401098 on Build GRCh38
rs1012419266 Chr.11: 36401007-36401007 on Build GRCh38
rs762386719 Chr.11: 36401132-36401132 on Build GRCh38
rs777491542 Chr.11: 36401059-36401059 on Build GRCh38

View Full Product Details