Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45811479-45811977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35C1
Entrez Gene ID
55343
Gene Name
solute carrier family 35 member C1
Gene Aliases
CDG2C, FUCT1
UniGene
Hs.12211
Gene Chromosome Location
Chr.11: 45804072-45813016 on Build GRCh38
Transcripts
NM_001145265.1 NM_001145266.1 NM_018389.4 XM_011520202.2

SNP Detail

SNP ID Chromosome Location
rs951755252 Chr.11: 45811604-45811604 on Build GRCh38
rs140259547 Chr.11: 45811526-45811526 on Build GRCh38
rs951357006 Chr.11: 45811944-45811944 on Build GRCh38
rs929858090 Chr.11: 45811661-45811661 on Build GRCh38
rs145216107 Chr.11: 45811653-45811653 on Build GRCh38
rs1031203954 Chr.11: 45811825-45811825 on Build GRCh38
rs137953810 Chr.11: 45811797-45811797 on Build GRCh38
rs906977597 Chr.11: 45811792-45811792 on Build GRCh38
rs4362111 Chr.11: 45811516-45811516 on Build GRCh38
rs982939334 Chr.11: 45811613-45811613 on Build GRCh38
rs910057772 Chr.11: 45811620-45811620 on Build GRCh38
rs983032874 Chr.11: 45811827-45811827 on Build GRCh38
rs1042723264 Chr.11: 45811781-45811781 on Build GRCh38
rs749186871 Chr.11: 45811743-45811744 on Build GRCh38
rs575802365 Chr.11: 45811859-45811859 on Build GRCh38
rs886048316 Chr.11: 45811579-45811579 on Build GRCh38
rs538005835 Chr.11: 45811877-45811877 on Build GRCh38
rs546984925 Chr.11: 45811798-45811798 on Build GRCh38
rs924615304 Chr.11: 45811918-45811918 on Build GRCh38
rs921339888 Chr.11: 45811885-45811885 on Build GRCh38
rs1054185570 Chr.11: 45811729-45811729 on Build GRCh38
rs1139267 Chr.11: 45811749-45811749 on Build GRCh38
rs186869966 Chr.11: 45811673-45811673 on Build GRCh38
rs1054319905 Chr.11: 45811928-45811928 on Build GRCh38
rs867299792 Chr.11: 45811800-45811800 on Build GRCh38
rs539129340 Chr.11: 45811834-45811834 on Build GRCh38
rs974010453 Chr.11: 45811600-45811600 on Build GRCh38
rs903098145 Chr.11: 45811708-45811708 on Build GRCh38
rs573426566 Chr.11: 45811882-45811882 on Build GRCh38
rs886048318 Chr.11: 45811907-45811907 on Build GRCh38
rs919902203 Chr.11: 45811934-45811934 on Build GRCh38
rs567352248 Chr.11: 45811894-45811894 on Build GRCh38
rs754318164 Chr.11: 45811692-45811692 on Build GRCh38
rs762251457 Chr.11: 45811732-45811732 on Build GRCh38
rs934540294 Chr.11: 45811919-45811919 on Build GRCh38
rs911261773 Chr.11: 45811569-45811569 on Build GRCh38
rs1026431099 Chr.11: 45811817-45811817 on Build GRCh38
rs955514015 Chr.11: 45811505-45811505 on Build GRCh38
rs974061537 Chr.11: 45811909-45811909 on Build GRCh38
rs921234821 Chr.11: 45811680-45811680 on Build GRCh38
rs764662834 Chr.11: 45811641-45811641 on Build GRCh38
rs938442940 Chr.11: 45811681-45811681 on Build GRCh38
rs951572635 Chr.11: 45811819-45811819 on Build GRCh38
rs934613642 Chr.11: 45811716-45811716 on Build GRCh38
rs530754257 Chr.11: 45811560-45811560 on Build GRCh38
rs550862505 Chr.11: 45811649-45811649 on Build GRCh38
rs886048317 Chr.11: 45811863-45811863 on Build GRCh38
rs1000173290 Chr.11: 45811799-45811799 on Build GRCh38
rs975606336 Chr.11: 45811849-45811849 on Build GRCh38
rs1018321380 Chr.11: 45811863-45811863 on Build GRCh38
rs567007782 Chr.11: 45811833-45811833 on Build GRCh38
rs933993294 Chr.11: 45811684-45811684 on Build GRCh38
rs934108329 Chr.11: 45811896-45811896 on Build GRCh38
rs559233131 Chr.11: 45811858-45811858 on Build GRCh38
rs892976674 Chr.11: 45811736-45811736 on Build GRCh38
rs986839015 Chr.11: 45811543-45811543 on Build GRCh38
rs1046914139 Chr.11: 45811662-45811662 on Build GRCh38
rs904130744 Chr.11: 45811793-45811793 on Build GRCh38
rs942705519 Chr.11: 45811587-45811587 on Build GRCh38

View Full Product Details