Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45931996-45932503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF21A
Entrez Gene ID
51317
Gene Name
PHD finger protein 21A
Gene Aliases
BHC80, BM-006
UniGene
Hs.502458
Gene Chromosome Location
Chr.11: 45929319-46121434 on Build GRCh38
Transcripts
NM_001101802.1 NM_016621.3 XM_011520158.2 XM_017017893.1 XM_017017901.1

SNP Detail

SNP ID Chromosome Location
rs574421119 Chr.11: 45932268-45932268 on Build GRCh38
rs576837086 Chr.11: 45932022-45932022 on Build GRCh38
rs927777571 Chr.11: 45932191-45932191 on Build GRCh38
rs1007023398 Chr.11: 45932347-45932347 on Build GRCh38
rs556437922 Chr.11: 45932100-45932100 on Build GRCh38
rs574608208 Chr.11: 45932414-45932415 on Build GRCh38
rs954442002 Chr.11: 45932059-45932059 on Build GRCh38
rs1030044200 Chr.11: 45932327-45932327 on Build GRCh38
rs201941105 Chr.11: 45932144-45932148 on Build GRCh38
rs1051163025 Chr.11: 45932340-45932340 on Build GRCh38
rs540323992 Chr.11: 45932301-45932301 on Build GRCh38
rs935422353 Chr.11: 45932337-45932337 on Build GRCh38
rs990670276 Chr.11: 45932454-45932454 on Build GRCh38
rs1027756029 Chr.11: 45932175-45932175 on Build GRCh38
rs1018486774 Chr.11: 45932090-45932090 on Build GRCh38
rs960499562 Chr.11: 45932444-45932444 on Build GRCh38
rs978164197 Chr.11: 45932075-45932075 on Build GRCh38
rs752614428 Chr.11: 45932147-45932148 on Build GRCh38
rs983182129 Chr.11: 45932418-45932418 on Build GRCh38
rs939169610 Chr.11: 45932206-45932206 on Build GRCh38
rs941951424 Chr.11: 45932302-45932302 on Build GRCh38
rs948805815 Chr.11: 45932160-45932160 on Build GRCh38
rs559850728 Chr.11: 45932412-45932412 on Build GRCh38
rs964315141 Chr.11: 45932106-45932106 on Build GRCh38
rs898770789 Chr.11: 45932366-45932366 on Build GRCh38
rs142413568 Chr.11: 45932205-45932205 on Build GRCh38
rs996327864 Chr.11: 45932325-45932325 on Build GRCh38
rs562321892 Chr.11: 45932267-45932267 on Build GRCh38
rs1018454212 Chr.11: 45932363-45932363 on Build GRCh38
rs573949750 Chr.11: 45932063-45932063 on Build GRCh38
rs999119868 Chr.11: 45932356-45932356 on Build GRCh38
rs928712844 Chr.11: 45932187-45932187 on Build GRCh38
rs990701339 Chr.11: 45932238-45932238 on Build GRCh38
rs141686809 Chr.11: 45932148-45932152 on Build GRCh38
rs779461972 Chr.11: 45932052-45932052 on Build GRCh38
rs1027311293 Chr.11: 45932414-45932414 on Build GRCh38
rs891257254 Chr.11: 45932341-45932341 on Build GRCh38
rs916513413 Chr.11: 45932278-45932278 on Build GRCh38
rs916441854 Chr.11: 45932467-45932467 on Build GRCh38
rs576355672 Chr.11: 45932190-45932190 on Build GRCh38
rs950482054 Chr.11: 45932176-45932176 on Build GRCh38
rs949790431 Chr.11: 45932417-45932417 on Build GRCh38
rs1043715357 Chr.11: 45932318-45932318 on Build GRCh38
rs532133035 Chr.11: 45932436-45932436 on Build GRCh38
rs200068069 Chr.11: 45932143-45932146 on Build GRCh38
rs886333510 Chr.11: 45932259-45932259 on Build GRCh38
rs1007095663 Chr.11: 45932077-45932077 on Build GRCh38
rs746377849 Chr.11: 45932338-45932338 on Build GRCh38
rs990258398 Chr.11: 45932141-45932141 on Build GRCh38
rs902543012 Chr.11: 45932322-45932322 on Build GRCh38
rs973107468 Chr.11: 45932170-45932170 on Build GRCh38
rs1034776112 Chr.11: 45932430-45932430 on Build GRCh38
rs182477810 Chr.11: 45932069-45932069 on Build GRCh38
rs188698988 Chr.11: 45932148-45932148 on Build GRCh38
rs772509051 Chr.11: 45932361-45932361 on Build GRCh38
rs946682944 Chr.11: 45932294-45932294 on Build GRCh38

View Full Product Details