Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45933973-45934470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF21A
Entrez Gene ID
51317
Gene Name
PHD finger protein 21A
Gene Aliases
BHC80, BM-006
UniGene
Hs.502458
Gene Chromosome Location
Chr.11: 45929319-46121434 on Build GRCh38
Transcripts
NM_001101802.1 NM_016621.3 XM_005252965.4 XM_011520157.2 XM_011520158.2 XM_011520160.2 XM_011520161.2 XM_011520162.2 XM_011520164.2 XM_011520165.2 XM_011520166.2 XM_011520167.2 XM_011520168.2 XM_011520169.2 XM_011520173.2 XM_011520174.2 XM_011520175.2 XM_011520179.2 XM_017017885.1 XM_017017886.1 XM_017017887.1 XM_017017888.1 XM_017017889.1 XM_017017890.1 XM_017017891.1 XM_017017892.1 XM_017017893.1 XM_017017894.1 XM_017017895.1 XM_017017896.1 XM_017017897.1 XM_017017898.1 XM_017017899.1 XM_017017900.1 XM_017017901.1 XM_017017902.1 XM_017017903.1 XM_017017905.1 XM_017017906.1 XM_017017907.1 XM_017017908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
927107 c.1839G>A p.T613T Chr.11: 45934034-45934034 on Build GRCh38
4032939 c.1682G>A p.R561Q Chr.11: 45934191-45934191 on Build GRCh38
6868361 c.2027G>T p.G676V Chr.11: 45933984-45933984 on Build GRCh38
4833925 c.2012C>T p.A671V Chr.11: 45933999-45933999 on Build GRCh38
4032937 c.1742G>A p.G581D Chr.11: 45934131-45934131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143570530 Chr.11: 45934300-45934300 on Build GRCh38
rs760373236 Chr.11: 45934239-45934239 on Build GRCh38
rs1033543117 Chr.11: 45934120-45934120 on Build GRCh38
rs540930082 Chr.11: 45934436-45934436 on Build GRCh38
rs892558907 Chr.11: 45934301-45934301 on Build GRCh38
rs370907044 Chr.11: 45934242-45934242 on Build GRCh38
rs774003990 Chr.11: 45934124-45934124 on Build GRCh38
rs375495711 Chr.11: 45934191-45934191 on Build GRCh38
rs149898933 Chr.11: 45933999-45933999 on Build GRCh38
rs755214673 Chr.11: 45934175-45934175 on Build GRCh38
rs567998784 Chr.11: 45934145-45934145 on Build GRCh38
rs556291726 Chr.11: 45934383-45934383 on Build GRCh38
rs184149212 Chr.11: 45934270-45934270 on Build GRCh38
rs943812963 Chr.11: 45934021-45934021 on Build GRCh38
rs374553564 Chr.11: 45934023-45934023 on Build GRCh38
rs976437955 Chr.11: 45934437-45934437 on Build GRCh38
rs151090965 Chr.11: 45934057-45934057 on Build GRCh38
rs188297957 Chr.11: 45934406-45934406 on Build GRCh38
rs748739458 Chr.11: 45934094-45934094 on Build GRCh38
rs752969599 Chr.11: 45934177-45934177 on Build GRCh38
rs866264867 Chr.11: 45934203-45934203 on Build GRCh38
rs965017857 Chr.11: 45934433-45934433 on Build GRCh38
rs759310849 Chr.11: 45934051-45934051 on Build GRCh38
rs373411765 Chr.11: 45934259-45934259 on Build GRCh38
rs761749780 Chr.11: 45934244-45934244 on Build GRCh38
rs751580003 Chr.11: 45934417-45934417 on Build GRCh38
rs149588947 Chr.11: 45934223-45934223 on Build GRCh38
rs377700825 Chr.11: 45934031-45934031 on Build GRCh38
rs756441738 Chr.11: 45934271-45934271 on Build GRCh38
rs751839230 Chr.11: 45934160-45934160 on Build GRCh38
rs144966693 Chr.11: 45934034-45934034 on Build GRCh38
rs747382214 Chr.11: 45934151-45934151 on Build GRCh38
rs145172014 Chr.11: 45934093-45934093 on Build GRCh38
rs35170501 Chr.11: 45933998-45933998 on Build GRCh38
rs752635361 Chr.11: 45934055-45934055 on Build GRCh38
rs750447716 Chr.11: 45934061-45934061 on Build GRCh38
rs76343926 Chr.11: 45934084-45934084 on Build GRCh38
rs141388105 Chr.11: 45934062-45934062 on Build GRCh38
rs758501208 Chr.11: 45934002-45934002 on Build GRCh38
rs970251628 Chr.11: 45934085-45934085 on Build GRCh38
rs750584861 Chr.11: 45934141-45934141 on Build GRCh38
rs377133610 Chr.11: 45934250-45934250 on Build GRCh38
rs776435004 Chr.11: 45934243-45934243 on Build GRCh38
rs555082497 Chr.11: 45934405-45934405 on Build GRCh38
rs767200214 Chr.11: 45934056-45934056 on Build GRCh38
rs979386319 Chr.11: 45934109-45934109 on Build GRCh38
rs767842970 Chr.11: 45934253-45934253 on Build GRCh38
rs767330804 Chr.11: 45934127-45934127 on Build GRCh38
rs755410646 Chr.11: 45934063-45934063 on Build GRCh38
rs1037022503 Chr.11: 45934350-45934350 on Build GRCh38
rs140543084 Chr.11: 45934131-45934131 on Build GRCh38
rs1063810 Chr.11: 45934029-45934029 on Build GRCh38
rs370166935 Chr.11: 45934218-45934218 on Build GRCh38
rs895856189 Chr.11: 45934354-45934354 on Build GRCh38
rs146779650 Chr.11: 45934129-45934129 on Build GRCh38
rs576086040 Chr.11: 45934384-45934384 on Build GRCh38
rs756707290 Chr.11: 45934184-45934184 on Build GRCh38
rs370627435 Chr.11: 45934143-45934143 on Build GRCh38
rs147424018 Chr.11: 45934199-45934199 on Build GRCh38
rs374586100 Chr.11: 45934144-45934144 on Build GRCh38
rs778144530 Chr.11: 45934276-45934276 on Build GRCh38
rs769053462 Chr.11: 45934163-45934163 on Build GRCh38
rs769354151 Chr.11: 45934033-45934033 on Build GRCh38
rs563961413 Chr.11: 45934045-45934045 on Build GRCh38
rs1020574413 Chr.11: 45934407-45934407 on Build GRCh38
rs1015665015 Chr.11: 45934069-45934069 on Build GRCh38
rs138242314 Chr.11: 45934201-45934201 on Build GRCh38
rs765913676 Chr.11: 45934043-45934043 on Build GRCh38
rs770443412 Chr.11: 45934194-45934194 on Build GRCh38
rs759120687 Chr.11: 45934126-45934126 on Build GRCh38
rs1000385557 Chr.11: 45934401-45934401 on Build GRCh38
rs1063809 Chr.11: 45934030-45934030 on Build GRCh38
rs148557580 Chr.11: 45934035-45934035 on Build GRCh38
rs903753167 Chr.11: 45934317-45934317 on Build GRCh38
rs146821133 Chr.11: 45934336-45934336 on Build GRCh38
rs921037346 Chr.11: 45934447-45934447 on Build GRCh38
rs763907329 Chr.11: 45934133-45934133 on Build GRCh38
rs112782705 Chr.11: 45934142-45934142 on Build GRCh38
rs1033924569 Chr.11: 45934389-45934389 on Build GRCh38
rs913597392 Chr.11: 45934202-45934202 on Build GRCh38
rs748610955 Chr.11: 45934032-45934032 on Build GRCh38
rs761575760 Chr.11: 45934055-45934056 on Build GRCh38
rs376981110 Chr.11: 45934132-45934132 on Build GRCh38
rs762102676 Chr.11: 45934213-45934213 on Build GRCh38
rs771682088 Chr.11: 45934215-45934215 on Build GRCh38
rs889181494 Chr.11: 45934046-45934046 on Build GRCh38
rs571882152 Chr.11: 45934325-45934325 on Build GRCh38
rs763090637 Chr.11: 45934358-45934358 on Build GRCh38
rs888389596 Chr.11: 45934316-45934316 on Build GRCh38
rs1009495129 Chr.11: 45934306-45934306 on Build GRCh38
rs199802844 Chr.11: 45934070-45934070 on Build GRCh38
rs373767892 Chr.11: 45934119-45934119 on Build GRCh38
rs749740019 Chr.11: 45934192-45934192 on Build GRCh38
rs75019001 Chr.11: 45934402-45934402 on Build GRCh38
rs926237474 Chr.11: 45934111-45934111 on Build GRCh38
rs191903911 Chr.11: 45934077-45934077 on Build GRCh38
rs748724067 Chr.11: 45934071-45934071 on Build GRCh38
rs766586238 Chr.11: 45934246-45934246 on Build GRCh38
rs150425973 Chr.11: 45934176-45934176 on Build GRCh38
rs759873830 Chr.11: 45934251-45934251 on Build GRCh38
rs973715337 Chr.11: 45934422-45934422 on Build GRCh38
rs1023133603 Chr.11: 45934081-45934081 on Build GRCh38
rs142948382 Chr.11: 45934054-45934054 on Build GRCh38
rs780614548 Chr.11: 45934017-45934017 on Build GRCh38

View Full Product Details