Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47242917-47243407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP2
Entrez Gene ID
53
Gene Name
acid phosphatase 2, lysosomal
Gene Aliases
LAP
UniGene
Hs.532492
Gene Chromosome Location
Chr.11: 47239302-47248847 on Build GRCh38
Transcripts
NM_001302489.1 NM_001302490.1 NM_001302491.1 NM_001302492.1 NM_001610.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1354038 c.878delT p.L293fs*89 Chr.11: 47243102-47243102 on Build GRCh38
3967535 c.807C>T p.T269T Chr.11: 47243287-47243287 on Build GRCh38
466848 c.789G>C p.Q263H Chr.11: 47243305-47243305 on Build GRCh38
158858 c.907G>A p.E303K Chr.11: 47243073-47243073 on Build GRCh38
348424 c.778C>T p.L260L Chr.11: 47243316-47243316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751170232 Chr.11: 47243081-47243081 on Build GRCh38
rs369629069 Chr.11: 47242977-47242977 on Build GRCh38
rs1027346860 Chr.11: 47243251-47243251 on Build GRCh38
rs376718590 Chr.11: 47243342-47243342 on Build GRCh38
rs757855546 Chr.11: 47242967-47242968 on Build GRCh38
rs201038164 Chr.11: 47243155-47243155 on Build GRCh38
rs34425282 Chr.11: 47243021-47243021 on Build GRCh38
rs770603142 Chr.11: 47243129-47243129 on Build GRCh38
rs776792605 Chr.11: 47243055-47243055 on Build GRCh38
rs373831783 Chr.11: 47242953-47242953 on Build GRCh38
rs188813852 Chr.11: 47243194-47243194 on Build GRCh38
rs746440989 Chr.11: 47243055-47243061 on Build GRCh38
rs376320899 Chr.11: 47243059-47243059 on Build GRCh38
rs146841506 Chr.11: 47242979-47242979 on Build GRCh38
rs374368485 Chr.11: 47243181-47243181 on Build GRCh38
rs762960271 Chr.11: 47243261-47243261 on Build GRCh38
rs542991252 Chr.11: 47243178-47243178 on Build GRCh38
rs752316501 Chr.11: 47243316-47243316 on Build GRCh38
rs755117624 Chr.11: 47243356-47243356 on Build GRCh38
rs758664326 Chr.11: 47243065-47243065 on Build GRCh38
rs565558461 Chr.11: 47243278-47243278 on Build GRCh38
rs769663317 Chr.11: 47243003-47243003 on Build GRCh38
rs776968759 Chr.11: 47243226-47243226 on Build GRCh38
rs192891764 Chr.11: 47243340-47243340 on Build GRCh38
rs1027381845 Chr.11: 47243071-47243071 on Build GRCh38
rs757897158 Chr.11: 47242969-47242969 on Build GRCh38
rs202217185 Chr.11: 47243337-47243337 on Build GRCh38
rs780439889 Chr.11: 47243103-47243103 on Build GRCh38
rs780723421 Chr.11: 47243107-47243107 on Build GRCh38
rs111618196 Chr.11: 47243324-47243324 on Build GRCh38
rs551025437 Chr.11: 47242967-47242967 on Build GRCh38
rs982431273 Chr.11: 47243371-47243371 on Build GRCh38
rs769986149 Chr.11: 47243240-47243240 on Build GRCh38
rs754721522 Chr.11: 47243083-47243083 on Build GRCh38
rs778860068 Chr.11: 47243351-47243351 on Build GRCh38
rs766802738 Chr.11: 47243067-47243067 on Build GRCh38
rs994482333 Chr.11: 47243245-47243245 on Build GRCh38
rs759897865 Chr.11: 47243279-47243279 on Build GRCh38
rs763580290 Chr.11: 47243169-47243169 on Build GRCh38
rs768855130 Chr.11: 47243146-47243146 on Build GRCh38
rs773820910 Chr.11: 47243168-47243168 on Build GRCh38
rs762697376 Chr.11: 47243017-47243017 on Build GRCh38
rs773141450 Chr.11: 47243004-47243004 on Build GRCh38
rs779574814 Chr.11: 47242975-47242975 on Build GRCh38
rs112195368 Chr.11: 47243376-47243376 on Build GRCh38
rs138306292 Chr.11: 47243272-47243272 on Build GRCh38
rs766815752 Chr.11: 47243233-47243233 on Build GRCh38
rs904524377 Chr.11: 47242973-47242973 on Build GRCh38
rs768889576 Chr.11: 47243240-47243241 on Build GRCh38
rs774260002 Chr.11: 47243136-47243136 on Build GRCh38
rs756747616 Chr.11: 47243346-47243346 on Build GRCh38
rs370819835 Chr.11: 47243204-47243204 on Build GRCh38
rs745663539 Chr.11: 47243140-47243140 on Build GRCh38
rs777139896 Chr.11: 47242984-47242984 on Build GRCh38
rs200711753 Chr.11: 47243047-47243047 on Build GRCh38
rs537427421 Chr.11: 47242956-47242956 on Build GRCh38
rs908775999 Chr.11: 47243177-47243177 on Build GRCh38
rs749209727 Chr.11: 47243214-47243214 on Build GRCh38
rs1057293362 Chr.11: 47243188-47243188 on Build GRCh38
rs761024108 Chr.11: 47243042-47243042 on Build GRCh38
rs111271272 Chr.11: 47243341-47243341 on Build GRCh38
rs761987849 Chr.11: 47243058-47243058 on Build GRCh38
rs750987173 Chr.11: 47243060-47243060 on Build GRCh38
rs777305288 Chr.11: 47243213-47243213 on Build GRCh38
rs986302772 Chr.11: 47243100-47243100 on Build GRCh38
rs748416777 Chr.11: 47243232-47243232 on Build GRCh38
rs111856045 Chr.11: 47243323-47243323 on Build GRCh38
rs201171124 Chr.11: 47243147-47243147 on Build GRCh38
rs745635185 Chr.11: 47243217-47243217 on Build GRCh38
rs113080778 Chr.11: 47242985-47242985 on Build GRCh38
rs11039145 Chr.11: 47242978-47242978 on Build GRCh38
rs140531425 Chr.11: 47242986-47242986 on Build GRCh38
rs770182549 Chr.11: 47243073-47243075 on Build GRCh38
rs952463842 Chr.11: 47243347-47243347 on Build GRCh38
rs755654381 Chr.11: 47243110-47243110 on Build GRCh38
rs35155532 Chr.11: 47242947-47242948 on Build GRCh38
rs770781950 Chr.11: 47243028-47243028 on Build GRCh38
rs754295344 Chr.11: 47242968-47242968 on Build GRCh38
rs755601392 Chr.11: 47243212-47243212 on Build GRCh38
rs755305861 Chr.11: 47243196-47243196 on Build GRCh38
rs773487765 Chr.11: 47243246-47243246 on Build GRCh38
rs367729654 Chr.11: 47243353-47243353 on Build GRCh38
rs764066850 Chr.11: 47243051-47243051 on Build GRCh38
rs775554240 Chr.11: 47243035-47243035 on Build GRCh38
rs111841808 Chr.11: 47243128-47243128 on Build GRCh38
rs938945319 Chr.11: 47243184-47243184 on Build GRCh38
rs908243259 Chr.11: 47243383-47243383 on Build GRCh38
rs963819716 Chr.11: 47243031-47243031 on Build GRCh38
rs747593925 Chr.11: 47243109-47243109 on Build GRCh38
rs760622622 Chr.11: 47242946-47242946 on Build GRCh38
rs777639992 Chr.11: 47243112-47243112 on Build GRCh38
rs200232951 Chr.11: 47243216-47243216 on Build GRCh38
rs528645202 Chr.11: 47243219-47243219 on Build GRCh38
rs757428208 Chr.11: 47243269-47243269 on Build GRCh38
rs975572308 Chr.11: 47243046-47243046 on Build GRCh38
rs953261589 Chr.11: 47243078-47243078 on Build GRCh38
rs754023477 Chr.11: 47243227-47243227 on Build GRCh38
rs765455336 Chr.11: 47243159-47243159 on Build GRCh38
rs140923968 Chr.11: 47243286-47243286 on Build GRCh38
rs768527012 Chr.11: 47243324-47243326 on Build GRCh38
rs542312797 Chr.11: 47243206-47243206 on Build GRCh38
rs749615901 Chr.11: 47243208-47243209 on Build GRCh38
rs867791378 Chr.11: 47243375-47243375 on Build GRCh38

View Full Product Details