Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 58624716-58625230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTF
Entrez Gene ID
1270
Gene Name
ciliary neurotrophic factor
Gene Aliases
HCNTF
UniGene
Hs.524920
Gene Chromosome Location
Chr.11: 58622673-58625733 on Build GRCh38
Transcripts
NM_000614.3 NR_024091.1

Gene Symbol
ZFP91-CNTF
Entrez Gene ID
386607
Gene Name
ZFP91-CNTF readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.524920
Gene Chromosome Location
Chr.11: 58579114-58625733 on Build GRCh38
Transcripts
NM_000614.3 NR_024091.1

SNP Detail

SNP ID Chromosome Location
rs775706851 Chr.11: 58624800-58624800 on Build GRCh38
rs867280335 Chr.11: 58624843-58624843 on Build GRCh38
rs1022964385 Chr.11: 58624997-58624997 on Build GRCh38
rs981476885 Chr.11: 58625053-58625053 on Build GRCh38
rs991862502 Chr.11: 58624975-58624975 on Build GRCh38
rs888822804 Chr.11: 58624902-58624902 on Build GRCh38
rs1040125347 Chr.11: 58625142-58625142 on Build GRCh38
rs7113847 Chr.11: 58624950-58624950 on Build GRCh38
rs190890301 Chr.11: 58625078-58625078 on Build GRCh38
rs547808853 Chr.11: 58625067-58625067 on Build GRCh38
rs61889535 Chr.11: 58624849-58624849 on Build GRCh38
rs112283734 Chr.11: 58624887-58624887 on Build GRCh38
rs964936988 Chr.11: 58624848-58624848 on Build GRCh38
rs915847015 Chr.11: 58624930-58624930 on Build GRCh38
rs1015761004 Chr.11: 58625193-58625193 on Build GRCh38
rs952340122 Chr.11: 58624973-58624973 on Build GRCh38
rs553738472 Chr.11: 58625179-58625179 on Build GRCh38
rs183568841 Chr.11: 58624918-58624918 on Build GRCh38
rs907219551 Chr.11: 58625005-58625005 on Build GRCh38
rs1031735313 Chr.11: 58624793-58624793 on Build GRCh38
rs534145800 Chr.11: 58625155-58625155 on Build GRCh38
rs183097999 Chr.11: 58625081-58625081 on Build GRCh38
rs1029364509 Chr.11: 58624850-58624850 on Build GRCh38
rs761435120 Chr.11: 58625061-58625061 on Build GRCh38
rs545924377 Chr.11: 58625102-58625102 on Build GRCh38
rs145693678 Chr.11: 58624988-58624988 on Build GRCh38
rs544460185 Chr.11: 58625075-58625075 on Build GRCh38
rs1053622872 Chr.11: 58625065-58625065 on Build GRCh38
rs973837184 Chr.11: 58624929-58624929 on Build GRCh38
rs141722669 Chr.11: 58624907-58624907 on Build GRCh38
rs764553350 Chr.11: 58624913-58624913 on Build GRCh38
rs555297295 Chr.11: 58625052-58625052 on Build GRCh38
rs993633573 Chr.11: 58625159-58625160 on Build GRCh38
rs568660082 Chr.11: 58624757-58624757 on Build GRCh38
rs908278485 Chr.11: 58624914-58624914 on Build GRCh38
rs187924262 Chr.11: 58625072-58625072 on Build GRCh38
rs557865192 Chr.11: 58624928-58624928 on Build GRCh38
rs1011714667 Chr.11: 58624803-58624803 on Build GRCh38
rs751001871 Chr.11: 58624915-58624915 on Build GRCh38
rs950166963 Chr.11: 58624782-58624783 on Build GRCh38
rs897592338 Chr.11: 58625145-58625145 on Build GRCh38
rs887984196 Chr.11: 58625180-58625180 on Build GRCh38
rs1030586365 Chr.11: 58625124-58625124 on Build GRCh38
rs201782463 Chr.11: 58624912-58624912 on Build GRCh38
rs190999960 Chr.11: 58624741-58624741 on Build GRCh38
rs999371496 Chr.11: 58624783-58624783 on Build GRCh38
rs934673412 Chr.11: 58625046-58625046 on Build GRCh38
rs564229572 Chr.11: 58625135-58625135 on Build GRCh38
rs767030073 Chr.11: 58625107-58625107 on Build GRCh38
rs982434766 Chr.11: 58625159-58625159 on Build GRCh38
rs944378829 Chr.11: 58625122-58625122 on Build GRCh38
rs955248777 Chr.11: 58625148-58625148 on Build GRCh38
rs1017758459 Chr.11: 58625080-58625080 on Build GRCh38
rs1017874518 Chr.11: 58624816-58624816 on Build GRCh38
rs1023665638 Chr.11: 58625204-58625204 on Build GRCh38
rs1050811818 Chr.11: 58625113-58625113 on Build GRCh38
rs997791388 Chr.11: 58625121-58625121 on Build GRCh38
rs907309124 Chr.11: 58624758-58624758 on Build GRCh38
rs567700940 Chr.11: 58625081-58625082 on Build GRCh38

View Full Product Details