Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60900493-60900985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPF19
Entrez Gene ID
27339
Gene Name
pre-mRNA processing factor 19
Gene Aliases
NMP200, PRP19, PSO4, SNEV, UBOX4, hPSO4
UniGene
Hs.502705
Gene Chromosome Location
Chr.11: 60890547-60906589 on Build GRCh38
Transcripts
NM_014502.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6756540 c.704A>C p.N235T Chr.11: 60900868-60900868 on Build GRCh38
6756548 c.693G>A p.P231P Chr.11: 60900879-60900879 on Build GRCh38
929320 c.787C>A p.H263N Chr.11: 60900623-60900623 on Build GRCh38
4974889 c.813G>T p.V271V Chr.11: 60900597-60900597 on Build GRCh38
929322 c.725C>T p.A242V Chr.11: 60900685-60900685 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764360216 Chr.11: 60900547-60900549 on Build GRCh38
rs764428172 Chr.11: 60900672-60900672 on Build GRCh38
rs919781715 Chr.11: 60900545-60900545 on Build GRCh38
rs148553971 Chr.11: 60900766-60900766 on Build GRCh38
rs752455658 Chr.11: 60900880-60900880 on Build GRCh38
rs758308868 Chr.11: 60900713-60900713 on Build GRCh38
rs972953309 Chr.11: 60900518-60900518 on Build GRCh38
rs368154972 Chr.11: 60900865-60900865 on Build GRCh38
rs756723140 Chr.11: 60900940-60900940 on Build GRCh38
rs377204037 Chr.11: 60900830-60900830 on Build GRCh38
rs551923225 Chr.11: 60900784-60900784 on Build GRCh38
rs904023865 Chr.11: 60900684-60900684 on Build GRCh38
rs777017929 Chr.11: 60900841-60900841 on Build GRCh38
rs763076277 Chr.11: 60900861-60900861 on Build GRCh38
rs759269639 Chr.11: 60900876-60900876 on Build GRCh38
rs1001021040 Chr.11: 60900950-60900950 on Build GRCh38
rs990341293 Chr.11: 60900619-60900619 on Build GRCh38
rs767347492 Chr.11: 60900879-60900879 on Build GRCh38
rs762178076 Chr.11: 60900845-60900845 on Build GRCh38
rs1008624103 Chr.11: 60900728-60900728 on Build GRCh38
rs370963167 Chr.11: 60900812-60900812 on Build GRCh38
rs200956536 Chr.11: 60900819-60900819 on Build GRCh38
rs1028571586 Chr.11: 60900715-60900715 on Build GRCh38
rs560231981 Chr.11: 60900569-60900569 on Build GRCh38
rs145883057 Chr.11: 60900765-60900765 on Build GRCh38
rs914686397 Chr.11: 60900635-60900635 on Build GRCh38
rs755828527 Chr.11: 60900927-60900927 on Build GRCh38
rs370679425 Chr.11: 60900956-60900956 on Build GRCh38
rs772598122 Chr.11: 60900591-60900591 on Build GRCh38
rs938337000 Chr.11: 60900575-60900575 on Build GRCh38
rs865974567 Chr.11: 60900620-60900620 on Build GRCh38
rs374417404 Chr.11: 60900685-60900685 on Build GRCh38
rs769105836 Chr.11: 60900837-60900837 on Build GRCh38
rs763891609 Chr.11: 60900666-60900666 on Build GRCh38
rs377469354 Chr.11: 60900718-60900718 on Build GRCh38
rs891516684 Chr.11: 60900584-60900584 on Build GRCh38
rs747503749 Chr.11: 60900960-60900960 on Build GRCh38
rs761083429 Chr.11: 60900671-60900671 on Build GRCh38
rs201654787 Chr.11: 60900832-60900832 on Build GRCh38
rs17155466 Chr.11: 60900561-60900561 on Build GRCh38
rs1026856328 Chr.11: 60900729-60900729 on Build GRCh38
rs180951671 Chr.11: 60900517-60900517 on Build GRCh38
rs781720049 Chr.11: 60900962-60900962 on Build GRCh38
rs549140900 Chr.11: 60900737-60900737 on Build GRCh38
rs904307839 Chr.11: 60900944-60900944 on Build GRCh38
rs1001072871 Chr.11: 60900694-60900694 on Build GRCh38
rs773232408 Chr.11: 60900855-60900855 on Build GRCh38
rs897089652 Chr.11: 60900719-60900719 on Build GRCh38
rs375416457 Chr.11: 60900566-60900566 on Build GRCh38
rs751805488 Chr.11: 60900713-60900716 on Build GRCh38
rs970426260 Chr.11: 60900771-60900771 on Build GRCh38
rs529175846 Chr.11: 60900696-60900696 on Build GRCh38
rs370112740 Chr.11: 60900603-60900603 on Build GRCh38
rs754735706 Chr.11: 60900734-60900734 on Build GRCh38
rs894962628 Chr.11: 60900745-60900745 on Build GRCh38
rs185619786 Chr.11: 60900961-60900961 on Build GRCh38
rs750510067 Chr.11: 60900711-60900711 on Build GRCh38
rs1019629536 Chr.11: 60900736-60900736 on Build GRCh38
rs765361944 Chr.11: 60900703-60900703 on Build GRCh38
rs970352585 Chr.11: 60900524-60900524 on Build GRCh38
rs981681832 Chr.11: 60900526-60900526 on Build GRCh38
rs765636008 Chr.11: 60900875-60900875 on Build GRCh38
rs868116322 Chr.11: 60900655-60900655 on Build GRCh38
rs372516464 Chr.11: 60900565-60900565 on Build GRCh38
rs753298628 Chr.11: 60900938-60900938 on Build GRCh38
rs920050648 Chr.11: 60900797-60900797 on Build GRCh38
rs747342987 Chr.11: 60900833-60900833 on Build GRCh38
rs375491765 Chr.11: 60900816-60900816 on Build GRCh38
rs1056823673 Chr.11: 60900576-60900576 on Build GRCh38
rs868158781 Chr.11: 60900650-60900650 on Build GRCh38
rs766336538 Chr.11: 60900717-60900717 on Build GRCh38
rs144450563 Chr.11: 60900798-60900798 on Build GRCh38
rs770111586 Chr.11: 60900847-60900847 on Build GRCh38

View Full Product Details