Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63597977-63598430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G16
Entrez Gene ID
11145
Gene Name
phospholipase A2 group XVI
Gene Aliases
AdPLA, H-REV107-1, HRASLS3, HREV107, HREV107-1, HREV107-3, HRSL3
UniGene
Hs.502775
Gene Chromosome Location
Chr.11: 63574462-63616926 on Build GRCh38
Transcripts
NM_001128203.1 NM_007069.3 XM_006718426.1 XM_011544741.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4543016 c.32G>A p.G11E Chr.11: 63598147-63598147 on Build GRCh38
4562515 c.92G>A p.G31E Chr.11: 63598087-63598087 on Build GRCh38
2162537 c.54C>T p.R18R Chr.11: 63598125-63598125 on Build GRCh38
3451302 c.29C>T p.P10L Chr.11: 63598150-63598150 on Build GRCh38
4930103 c.93A>G p.G31G Chr.11: 63598086-63598086 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372625520 Chr.11: 63598117-63598117 on Build GRCh38
rs924434393 Chr.11: 63598069-63598069 on Build GRCh38
rs751283003 Chr.11: 63598197-63598197 on Build GRCh38
rs750267636 Chr.11: 63598205-63598205 on Build GRCh38
rs369064079 Chr.11: 63598109-63598109 on Build GRCh38
rs769405682 Chr.11: 63598097-63598097 on Build GRCh38
rs568317054 Chr.11: 63598346-63598346 on Build GRCh38
rs1049915039 Chr.11: 63598183-63598183 on Build GRCh38
rs551883033 Chr.11: 63598186-63598186 on Build GRCh38
rs952022749 Chr.11: 63598350-63598350 on Build GRCh38
rs775365894 Chr.11: 63598106-63598106 on Build GRCh38
rs753136861 Chr.11: 63598006-63598006 on Build GRCh38
rs192601907 Chr.11: 63598055-63598055 on Build GRCh38
rs543397953 Chr.11: 63598049-63598049 on Build GRCh38
rs756394625 Chr.11: 63598026-63598026 on Build GRCh38
rs112530400 Chr.11: 63598164-63598164 on Build GRCh38
rs970265236 Chr.11: 63598066-63598066 on Build GRCh38
rs985238919 Chr.11: 63598050-63598050 on Build GRCh38
rs1041316126 Chr.11: 63598304-63598304 on Build GRCh38
rs887261345 Chr.11: 63598254-63598254 on Build GRCh38
rs1026273915 Chr.11: 63598338-63598338 on Build GRCh38
rs1004443119 Chr.11: 63598261-63598261 on Build GRCh38
rs955972742 Chr.11: 63598075-63598075 on Build GRCh38
rs372985043 Chr.11: 63598199-63598199 on Build GRCh38
rs1008163907 Chr.11: 63598275-63598275 on Build GRCh38
rs575235692 Chr.11: 63598028-63598028 on Build GRCh38
rs758085456 Chr.11: 63598064-63598064 on Build GRCh38
rs993675510 Chr.11: 63598328-63598328 on Build GRCh38
rs996529477 Chr.11: 63598318-63598318 on Build GRCh38
rs766854304 Chr.11: 63598143-63598143 on Build GRCh38
rs1052780406 Chr.11: 63598105-63598105 on Build GRCh38
rs377349187 Chr.11: 63598188-63598188 on Build GRCh38
rs981283061 Chr.11: 63598084-63598084 on Build GRCh38
rs867241335 Chr.11: 63598326-63598326 on Build GRCh38
rs372138391 Chr.11: 63598070-63598070 on Build GRCh38
rs901391511 Chr.11: 63598314-63598314 on Build GRCh38
rs145543963 Chr.11: 63598206-63598206 on Build GRCh38
rs369971730 Chr.11: 63598182-63598182 on Build GRCh38
rs548098641 Chr.11: 63598344-63598344 on Build GRCh38
rs761857748 Chr.11: 63598121-63598121 on Build GRCh38
rs1040544049 Chr.11: 63598310-63598310 on Build GRCh38
rs780758240 Chr.11: 63598386-63598386 on Build GRCh38
rs763820840 Chr.11: 63598184-63598184 on Build GRCh38
rs150138244 Chr.11: 63598010-63598010 on Build GRCh38
rs752341771 Chr.11: 63598046-63598046 on Build GRCh38
rs746491288 Chr.11: 63598089-63598089 on Build GRCh38
rs749138204 Chr.11: 63598107-63598107 on Build GRCh38
rs768858995 Chr.11: 63598113-63598113 on Build GRCh38
rs369058482 Chr.11: 63598092-63598092 on Build GRCh38
rs532186243 Chr.11: 63598126-63598126 on Build GRCh38
rs917131014 Chr.11: 63598099-63598099 on Build GRCh38
rs750986828 Chr.11: 63598023-63598023 on Build GRCh38
rs776545687 Chr.11: 63598379-63598379 on Build GRCh38
rs765867760 Chr.11: 63598028-63598029 on Build GRCh38
rs761259464 Chr.11: 63598127-63598127 on Build GRCh38
rs201688333 Chr.11: 63598091-63598091 on Build GRCh38

View Full Product Details