Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64790099-64790560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP4K2
Entrez Gene ID
5871
Gene Name
mitogen-activated protein kinase kinase kinase kinase 2
Gene Aliases
BL44, GCK, RAB8IP
UniGene
Hs.534341
Gene Chromosome Location
Chr.11: 64786914-64803254 on Build GRCh38
Transcripts
NM_001307990.1 NM_004579.4 XM_011545203.2 XM_011545204.2 XM_017018093.1 XM_017018094.1 XM_017018095.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
930201 c.2210C>T p.A737V Chr.11: 64790226-64790226 on Build GRCh38
3398025 c.2238C>T p.I746I Chr.11: 64790198-64790198 on Build GRCh38
4387603 c.2162-8C>T p.? Chr.11: 64790282-64790282 on Build GRCh38
415463 c.2239G>C p.E747Q Chr.11: 64790197-64790197 on Build GRCh38
5570325 c.2198C>T p.T733M Chr.11: 64790238-64790238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747649893 Chr.11: 64790153-64790153 on Build GRCh38
rs925579096 Chr.11: 64790342-64790342 on Build GRCh38
rs368600929 Chr.11: 64790130-64790130 on Build GRCh38
rs750957955 Chr.11: 64790384-64790384 on Build GRCh38
rs200063305 Chr.11: 64790273-64790273 on Build GRCh38
rs965208178 Chr.11: 64790299-64790299 on Build GRCh38
rs1045210408 Chr.11: 64790402-64790402 on Build GRCh38
rs949530556 Chr.11: 64790371-64790371 on Build GRCh38
rs771053940 Chr.11: 64790301-64790301 on Build GRCh38
rs746465394 Chr.11: 64790492-64790493 on Build GRCh38
rs376881891 Chr.11: 64790139-64790139 on Build GRCh38
rs151089831 Chr.11: 64790394-64790394 on Build GRCh38
rs543617419 Chr.11: 64790126-64790126 on Build GRCh38
rs377145378 Chr.11: 64790536-64790546 on Build GRCh38
rs376417763 Chr.11: 64790526-64790526 on Build GRCh38
rs777148489 Chr.11: 64790476-64790477 on Build GRCh38
rs776635800 Chr.11: 64790182-64790182 on Build GRCh38
rs770925455 Chr.11: 64790181-64790181 on Build GRCh38
rs79516942 Chr.11: 64790263-64790263 on Build GRCh38
rs777341499 Chr.11: 64790286-64790286 on Build GRCh38
rs114846214 Chr.11: 64790333-64790333 on Build GRCh38
rs200441498 Chr.11: 64790225-64790225 on Build GRCh38
rs1004545269 Chr.11: 64790529-64790529 on Build GRCh38
rs71581723 Chr.11: 64790532-64790542 on Build GRCh38
rs751568547 Chr.11: 64790237-64790237 on Build GRCh38
rs745699530 Chr.11: 64790306-64790306 on Build GRCh38
rs112459115 Chr.11: 64790295-64790295 on Build GRCh38
rs1040928778 Chr.11: 64790536-64790536 on Build GRCh38
rs1026416965 Chr.11: 64790177-64790177 on Build GRCh38
rs768353673 Chr.11: 64790140-64790140 on Build GRCh38
rs768410984 Chr.11: 64790540-64790540 on Build GRCh38
rs145194053 Chr.11: 64790531-64790541 on Build GRCh38
rs143967615 Chr.11: 64790230-64790230 on Build GRCh38
rs746570975 Chr.11: 64790417-64790417 on Build GRCh38
rs775752364 Chr.11: 64790218-64790218 on Build GRCh38
rs750835374 Chr.11: 64790246-64790246 on Build GRCh38
rs1001834841 Chr.11: 64790355-64790355 on Build GRCh38
rs756800455 Chr.11: 64790431-64790431 on Build GRCh38
rs749015852 Chr.11: 64790491-64790491 on Build GRCh38
rs372923150 Chr.11: 64790291-64790291 on Build GRCh38
rs768052793 Chr.11: 64790362-64790362 on Build GRCh38
rs906232260 Chr.11: 64790341-64790341 on Build GRCh38
rs764214259 Chr.11: 64790348-64790348 on Build GRCh38
rs994631220 Chr.11: 64790167-64790167 on Build GRCh38
rs202021353 Chr.11: 64790245-64790245 on Build GRCh38
rs926682182 Chr.11: 64790447-64790447 on Build GRCh38
rs568345409 Chr.11: 64790527-64790527 on Build GRCh38
rs761857818 Chr.11: 64790358-64790358 on Build GRCh38
rs139845154 Chr.11: 64790254-64790254 on Build GRCh38
rs369251587 Chr.11: 64790356-64790356 on Build GRCh38
rs372406359 Chr.11: 64790343-64790343 on Build GRCh38
rs148398112 Chr.11: 64790197-64790197 on Build GRCh38
rs371578192 Chr.11: 64790366-64790366 on Build GRCh38
rs1028025801 Chr.11: 64790316-64790316 on Build GRCh38
rs775086112 Chr.11: 64790344-64790344 on Build GRCh38
rs775417235 Chr.11: 64790514-64790514 on Build GRCh38
rs913376927 Chr.11: 64790293-64790293 on Build GRCh38
rs781537229 Chr.11: 64790138-64790138 on Build GRCh38
rs746314836 Chr.11: 64790198-64790198 on Build GRCh38
rs780240623 Chr.11: 64790262-64790262 on Build GRCh38
rs780812507 Chr.11: 64790446-64790446 on Build GRCh38
rs376776439 Chr.11: 64790345-64790345 on Build GRCh38
rs370193525 Chr.11: 64790161-64790161 on Build GRCh38
rs771642278 Chr.11: 64790158-64790158 on Build GRCh38
rs769549085 Chr.11: 64790470-64790470 on Build GRCh38
rs949510507 Chr.11: 64790200-64790200 on Build GRCh38
rs758137477 Chr.11: 64790409-64790409 on Build GRCh38
rs978383986 Chr.11: 64790337-64790337 on Build GRCh38
rs766782005 Chr.11: 64790405-64790405 on Build GRCh38
rs887009307 Chr.11: 64790257-64790257 on Build GRCh38
rs760704240 Chr.11: 64790163-64790163 on Build GRCh38
rs144446553 Chr.11: 64790271-64790271 on Build GRCh38
rs374254798 Chr.11: 64790294-64790294 on Build GRCh38
rs201019245 Chr.11: 64790458-64790458 on Build GRCh38
rs138274379 Chr.11: 64790305-64790305 on Build GRCh38
rs375927430 Chr.11: 64790408-64790408 on Build GRCh38
rs918084538 Chr.11: 64790370-64790370 on Build GRCh38
rs920937961 Chr.11: 64790118-64790118 on Build GRCh38
rs777555573 Chr.11: 64790411-64790411 on Build GRCh38
rs756467759 Chr.11: 64790250-64790250 on Build GRCh38
rs758029001 Chr.11: 64790295-64790295 on Build GRCh38
rs145971059 Chr.11: 64790238-64790238 on Build GRCh38
rs778604824 Chr.11: 64790146-64790146 on Build GRCh38

View Full Product Details