Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53046868-53047418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC283335
Entrez Gene ID
283335
Gene Name
uncharacterized LOC283335
Gene Aliases
-
UniGene
Hs.657060
Gene Chromosome Location
Chr.12: 53043189-53054438 on Build GRCh38
Transcripts
NR_033854.1 NM_198316.1 XM_011538079.1 XM_017019088.1 XM_017019089.1

Gene Symbol
TNS2
Entrez Gene ID
23371
Gene Name
tensin 2
Gene Aliases
C1-TEN, C1TEN, TENC1
UniGene
Hs.343334
Gene Chromosome Location
Chr.12: 53046980-53064379 on Build GRCh38
Transcripts
NR_033854.1 NM_198316.1 XM_011538079.1 XM_017019088.1 XM_017019089.1

SNP Detail

SNP ID Chromosome Location
rs200862018 Chr.12: 53047149-53047150 on Build GRCh38
rs1038600480 Chr.12: 53047295-53047295 on Build GRCh38
rs76359458 Chr.12: 53046914-53046914 on Build GRCh38
rs555845852 Chr.12: 53047067-53047068 on Build GRCh38
rs976150589 Chr.12: 53047206-53047206 on Build GRCh38
rs906975013 Chr.12: 53047012-53047012 on Build GRCh38
rs141050814 Chr.12: 53047176-53047181 on Build GRCh38
rs1011781763 Chr.12: 53047385-53047385 on Build GRCh38
rs941170502 Chr.12: 53046968-53046968 on Build GRCh38
rs1052859275 Chr.12: 53047360-53047360 on Build GRCh38
rs957545118 Chr.12: 53047032-53047032 on Build GRCh38
rs1016777402 Chr.12: 53046922-53046922 on Build GRCh38
rs998497639 Chr.12: 53047349-53047349 on Build GRCh38
rs1002404532 Chr.12: 53047335-53047335 on Build GRCh38
rs1046905967 Chr.12: 53046925-53046925 on Build GRCh38
rs983514622 Chr.12: 53047280-53047281 on Build GRCh38
rs557779732 Chr.12: 53046999-53047000 on Build GRCh38
rs906880120 Chr.12: 53047320-53047320 on Build GRCh38
rs868127674 Chr.12: 53047321-53047321 on Build GRCh38
rs73099903 Chr.12: 53046995-53046995 on Build GRCh38
rs950833099 Chr.12: 53047258-53047258 on Build GRCh38
rs547904617 Chr.12: 53047020-53047020 on Build GRCh38
rs544823724 Chr.12: 53047111-53047112 on Build GRCh38
rs74891301 Chr.12: 53046903-53046903 on Build GRCh38
rs1038291186 Chr.12: 53046981-53046981 on Build GRCh38
rs948625611 Chr.12: 53047310-53047310 on Build GRCh38
rs946972925 Chr.12: 53047039-53047039 on Build GRCh38
rs927687108 Chr.12: 53046990-53046990 on Build GRCh38
rs990906365 Chr.12: 53047004-53047004 on Build GRCh38
rs1045430542 Chr.12: 53047313-53047313 on Build GRCh38
rs972508270 Chr.12: 53046955-53046955 on Build GRCh38
rs530603809 Chr.12: 53047092-53047092 on Build GRCh38
rs368930903 Chr.12: 53047117-53047117 on Build GRCh38
rs976455192 Chr.12: 53046906-53046906 on Build GRCh38
rs915867229 Chr.12: 53047297-53047297 on Build GRCh38
rs954110110 Chr.12: 53046912-53046912 on Build GRCh38
rs373447124 Chr.12: 53047145-53047145 on Build GRCh38
rs776192869 Chr.12: 53046977-53046977 on Build GRCh38
rs961194188 Chr.12: 53046938-53046938 on Build GRCh38
rs552350299 Chr.12: 53047144-53047144 on Build GRCh38
rs892858885 Chr.12: 53047364-53047364 on Build GRCh38
rs923248582 Chr.12: 53047233-53047233 on Build GRCh38
rs888007118 Chr.12: 53047323-53047323 on Build GRCh38
rs941204569 Chr.12: 53047294-53047294 on Build GRCh38
rs867884528 Chr.12: 53047286-53047286 on Build GRCh38
rs10783559 Chr.12: 53046890-53046890 on Build GRCh38

View Full Product Details