Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38895207-38895720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R16B
Entrez Gene ID
26051
Gene Name
protein phosphatase 1 regulatory subunit 16B
Gene Aliases
ANKRD4, TIMAP
UniGene
Hs.45719
Gene Chromosome Location
Chr.20: 38805693-38923024 on Build GRCh38
Transcripts
NM_001172735.2 NM_015568.3 XM_011528768.2 XM_011528769.2 XM_017027785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3963558 c.322-1G>T p.? Chr.20: 38895564-38895564 on Build GRCh38
1713461 c.325T>G p.C109G Chr.20: 38895568-38895568 on Build GRCh38
3546248 c.343G>A p.E115K Chr.20: 38895586-38895586 on Build GRCh38
5857846 c.356T>A p.L119Q Chr.20: 38895599-38895599 on Build GRCh38
5004242 c.374C>T p.A125V Chr.20: 38895617-38895617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756483227 Chr.20: 38895573-38895573 on Build GRCh38
rs753128677 Chr.20: 38895567-38895567 on Build GRCh38
rs1032837182 Chr.20: 38895261-38895261 on Build GRCh38
rs1042584208 Chr.20: 38895414-38895414 on Build GRCh38
rs369792119 Chr.20: 38895604-38895604 on Build GRCh38
rs1049846774 Chr.20: 38895491-38895491 on Build GRCh38
rs920123945 Chr.20: 38895242-38895242 on Build GRCh38
rs771658390 Chr.20: 38895602-38895602 on Build GRCh38
rs550711807 Chr.20: 38895600-38895600 on Build GRCh38
rs376584312 Chr.20: 38895362-38895362 on Build GRCh38
rs778459886 Chr.20: 38895581-38895581 on Build GRCh38
rs998927542 Chr.20: 38895511-38895511 on Build GRCh38
rs917806300 Chr.20: 38895405-38895405 on Build GRCh38
rs550776658 Chr.20: 38895484-38895484 on Build GRCh38
rs774283618 Chr.20: 38895675-38895675 on Build GRCh38
rs530907128 Chr.20: 38895699-38895699 on Build GRCh38
rs998727998 Chr.20: 38895254-38895254 on Build GRCh38
rs752930397 Chr.20: 38895688-38895688 on Build GRCh38
rs148704863 Chr.20: 38895603-38895603 on Build GRCh38
rs200761547 Chr.20: 38895515-38895515 on Build GRCh38
rs997185400 Chr.20: 38895294-38895294 on Build GRCh38
rs771589707 Chr.20: 38895547-38895547 on Build GRCh38
rs75333928 Chr.20: 38895513-38895513 on Build GRCh38
rs1056170361 Chr.20: 38895401-38895401 on Build GRCh38
rs763051410 Chr.20: 38895523-38895523 on Build GRCh38
rs111456997 Chr.20: 38895465-38895465 on Build GRCh38
rs1008642071 Chr.20: 38895510-38895510 on Build GRCh38
rs773746899 Chr.20: 38895338-38895338 on Build GRCh38
rs374089812 Chr.20: 38895531-38895531 on Build GRCh38
rs117084948 Chr.20: 38895527-38895527 on Build GRCh38
rs781664009 Chr.20: 38895545-38895545 on Build GRCh38
rs77208150 Chr.20: 38895431-38895431 on Build GRCh38
rs769537525 Chr.20: 38895628-38895628 on Build GRCh38
rs748285027 Chr.20: 38895306-38895306 on Build GRCh38
rs762846601 Chr.20: 38895645-38895645 on Build GRCh38
rs16987662 Chr.20: 38895408-38895408 on Build GRCh38
rs972236046 Chr.20: 38895360-38895360 on Build GRCh38
rs765172273 Chr.20: 38895514-38895514 on Build GRCh38
rs773155575 Chr.20: 38895639-38895639 on Build GRCh38
rs368407791 Chr.20: 38895322-38895322 on Build GRCh38
rs140536087 Chr.20: 38895345-38895345 on Build GRCh38
rs1000911730 Chr.20: 38895485-38895485 on Build GRCh38
rs868634949 Chr.20: 38895310-38895310 on Build GRCh38
rs766319395 Chr.20: 38895525-38895525 on Build GRCh38
rs555711457 Chr.20: 38895400-38895400 on Build GRCh38
rs199729917 Chr.20: 38895687-38895687 on Build GRCh38
rs759604187 Chr.20: 38895676-38895676 on Build GRCh38
rs147809049 Chr.20: 38895535-38895535 on Build GRCh38
rs930802537 Chr.20: 38895366-38895366 on Build GRCh38
rs538879073 Chr.20: 38895299-38895300 on Build GRCh38
rs532102094 Chr.20: 38895289-38895289 on Build GRCh38
rs903227941 Chr.20: 38895238-38895239 on Build GRCh38
rs1028214072 Chr.20: 38895304-38895305 on Build GRCh38
rs1045857911 Chr.20: 38895237-38895237 on Build GRCh38
rs759942653 Chr.20: 38895486-38895486 on Build GRCh38
rs776206651 Chr.20: 38895621-38895621 on Build GRCh38
rs35037544 Chr.20: 38895627-38895627 on Build GRCh38
rs764859836 Chr.20: 38895501-38895501 on Build GRCh38
rs766547256 Chr.20: 38895663-38895663 on Build GRCh38
rs6028183 Chr.20: 38895365-38895365 on Build GRCh38

View Full Product Details