Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122912537-122913116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDT6
Entrez Gene ID
11162
Gene Name
nudix hydrolase 6
Gene Aliases
ASFGF2, FGF-AS, FGF2AS, GFG-1, GFG1
UniGene
Hs.558459
Gene Chromosome Location
Chr.4: 122892644-122923004 on Build GRCh38
Transcripts
NM_007083.4 NM_198041.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1637444 c.498A>T p.K166N Chr.4: 122912568-122912568 on Build GRCh38
5865675 c.478G>A p.V160I Chr.4: 122912588-122912588 on Build GRCh38
1050743 c.470A>G p.K157R Chr.4: 122912596-122912596 on Build GRCh38
5508040 c.459A>T p.E153D Chr.4: 122912607-122912607 on Build GRCh38
3428109 c.496A>G p.K166E Chr.4: 122912570-122912570 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1006597711 Chr.4: 122912981-122912981 on Build GRCh38
rs755315157 Chr.4: 122912688-122912688 on Build GRCh38
rs1008512291 Chr.4: 122912700-122912700 on Build GRCh38
rs1026259000 Chr.4: 122912626-122912626 on Build GRCh38
rs988133733 Chr.4: 122912911-122912911 on Build GRCh38
rs996037272 Chr.4: 122912616-122912616 on Build GRCh38
rs561668666 Chr.4: 122912631-122912631 on Build GRCh38
rs374978429 Chr.4: 122912645-122912645 on Build GRCh38
rs112984678 Chr.4: 122912887-122912887 on Build GRCh38
rs761748125 Chr.4: 122912590-122912590 on Build GRCh38
rs56211136 Chr.4: 122912956-122912956 on Build GRCh38
rs1016308823 Chr.4: 122912990-122912990 on Build GRCh38
rs990986036 Chr.4: 122912731-122912731 on Build GRCh38
rs949769237 Chr.4: 122912765-122912765 on Build GRCh38
rs201613726 Chr.4: 122912653-122912653 on Build GRCh38
rs539520210 Chr.4: 122912753-122912753 on Build GRCh38
rs1016895868 Chr.4: 122912697-122912697 on Build GRCh38
rs776736494 Chr.4: 122912589-122912589 on Build GRCh38
rs763387961 Chr.4: 122912636-122912636 on Build GRCh38
rs959884344 Chr.4: 122913089-122913089 on Build GRCh38
rs984824443 Chr.4: 122912682-122912682 on Build GRCh38
rs201932134 Chr.4: 122912823-122912823 on Build GRCh38
rs768832119 Chr.4: 122912575-122912575 on Build GRCh38
rs1007770021 Chr.4: 122912576-122912576 on Build GRCh38
rs890400291 Chr.4: 122912693-122912693 on Build GRCh38
rs748035982 Chr.4: 122912822-122912823 on Build GRCh38
rs1038833280 Chr.4: 122912917-122912917 on Build GRCh38
rs901711012 Chr.4: 122912943-122912943 on Build GRCh38
rs570591384 Chr.4: 122912706-122912706 on Build GRCh38
rs187979958 Chr.4: 122912972-122912972 on Build GRCh38
rs779688565 Chr.4: 122912986-122912986 on Build GRCh38
rs566225812 Chr.4: 122912820-122912820 on Build GRCh38
rs1044171638 Chr.4: 122912977-122912977 on Build GRCh38
rs548946311 Chr.4: 122912863-122912863 on Build GRCh38
rs760454363 Chr.4: 122912672-122912672 on Build GRCh38
rs56037335 Chr.4: 122912896-122912896 on Build GRCh38
rs1054134983 Chr.4: 122912870-122912870 on Build GRCh38
rs746791314 Chr.4: 122913059-122913059 on Build GRCh38
rs1027426345 Chr.4: 122912776-122912776 on Build GRCh38
rs934291258 Chr.4: 122912845-122912845 on Build GRCh38
rs753209423 Chr.4: 122912823-122912823 on Build GRCh38
rs1026331284 Chr.4: 122912962-122912962 on Build GRCh38
rs997588212 Chr.4: 122912622-122912622 on Build GRCh38
rs980994589 Chr.4: 122912837-122912837 on Build GRCh38
rs751967679 Chr.4: 122912664-122912664 on Build GRCh38
rs961694770 Chr.4: 122912723-122912723 on Build GRCh38
rs55691042 Chr.4: 122912763-122912763 on Build GRCh38
rs371981022 Chr.4: 122912635-122912635 on Build GRCh38
rs548028119 Chr.4: 122912994-122912994 on Build GRCh38
rs904329811 Chr.4: 122912603-122912603 on Build GRCh38
rs550707462 Chr.4: 122912694-122912694 on Build GRCh38

View Full Product Details