Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 40761082-40761638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKAA1
Entrez Gene ID
5562
Gene Name
protein kinase AMP-activated catalytic subunit alpha 1
Gene Aliases
AMPK, AMPKa1
UniGene
Hs.43322
Gene Chromosome Location
Chr.5: 40759379-40798195 on Build GRCh38
Transcripts
NM_006251.5 NM_206907.3 XM_006714482.1 XM_017009624.1 XM_017009625.1

SNP Detail

SNP ID Chromosome Location
rs990762351 Chr.5: 40761250-40761250 on Build GRCh38
rs1036821500 Chr.5: 40761354-40761354 on Build GRCh38
rs564824934 Chr.5: 40761377-40761378 on Build GRCh38
rs996711995 Chr.5: 40761613-40761613 on Build GRCh38
rs913390134 Chr.5: 40761260-40761260 on Build GRCh38
rs897876733 Chr.5: 40761566-40761566 on Build GRCh38
rs183113419 Chr.5: 40761376-40761376 on Build GRCh38
rs994382253 Chr.5: 40761371-40761371 on Build GRCh38
rs891086112 Chr.5: 40761612-40761612 on Build GRCh38
rs896990167 Chr.5: 40761357-40761357 on Build GRCh38
rs924603172 Chr.5: 40761325-40761325 on Build GRCh38
rs909225382 Chr.5: 40761553-40761553 on Build GRCh38
rs573055558 Chr.5: 40761373-40761373 on Build GRCh38
rs556093072 Chr.5: 40761557-40761557 on Build GRCh38
rs957089442 Chr.5: 40761179-40761179 on Build GRCh38
rs1055705819 Chr.5: 40761311-40761311 on Build GRCh38
rs955260829 Chr.5: 40761377-40761377 on Build GRCh38
rs745530996 Chr.5: 40761227-40761227 on Build GRCh38
rs951397646 Chr.5: 40761157-40761157 on Build GRCh38
rs968777057 Chr.5: 40761316-40761316 on Build GRCh38
rs938964712 Chr.5: 40761364-40761364 on Build GRCh38
rs963887896 Chr.5: 40761283-40761283 on Build GRCh38
rs756469301 Chr.5: 40761143-40761147 on Build GRCh38
rs191771027 Chr.5: 40761302-40761302 on Build GRCh38
rs1029593031 Chr.5: 40761372-40761372 on Build GRCh38
rs1006583108 Chr.5: 40761397-40761397 on Build GRCh38
rs760301103 Chr.5: 40761294-40761294 on Build GRCh38
rs985094952 Chr.5: 40761123-40761123 on Build GRCh38
rs887902922 Chr.5: 40761319-40761319 on Build GRCh38
rs898276847 Chr.5: 40761444-40761445 on Build GRCh38
rs530852178 Chr.5: 40761607-40761607 on Build GRCh38
rs201036313 Chr.5: 40761276-40761276 on Build GRCh38
rs763633699 Chr.5: 40761264-40761267 on Build GRCh38
rs991485793 Chr.5: 40761524-40761524 on Build GRCh38
rs928916555 Chr.5: 40761505-40761505 on Build GRCh38
rs1029953729 Chr.5: 40761112-40761112 on Build GRCh38
rs749973791 Chr.5: 40761446-40761446 on Build GRCh38
rs142677351 Chr.5: 40761439-40761439 on Build GRCh38
rs1025530065 Chr.5: 40761277-40761277 on Build GRCh38
rs527574612 Chr.5: 40761419-40761419 on Build GRCh38
rs941996762 Chr.5: 40761554-40761554 on Build GRCh38
rs112364555 Chr.5: 40761107-40761107 on Build GRCh38
rs929853840 Chr.5: 40761572-40761572 on Build GRCh38
rs571778582 Chr.5: 40761121-40761121 on Build GRCh38
rs563699720 Chr.5: 40761552-40761552 on Build GRCh38
rs995216224 Chr.5: 40761482-40761482 on Build GRCh38
rs147350847 Chr.5: 40761493-40761493 on Build GRCh38
rs567650373 Chr.5: 40761473-40761473 on Build GRCh38
rs78955876 Chr.5: 40761320-40761320 on Build GRCh38
rs955043514 Chr.5: 40761118-40761118 on Build GRCh38
rs41271117 Chr.5: 40761284-40761284 on Build GRCh38
rs911002559 Chr.5: 40761156-40761156 on Build GRCh38
rs1017949809 Chr.5: 40761412-40761412 on Build GRCh38
rs766223652 Chr.5: 40761490-40761490 on Build GRCh38
rs761427602 Chr.5: 40761432-40761435 on Build GRCh38
rs188251829 Chr.5: 40761435-40761435 on Build GRCh38
rs34482915 Chr.5: 40761608-40761609 on Build GRCh38
rs763565371 Chr.5: 40761518-40761518 on Build GRCh38
rs771680417 Chr.5: 40761273-40761273 on Build GRCh38
rs1008565187 Chr.5: 40761540-40761540 on Build GRCh38

View Full Product Details