Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117224000-117224513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCSK7
Entrez Gene ID
9159
Gene Name
proprotein convertase subtilisin/kexin type 7
Gene Aliases
LPC, PC7, PC8, SPC7
UniGene
Hs.648612
Gene Chromosome Location
Chr.11: 117205071-117232525 on Build GRCh38
Transcripts
NM_004716.3 XM_017018546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3368229 c.1001A>G p.N334S Chr.11: 117224131-117224131 on Build GRCh38
6131293 c.989G>C p.G330A Chr.11: 117224143-117224143 on Build GRCh38
4712980 c.1023C>T p.G341G Chr.11: 117224109-117224109 on Build GRCh38
1159563 c.954C>G p.G318G Chr.11: 117224178-117224178 on Build GRCh38
6742140 c.916G>T p.A306S Chr.11: 117224216-117224216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138818713 Chr.11: 117224329-117224329 on Build GRCh38
rs772874563 Chr.11: 117224229-117224229 on Build GRCh38
rs758956991 Chr.11: 117224030-117224030 on Build GRCh38
rs778079838 Chr.11: 117224264-117224264 on Build GRCh38
rs755108938 Chr.11: 117224044-117224044 on Build GRCh38
rs917734892 Chr.11: 117224220-117224220 on Build GRCh38
rs569662186 Chr.11: 117224115-117224115 on Build GRCh38
rs559742385 Chr.11: 117224272-117224272 on Build GRCh38
rs111680516 Chr.11: 117224414-117224414 on Build GRCh38
rs34117974 Chr.11: 117224065-117224066 on Build GRCh38
rs552155438 Chr.11: 117224270-117224270 on Build GRCh38
rs752121996 Chr.11: 117224110-117224110 on Build GRCh38
rs775657882 Chr.11: 117224114-117224114 on Build GRCh38
rs531193281 Chr.11: 117224138-117224138 on Build GRCh38
rs377246346 Chr.11: 117224392-117224392 on Build GRCh38
rs142989718 Chr.11: 117224120-117224120 on Build GRCh38
rs765567568 Chr.11: 117224088-117224088 on Build GRCh38
rs759664069 Chr.11: 117224448-117224448 on Build GRCh38
rs745605376 Chr.11: 117224145-117224145 on Build GRCh38
rs775366580 Chr.11: 117224481-117224481 on Build GRCh38
rs142825051 Chr.11: 117224397-117224397 on Build GRCh38
rs769400589 Chr.11: 117224222-117224222 on Build GRCh38
rs756280940 Chr.11: 117224260-117224260 on Build GRCh38
rs200527015 Chr.11: 117224048-117224048 on Build GRCh38
rs751994789 Chr.11: 117224237-117224237 on Build GRCh38
rs952115711 Chr.11: 117224405-117224405 on Build GRCh38
rs566498969 Chr.11: 117224476-117224476 on Build GRCh38
rs903531757 Chr.11: 117224393-117224393 on Build GRCh38
rs200039586 Chr.11: 117224257-117224257 on Build GRCh38
rs940965182 Chr.11: 117224170-117224170 on Build GRCh38
rs115599605 Chr.11: 117224130-117224130 on Build GRCh38
rs542157926 Chr.11: 117224079-117224079 on Build GRCh38
rs772416098 Chr.11: 117224041-117224044 on Build GRCh38
rs908085540 Chr.11: 117224168-117224168 on Build GRCh38
rs774336211 Chr.11: 117224058-117224058 on Build GRCh38
rs1000914807 Chr.11: 117224403-117224403 on Build GRCh38
rs758120801 Chr.11: 117224265-117224265 on Build GRCh38
rs1024669602 Chr.11: 117224363-117224363 on Build GRCh38
rs771910214 Chr.11: 117224166-117224166 on Build GRCh38
rs923559030 Chr.11: 117224463-117224463 on Build GRCh38
rs776147013 Chr.11: 117224086-117224086 on Build GRCh38
rs546376522 Chr.11: 117224437-117224437 on Build GRCh38
rs776170732 Chr.11: 117224204-117224204 on Build GRCh38
rs201586649 Chr.11: 117224172-117224172 on Build GRCh38
rs766773697 Chr.11: 117224233-117224233 on Build GRCh38
rs746264965 Chr.11: 117224040-117224041 on Build GRCh38
rs745768879 Chr.11: 117224061-117224061 on Build GRCh38
rs748357376 Chr.11: 117224122-117224122 on Build GRCh38
rs374854628 Chr.11: 117224106-117224106 on Build GRCh38
rs997756988 Chr.11: 117224282-117224282 on Build GRCh38
rs368923684 Chr.11: 117224070-117224070 on Build GRCh38
rs568437092 Chr.11: 117224335-117224335 on Build GRCh38
rs199595867 Chr.11: 117224104-117224104 on Build GRCh38
rs747020257 Chr.11: 117224037-117224037 on Build GRCh38
rs532625371 Chr.11: 117224269-117224269 on Build GRCh38
rs147004182 Chr.11: 117224423-117224423 on Build GRCh38
rs528465285 Chr.11: 117224281-117224281 on Build GRCh38
rs552984510 Chr.11: 117224465-117224465 on Build GRCh38
rs149062087 Chr.11: 117224065-117224065 on Build GRCh38
rs141025136 Chr.11: 117224186-117224186 on Build GRCh38
rs750831268 Chr.11: 117224094-117224094 on Build GRCh38
rs747032045 Chr.11: 117224185-117224185 on Build GRCh38
rs779088477 Chr.11: 117224041-117224042 on Build GRCh38
rs763662107 Chr.11: 117224085-117224085 on Build GRCh38
rs192060612 Chr.11: 117224449-117224449 on Build GRCh38
rs891686172 Chr.11: 117224338-117224338 on Build GRCh38
rs775516955 Chr.11: 117224066-117224066 on Build GRCh38
rs765373647 Chr.11: 117224453-117224453 on Build GRCh38
rs199538186 Chr.11: 117224129-117224129 on Build GRCh38
rs73580314 Chr.11: 117224249-117224249 on Build GRCh38
rs555462323 Chr.11: 117224483-117224483 on Build GRCh38
rs763171212 Chr.11: 117224232-117224232 on Build GRCh38
rs12288538 Chr.11: 117224036-117224036 on Build GRCh38
rs752783500 Chr.11: 117224259-117224259 on Build GRCh38
rs535083896 Chr.11: 117224477-117224477 on Build GRCh38
rs771389545 Chr.11: 117224383-117224383 on Build GRCh38
rs563654447 Chr.11: 117224267-117224267 on Build GRCh38
rs373226776 Chr.11: 117224219-117224219 on Build GRCh38
rs770835291 Chr.11: 117224052-117224052 on Build GRCh38
rs889536615 Chr.11: 117224263-117224263 on Build GRCh38
rs747425506 Chr.11: 117224367-117224367 on Build GRCh38
rs1010133778 Chr.11: 117224349-117224349 on Build GRCh38
rs1034393940 Chr.11: 117224396-117224396 on Build GRCh38
rs372878339 Chr.11: 117224087-117224087 on Build GRCh38

View Full Product Details