Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117822557-117823054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXYD2
Entrez Gene ID
486
Gene Name
FXYD domain containing ion transport regulator 2
Gene Aliases
ATP1G1, HOMG2
UniGene
Hs.731865
Gene Chromosome Location
Chr.11: 117820075-117828092 on Build GRCh38
Transcripts
NM_001680.4 NM_021603.3 NM_001204268.1 NM_001243598.2

Gene Symbol
FXYD6-FXYD2
Entrez Gene ID
100533181
Gene Name
FXYD6-FXYD2 readthrough
Gene Aliases
-
UniGene
Hs.744850
Gene Chromosome Location
Chr.11: 117820075-117877031 on Build GRCh38
Transcripts
NM_001680.4 NM_021603.3 NM_001204268.1 NM_001243598.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1660043 c.58+4A>T p.? Chr.11: 117822675-117822675 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs928889734 Chr.11: 117822624-117822624 on Build GRCh38
rs931480224 Chr.11: 117822954-117822954 on Build GRCh38
rs985678944 Chr.11: 117823016-117823016 on Build GRCh38
rs757376396 Chr.11: 117822888-117822888 on Build GRCh38
rs760295629 Chr.11: 117822590-117822590 on Build GRCh38
rs1049699406 Chr.11: 117822743-117822743 on Build GRCh38
rs781565039 Chr.11: 117822640-117822640 on Build GRCh38
rs866550911 Chr.11: 117822795-117822795 on Build GRCh38
rs766472507 Chr.11: 117822692-117822692 on Build GRCh38
rs368479505 Chr.11: 117822674-117822674 on Build GRCh38
rs375348085 Chr.11: 117822876-117822876 on Build GRCh38
rs372610059 Chr.11: 117822997-117822997 on Build GRCh38
rs182462094 Chr.11: 117822722-117822722 on Build GRCh38
rs377266813 Chr.11: 117822755-117822755 on Build GRCh38
rs575666477 Chr.11: 117822945-117822945 on Build GRCh38
rs533959280 Chr.11: 117822816-117822816 on Build GRCh38
rs891436883 Chr.11: 117822813-117822813 on Build GRCh38
rs763425417 Chr.11: 117822690-117822690 on Build GRCh38
rs1045467136 Chr.11: 117822818-117822818 on Build GRCh38
rs146614981 Chr.11: 117822715-117822715 on Build GRCh38
rs778952542 Chr.11: 117822659-117822659 on Build GRCh38
rs573639588 Chr.11: 117822894-117822894 on Build GRCh38
rs773513184 Chr.11: 117822753-117822753 on Build GRCh38
rs763375923 Chr.11: 117822754-117822754 on Build GRCh38
rs1020005731 Chr.11: 117822846-117822846 on Build GRCh38
rs371633046 Chr.11: 117822870-117822870 on Build GRCh38
rs754269604 Chr.11: 117822607-117822607 on Build GRCh38
rs985180049 Chr.11: 117822711-117822711 on Build GRCh38
rs889466585 Chr.11: 117822600-117822600 on Build GRCh38
rs535588993 Chr.11: 117822721-117822721 on Build GRCh38
rs750953668 Chr.11: 117822617-117822617 on Build GRCh38
rs1047141619 Chr.11: 117822955-117822956 on Build GRCh38
rs747841055 Chr.11: 117822737-117822737 on Build GRCh38
rs550904239 Chr.11: 117822635-117822635 on Build GRCh38
rs775179288 Chr.11: 117822587-117822587 on Build GRCh38
rs887191178 Chr.11: 117822975-117822975 on Build GRCh38
rs762119604 Chr.11: 117822683-117822683 on Build GRCh38
rs955272779 Chr.11: 117822990-117822990 on Build GRCh38
rs560526966 Chr.11: 117823024-117823024 on Build GRCh38
rs758335144 Chr.11: 117822716-117822716 on Build GRCh38
rs749032169 Chr.11: 117822646-117822646 on Build GRCh38
rs778901593 Chr.11: 117822707-117822707 on Build GRCh38
rs542360145 Chr.11: 117822905-117822905 on Build GRCh38
rs760372029 Chr.11: 117822673-117822673 on Build GRCh38
rs751789620 Chr.11: 117822634-117822634 on Build GRCh38
rs752997671 Chr.11: 117822701-117822701 on Build GRCh38
rs150608549 Chr.11: 117822688-117822688 on Build GRCh38
rs531015300 Chr.11: 117822726-117822726 on Build GRCh38
rs79521383 Chr.11: 117822867-117822867 on Build GRCh38
rs201735486 Chr.11: 117822660-117822660 on Build GRCh38
rs1014897829 Chr.11: 117822933-117822933 on Build GRCh38
rs776282287 Chr.11: 117822680-117822680 on Build GRCh38
rs755098516 Chr.11: 117822699-117822699 on Build GRCh38
rs117170428 Chr.11: 117822959-117822959 on Build GRCh38
rs991653163 Chr.11: 117822928-117822929 on Build GRCh38
rs771512838 Chr.11: 117822585-117822585 on Build GRCh38
rs41302381 Chr.11: 117823017-117823017 on Build GRCh38
rs774922528 Chr.11: 117822665-117822665 on Build GRCh38
rs562135238 Chr.11: 117822916-117822916 on Build GRCh38
rs952384889 Chr.11: 117822918-117822918 on Build GRCh38
rs767799368 Chr.11: 117822700-117822700 on Build GRCh38
rs764506296 Chr.11: 117822601-117822601 on Build GRCh38
rs981077218 Chr.11: 117822893-117822893 on Build GRCh38
rs1030363683 Chr.11: 117822976-117822976 on Build GRCh38
rs149796718 Chr.11: 117822728-117822728 on Build GRCh38
rs562154032 Chr.11: 117822619-117822619 on Build GRCh38
rs765679681 Chr.11: 117822611-117822611 on Build GRCh38
rs757085841 Chr.11: 117822702-117822702 on Build GRCh38
rs776228953 Chr.11: 117822732-117822732 on Build GRCh38
rs1006445179 Chr.11: 117822927-117822927 on Build GRCh38
rs191571857 Chr.11: 117822947-117822947 on Build GRCh38
rs773683745 Chr.11: 117822689-117822689 on Build GRCh38
rs765498478 Chr.11: 117822906-117822906 on Build GRCh38
rs558117050 Chr.11: 117822768-117822768 on Build GRCh38
rs199722279 Chr.11: 117822620-117822620 on Build GRCh38
rs187784200 Chr.11: 117822798-117822798 on Build GRCh38
rs989836044 Chr.11: 117822827-117822827 on Build GRCh38
rs770872034 Chr.11: 117822651-117822651 on Build GRCh38
rs544483613 Chr.11: 117822946-117822946 on Build GRCh38
rs1010006011 Chr.11: 117822817-117822817 on Build GRCh38
rs35705701 Chr.11: 117822698-117822698 on Build GRCh38

View Full Product Details