Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118659280-118659776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TREH
Entrez Gene ID
11181
Gene Name
trehalase
Gene Aliases
TRE, TREA
UniGene
Hs.129712
Gene Chromosome Location
Chr.11: 118658232-118679672 on Build GRCh38
Transcripts
NM_001301065.1 NM_007180.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3397456 c.967A>G p.T323A Chr.11: 118659466-118659466 on Build GRCh38
7326516 c.951+7_951+8insG p.? Chr.11: 118659739-118659740 on Build GRCh38
1351696 c.1320+1G>A p.? Chr.11: 118659746-118659746 on Build GRCh38
7180311 c.1002G>T p.K334N Chr.11: 118659431-118659431 on Build GRCh38
7326515 c.1320+7_1320+8insG p.? Chr.11: 118659739-118659740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374460069 Chr.11: 118659510-118659510 on Build GRCh38
rs577377304 Chr.11: 118659687-118659687 on Build GRCh38
rs546157639 Chr.11: 118659655-118659655 on Build GRCh38
rs536690290 Chr.11: 118659504-118659504 on Build GRCh38
rs782349165 Chr.11: 118659402-118659402 on Build GRCh38
rs781917462 Chr.11: 118659521-118659522 on Build GRCh38
rs782006844 Chr.11: 118659415-118659415 on Build GRCh38
rs782681922 Chr.11: 118659475-118659475 on Build GRCh38
rs782626070 Chr.11: 118659393-118659393 on Build GRCh38
rs782178104 Chr.11: 118659398-118659398 on Build GRCh38
rs112570360 Chr.11: 118659550-118659550 on Build GRCh38
rs782579310 Chr.11: 118659397-118659397 on Build GRCh38
rs782070070 Chr.11: 118659441-118659441 on Build GRCh38
rs781811872 Chr.11: 118659365-118659365 on Build GRCh38
rs782701404 Chr.11: 118659442-118659442 on Build GRCh38
rs573627789 Chr.11: 118659656-118659656 on Build GRCh38
rs367908002 Chr.11: 118659396-118659396 on Build GRCh38
rs782282504 Chr.11: 118659394-118659394 on Build GRCh38
rs782545765 Chr.11: 118659349-118659349 on Build GRCh38
rs782613056 Chr.11: 118659378-118659378 on Build GRCh38
rs186780088 Chr.11: 118659446-118659446 on Build GRCh38
rs117619140 Chr.11: 118659725-118659725 on Build GRCh38
rs781800491 Chr.11: 118659467-118659467 on Build GRCh38
rs782818824 Chr.11: 118659507-118659507 on Build GRCh38
rs782015297 Chr.11: 118659553-118659553 on Build GRCh38
rs782117647 Chr.11: 118659420-118659420 on Build GRCh38
rs544459106 Chr.11: 118659721-118659721 on Build GRCh38
rs782416144 Chr.11: 118659538-118659538 on Build GRCh38
rs1133780 Chr.11: 118659382-118659382 on Build GRCh38
rs782014731 Chr.11: 118659426-118659426 on Build GRCh38
rs782223111 Chr.11: 118659445-118659445 on Build GRCh38
rs782178282 Chr.11: 118659742-118659742 on Build GRCh38
rs781877579 Chr.11: 118659515-118659515 on Build GRCh38
rs546801778 Chr.11: 118659392-118659392 on Build GRCh38
rs782544797 Chr.11: 118659734-118659734 on Build GRCh38
rs782667217 Chr.11: 118659459-118659459 on Build GRCh38
rs540446493 Chr.11: 118659635-118659635 on Build GRCh38
rs782059500 Chr.11: 118659523-118659523 on Build GRCh38
rs782653438 Chr.11: 118659447-118659447 on Build GRCh38
rs779137111 Chr.11: 118659568-118659568 on Build GRCh38
rs782422052 Chr.11: 118659390-118659391 on Build GRCh38
rs782324008 Chr.11: 118659492-118659492 on Build GRCh38
rs781854435 Chr.11: 118659449-118659449 on Build GRCh38
rs35892163 Chr.11: 118659678-118659678 on Build GRCh38
rs7932296 Chr.11: 118659593-118659593 on Build GRCh38
rs557300358 Chr.11: 118659484-118659484 on Build GRCh38
rs782370291 Chr.11: 118659630-118659630 on Build GRCh38
rs11827611 Chr.11: 118659457-118659457 on Build GRCh38
rs559075998 Chr.11: 118659659-118659659 on Build GRCh38
rs149499402 Chr.11: 118659503-118659503 on Build GRCh38
rs781819385 Chr.11: 118659388-118659388 on Build GRCh38
rs377304435 Chr.11: 118659522-118659522 on Build GRCh38
rs782087146 Chr.11: 118659329-118659329 on Build GRCh38
rs369679930 Chr.11: 118659473-118659473 on Build GRCh38
rs782105811 Chr.11: 118659495-118659495 on Build GRCh38
rs782776630 Chr.11: 118659330-118659330 on Build GRCh38
rs782191193 Chr.11: 118659555-118659555 on Build GRCh38
rs782684164 Chr.11: 118659701-118659701 on Build GRCh38
rs377125906 Chr.11: 118659651-118659651 on Build GRCh38
rs782524459 Chr.11: 118659724-118659724 on Build GRCh38
rs781841527 Chr.11: 118659444-118659444 on Build GRCh38
rs112337443 Chr.11: 118659359-118659359 on Build GRCh38
rs548665440 Chr.11: 118659315-118659315 on Build GRCh38
rs782595383 Chr.11: 118659741-118659741 on Build GRCh38
rs528258475 Chr.11: 118659368-118659368 on Build GRCh38
rs782267995 Chr.11: 118659474-118659474 on Build GRCh38
rs781862844 Chr.11: 118659346-118659346 on Build GRCh38
rs573318072 Chr.11: 118659612-118659612 on Build GRCh38

View Full Product Details