Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118896040-118896545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL9L
Entrez Gene ID
283149
Gene Name
B-cell CLL/lymphoma 9-like
Gene Aliases
BCL9-2, DLNB11
UniGene
Hs.414740
Gene Chromosome Location
Chr.11: 118896140-118926072 on Build GRCh38
Transcripts
NM_182557.2 XM_005271511.2 XM_006718815.2 XM_017017572.1 NM_001716.4 NM_032966.2

Gene Symbol
CXCR5
Entrez Gene ID
643
Gene Name
C-X-C motif chemokine receptor 5
Gene Aliases
BLR1, CD185, MDR15
UniGene
Hs.113916
Gene Chromosome Location
Chr.11: 118883766-118896271 on Build GRCh38
Transcripts
NM_182557.2 XM_005271511.2 XM_006718815.2 XM_017017572.1 NM_001716.4 NM_032966.2

SNP Detail

SNP ID Chromosome Location
rs944131045 Chr.11: 118896521-118896521 on Build GRCh38
rs55938970 Chr.11: 118896178-118896178 on Build GRCh38
rs553580654 Chr.11: 118896404-118896404 on Build GRCh38
rs992473716 Chr.11: 118896117-118896117 on Build GRCh38
rs974320817 Chr.11: 118896282-118896293 on Build GRCh38
rs900068283 Chr.11: 118896323-118896323 on Build GRCh38
rs971076813 Chr.11: 118896401-118896401 on Build GRCh38
rs951234666 Chr.11: 118896461-118896461 on Build GRCh38
rs944726473 Chr.11: 118896502-118896502 on Build GRCh38
rs904649911 Chr.11: 118896366-118896366 on Build GRCh38
rs983966718 Chr.11: 118896465-118896465 on Build GRCh38
rs900749861 Chr.11: 118896326-118896326 on Build GRCh38
rs927209691 Chr.11: 118896263-118896263 on Build GRCh38
rs1053877 Chr.11: 118896135-118896135 on Build GRCh38
rs983300634 Chr.11: 118896491-118896491 on Build GRCh38
rs551370085 Chr.11: 118896290-118896290 on Build GRCh38
rs911925821 Chr.11: 118896292-118896292 on Build GRCh38
rs542970845 Chr.11: 118896488-118896488 on Build GRCh38
rs890438954 Chr.11: 118896082-118896082 on Build GRCh38
rs566165476 Chr.11: 118896090-118896090 on Build GRCh38
rs535234229 Chr.11: 118896369-118896369 on Build GRCh38
rs908607840 Chr.11: 118896513-118896513 on Build GRCh38
rs973307579 Chr.11: 118896420-118896420 on Build GRCh38
rs559739639 Chr.11: 118896294-118896294 on Build GRCh38
rs3782048 Chr.11: 118896328-118896328 on Build GRCh38
rs14811 Chr.11: 118896151-118896151 on Build GRCh38
rs908639077 Chr.11: 118896289-118896289 on Build GRCh38
rs1033144681 Chr.11: 118896191-118896191 on Build GRCh38
rs772705742 Chr.11: 118896400-118896400 on Build GRCh38
rs581063 Chr.11: 118896368-118896368 on Build GRCh38
rs55820058 Chr.11: 118896177-118896177 on Build GRCh38
rs539355358 Chr.11: 118896406-118896406 on Build GRCh38
rs970030862 Chr.11: 118896208-118896208 on Build GRCh38
rs554540102 Chr.11: 118896459-118896460 on Build GRCh38
rs982831692 Chr.11: 118896288-118896288 on Build GRCh38
rs973636826 Chr.11: 118896212-118896212 on Build GRCh38
rs71482158 Chr.11: 118896375-118896375 on Build GRCh38
rs929128944 Chr.11: 118896287-118896287 on Build GRCh38
rs533815643 Chr.11: 118896125-118896125 on Build GRCh38
rs1043613835 Chr.11: 118896359-118896359 on Build GRCh38
rs766106371 Chr.11: 118896463-118896463 on Build GRCh38
rs558537383 Chr.11: 118896278-118896278 on Build GRCh38
rs575646091 Chr.11: 118896446-118896446 on Build GRCh38
rs35022133 Chr.11: 118896270-118896270 on Build GRCh38
rs1047779575 Chr.11: 118896287-118896288 on Build GRCh38
rs904261069 Chr.11: 118896072-118896072 on Build GRCh38
rs557350357 Chr.11: 118896412-118896412 on Build GRCh38
rs1041072773 Chr.11: 118896312-118896312 on Build GRCh38
rs552272713 Chr.11: 118896248-118896250 on Build GRCh38
rs762624114 Chr.11: 118896450-118896450 on Build GRCh38
rs183762696 Chr.11: 118896367-118896367 on Build GRCh38
rs959672296 Chr.11: 118896099-118896099 on Build GRCh38
rs1031383990 Chr.11: 118896096-118896096 on Build GRCh38
rs973994646 Chr.11: 118896441-118896442 on Build GRCh38
rs962950567 Chr.11: 118896426-118896426 on Build GRCh38
rs998599613 Chr.11: 118896087-118896087 on Build GRCh38
rs772504922 Chr.11: 118896091-118896091 on Build GRCh38
rs1000771263 Chr.11: 118896143-118896143 on Build GRCh38
rs918388217 Chr.11: 118896284-118896284 on Build GRCh38
rs1027995218 Chr.11: 118896430-118896430 on Build GRCh38
rs980011057 Chr.11: 118896250-118896250 on Build GRCh38
rs921902817 Chr.11: 118896522-118896522 on Build GRCh38
rs578105206 Chr.11: 118896405-118896405 on Build GRCh38
rs1024403064 Chr.11: 118896222-118896222 on Build GRCh38

View Full Product Details