Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 121307364-121307873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SC5D
Entrez Gene ID
6309
Gene Name
sterol-C5-desaturase
Gene Aliases
ERG3, S5DES, SC5DL
UniGene
Hs.287749
Gene Chromosome Location
Chr.11: 121292679-121313410 on Build GRCh38
Transcripts
NM_001024956.2 NM_006918.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
218298 c.754G>A p.G252S Chr.11: 121307366-121307366 on Build GRCh38
6772298 c.834C>T p.S278S Chr.11: 121307446-121307446 on Build GRCh38
3444347 c.829C>T p.R277C Chr.11: 121307441-121307441 on Build GRCh38
3444346 c.776C>T p.S259F Chr.11: 121307388-121307388 on Build GRCh38
924036 c.761T>G p.F254C Chr.11: 121307373-121307373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778722898 Chr.11: 121307555-121307555 on Build GRCh38
rs956181781 Chr.11: 121307419-121307419 on Build GRCh38
rs377729279 Chr.11: 121307643-121307643 on Build GRCh38
rs768181937 Chr.11: 121307493-121307493 on Build GRCh38
rs537561364 Chr.11: 121307624-121307624 on Build GRCh38
rs141523614 Chr.11: 121307597-121307597 on Build GRCh38
rs370547683 Chr.11: 121307523-121307523 on Build GRCh38
rs767759025 Chr.11: 121307539-121307539 on Build GRCh38
rs762375370 Chr.11: 121307431-121307432 on Build GRCh38
rs1031911283 Chr.11: 121307669-121307669 on Build GRCh38
rs185522407 Chr.11: 121307787-121307787 on Build GRCh38
rs118099079 Chr.11: 121307441-121307441 on Build GRCh38
rs770323739 Chr.11: 121307532-121307535 on Build GRCh38
rs942908119 Chr.11: 121307766-121307766 on Build GRCh38
rs776041178 Chr.11: 121307494-121307494 on Build GRCh38
rs374568188 Chr.11: 121307527-121307527 on Build GRCh38
rs187447148 Chr.11: 121307562-121307562 on Build GRCh38
rs1008454073 Chr.11: 121307649-121307649 on Build GRCh38
rs778899115 Chr.11: 121307455-121307455 on Build GRCh38
rs745380178 Chr.11: 121307406-121307406 on Build GRCh38
rs758926625 Chr.11: 121307478-121307478 on Build GRCh38
rs750306836 Chr.11: 121307556-121307556 on Build GRCh38
rs751763860 Chr.11: 121307438-121307438 on Build GRCh38
rs146228207 Chr.11: 121307775-121307775 on Build GRCh38
rs750799609 Chr.11: 121307398-121307398 on Build GRCh38
rs928598332 Chr.11: 121307741-121307741 on Build GRCh38
rs184677354 Chr.11: 121307553-121307553 on Build GRCh38
rs980102323 Chr.11: 121307400-121307400 on Build GRCh38
rs972740616 Chr.11: 121307700-121307700 on Build GRCh38
rs574865551 Chr.11: 121307560-121307560 on Build GRCh38
rs766772523 Chr.11: 121307427-121307427 on Build GRCh38
rs553818571 Chr.11: 121307825-121307825 on Build GRCh38
rs192695606 Chr.11: 121307732-121307732 on Build GRCh38
rs867806506 Chr.11: 121307729-121307729 on Build GRCh38
rs960026200 Chr.11: 121307689-121307689 on Build GRCh38
rs1049044204 Chr.11: 121307497-121307497 on Build GRCh38
rs1052104199 Chr.11: 121307833-121307833 on Build GRCh38
rs1031818443 Chr.11: 121307407-121307407 on Build GRCh38
rs368283320 Chr.11: 121307547-121307547 on Build GRCh38
rs369195944 Chr.11: 121307520-121307520 on Build GRCh38
rs1061332 Chr.11: 121307593-121307593 on Build GRCh38
rs759878140 Chr.11: 121307533-121307533 on Build GRCh38
rs568068471 Chr.11: 121307672-121307672 on Build GRCh38
rs939838511 Chr.11: 121307554-121307554 on Build GRCh38
rs914640787 Chr.11: 121307465-121307465 on Build GRCh38
rs367727061 Chr.11: 121307425-121307425 on Build GRCh38
rs576544384 Chr.11: 121307842-121307842 on Build GRCh38
rs761361460 Chr.11: 121307501-121307501 on Build GRCh38
rs898230477 Chr.11: 121307584-121307584 on Build GRCh38
rs745732638 Chr.11: 121307462-121307462 on Build GRCh38
rs980144975 Chr.11: 121307740-121307740 on Build GRCh38
rs911368306 Chr.11: 121307759-121307759 on Build GRCh38
rs763414117 Chr.11: 121307525-121307525 on Build GRCh38
rs942960903 Chr.11: 121307516-121307516 on Build GRCh38
rs566470203 Chr.11: 121307577-121307577 on Build GRCh38
rs960058373 Chr.11: 121307415-121307415 on Build GRCh38
rs768458346 Chr.11: 121307755-121307755 on Build GRCh38
rs111941010 Chr.11: 121307674-121307674 on Build GRCh38
rs546495208 Chr.11: 121307782-121307782 on Build GRCh38
rs751861611 Chr.11: 121307528-121307528 on Build GRCh38
rs771792307 Chr.11: 121307483-121307483 on Build GRCh38
rs757191362 Chr.11: 121307445-121307445 on Build GRCh38
rs1011746511 Chr.11: 121307717-121307717 on Build GRCh38
rs939890940 Chr.11: 121307831-121307831 on Build GRCh38
rs1010701046 Chr.11: 121307574-121307574 on Build GRCh38
rs919685649 Chr.11: 121307845-121307845 on Build GRCh38
rs570600004 Chr.11: 121307612-121307612 on Build GRCh38
rs771541681 Chr.11: 121307486-121307486 on Build GRCh38
rs535487230 Chr.11: 121307720-121307720 on Build GRCh38
rs372543290 Chr.11: 121307561-121307561 on Build GRCh38
rs970507680 Chr.11: 121307731-121307731 on Build GRCh38
rs779699551 Chr.11: 121307485-121307485 on Build GRCh38
rs556185890 Chr.11: 121307657-121307657 on Build GRCh38
rs970120462 Chr.11: 121307469-121307469 on Build GRCh38
rs367608610 Chr.11: 121307487-121307487 on Build GRCh38
rs114578771 Chr.11: 121307442-121307442 on Build GRCh38

View Full Product Details