Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 43783175-43783721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRAK4
Entrez Gene ID
51135
Gene Name
interleukin 1 receptor associated kinase 4
Gene Aliases
IPD1, IRAK-4, NY-REN-64, REN64
UniGene
Hs.138499
Gene Chromosome Location
Chr.12: 43758913-43789543 on Build GRCh38
Transcripts
NM_001114182.2 NM_001145256.1 NM_001145257.1 NM_001145258.1 NM_016123.3 XM_005268943.3 XM_005268944.4 XM_005268945.4 XM_005268947.4 XM_005268948.2 XM_005268949.2 XM_006719438.3 XM_006719439.2 XM_006719440.2 XM_006719441.2 XM_006719442.2 XM_006719443.3 XM_011538431.2 XM_011538432.1 XM_011538433.2 XM_017019390.1 XM_017019391.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4564378 c.1161_1162GG>AA p.D388N Chr.12: 43783697-43783698 on Build GRCh38
6398819 c.1162G>T p.D388Y Chr.12: 43783698-43783698 on Build GRCh38
6462600 c.1167A>G p.E389E Chr.12: 43783703-43783703 on Build GRCh38
4041869 c.1181A>C p.Q394P Chr.12: 43783717-43783717 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs4251529 Chr.12: 43783551-43783551 on Build GRCh38
rs986445433 Chr.12: 43783324-43783324 on Build GRCh38
rs759683832 Chr.12: 43783625-43783625 on Build GRCh38
rs774384959 Chr.12: 43783642-43783642 on Build GRCh38
rs765296895 Chr.12: 43783487-43783487 on Build GRCh38
rs771146816 Chr.12: 43783619-43783619 on Build GRCh38
rs755772831 Chr.12: 43783684-43783684 on Build GRCh38
rs776634104 Chr.12: 43783620-43783620 on Build GRCh38
rs996206638 Chr.12: 43783350-43783350 on Build GRCh38
rs890875157 Chr.12: 43783637-43783637 on Build GRCh38
rs547883821 Chr.12: 43783468-43783468 on Build GRCh38
rs186857922 Chr.12: 43783601-43783601 on Build GRCh38
rs560045790 Chr.12: 43783483-43783483 on Build GRCh38
rs914353281 Chr.12: 43783494-43783494 on Build GRCh38
rs891958168 Chr.12: 43783246-43783246 on Build GRCh38
rs955135177 Chr.12: 43783310-43783310 on Build GRCh38
rs767476422 Chr.12: 43783678-43783678 on Build GRCh38
rs572443308 Chr.12: 43783317-43783318 on Build GRCh38
rs1009448073 Chr.12: 43783275-43783275 on Build GRCh38
rs746640027 Chr.12: 43783544-43783544 on Build GRCh38
rs753251856 Chr.12: 43783685-43783685 on Build GRCh38
rs4251530 Chr.12: 43783599-43783599 on Build GRCh38
rs761095560 Chr.12: 43783644-43783644 on Build GRCh38
rs183014058 Chr.12: 43783223-43783223 on Build GRCh38
rs11182263 Chr.12: 43783643-43783643 on Build GRCh38
rs768082448 Chr.12: 43783614-43783614 on Build GRCh38
rs1028087401 Chr.12: 43783304-43783304 on Build GRCh38
rs924338813 Chr.12: 43783405-43783405 on Build GRCh38
rs747299143 Chr.12: 43783616-43783616 on Build GRCh38
rs769668046 Chr.12: 43783517-43783517 on Build GRCh38
rs939462816 Chr.12: 43783547-43783547 on Build GRCh38
rs750092963 Chr.12: 43783681-43783681 on Build GRCh38
rs4251528 Chr.12: 43783539-43783539 on Build GRCh38
rs763896878 Chr.12: 43783629-43783629 on Build GRCh38
rs753200898 Chr.12: 43783643-43783644 on Build GRCh38
rs1040498038 Chr.12: 43783327-43783327 on Build GRCh38
rs1015892869 Chr.12: 43783527-43783527 on Build GRCh38
rs527283663 Chr.12: 43783519-43783519 on Build GRCh38
rs1053736675 Chr.12: 43783205-43783205 on Build GRCh38
rs1031721401 Chr.12: 43783694-43783694 on Build GRCh38
rs529759570 Chr.12: 43783453-43783453 on Build GRCh38
rs764404378 Chr.12: 43783682-43783683 on Build GRCh38
rs997002877 Chr.12: 43783607-43783607 on Build GRCh38
rs544797990 Chr.12: 43783201-43783201 on Build GRCh38
rs1056866576 Chr.12: 43783548-43783548 on Build GRCh38
rs940226495 Chr.12: 43783559-43783559 on Build GRCh38
rs757918302 Chr.12: 43783697-43783697 on Build GRCh38
rs961239829 Chr.12: 43783533-43783533 on Build GRCh38
rs900558188 Chr.12: 43783342-43783342 on Build GRCh38
rs144296954 Chr.12: 43783644-43783644 on Build GRCh38
rs1027638520 Chr.12: 43783392-43783392 on Build GRCh38
rs142891158 Chr.12: 43783249-43783249 on Build GRCh38
rs913448868 Chr.12: 43783605-43783605 on Build GRCh38
rs531426729 Chr.12: 43783598-43783598 on Build GRCh38

View Full Product Details