Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72160924-72161426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAMD2
Entrez Gene ID
196996
Gene Name
GRAM domain containing 2
Gene Aliases
-
UniGene
Hs.596332
Gene Chromosome Location
Chr.15: 72159806-72197814 on Build GRCh38
Transcripts
NM_001012642.2 XM_011521327.2 XM_011521328.2 XM_011521329.2 XM_011521330.2 XM_011521331.2 XM_011521332.2 XM_011521333.2 XM_017021997.1 XM_017021998.1

SNP Detail

SNP ID Chromosome Location
rs755746230 Chr.15: 72161179-72161179 on Build GRCh38
rs1016651898 Chr.15: 72160985-72160985 on Build GRCh38
rs1015346883 Chr.15: 72161031-72161031 on Build GRCh38
rs1017137572 Chr.15: 72161215-72161215 on Build GRCh38
rs1034699266 Chr.15: 72160946-72160946 on Build GRCh38
rs1030918700 Chr.15: 72161306-72161306 on Build GRCh38
rs982944036 Chr.15: 72160969-72160969 on Build GRCh38
rs1044908615 Chr.15: 72161121-72161121 on Build GRCh38
rs568679396 Chr.15: 72161030-72161030 on Build GRCh38
rs377432556 Chr.15: 72161274-72161274 on Build GRCh38
rs893888358 Chr.15: 72161087-72161087 on Build GRCh38
rs951057758 Chr.15: 72160960-72160960 on Build GRCh38
rs570584798 Chr.15: 72160982-72160982 on Build GRCh38
rs767214698 Chr.15: 72161058-72161058 on Build GRCh38
rs1034370181 Chr.15: 72161386-72161386 on Build GRCh38
rs569458584 Chr.15: 72161218-72161218 on Build GRCh38
rs991076408 Chr.15: 72161123-72161123 on Build GRCh38
rs147418513 Chr.15: 72161380-72161380 on Build GRCh38
rs953524253 Chr.15: 72161320-72161320 on Build GRCh38
rs886785277 Chr.15: 72161183-72161183 on Build GRCh38
rs557584961 Chr.15: 72161187-72161187 on Build GRCh38
rs936343330 Chr.15: 72161194-72161194 on Build GRCh38
rs546863698 Chr.15: 72161027-72161027 on Build GRCh38
rs1003462016 Chr.15: 72161137-72161137 on Build GRCh38
rs922214517 Chr.15: 72161008-72161008 on Build GRCh38
rs142941603 Chr.15: 72160989-72160989 on Build GRCh38
rs370137690 Chr.15: 72161327-72161327 on Build GRCh38
rs1053517868 Chr.15: 72161240-72161240 on Build GRCh38
rs558605972 Chr.15: 72161364-72161364 on Build GRCh38
rs752270285 Chr.15: 72161059-72161059 on Build GRCh38
rs932247439 Chr.15: 72161025-72161025 on Build GRCh38
rs987659240 Chr.15: 72161053-72161053 on Build GRCh38
rs112129833 Chr.15: 72161373-72161373 on Build GRCh38
rs1029120275 Chr.15: 72161307-72161307 on Build GRCh38
rs949331984 Chr.15: 72161149-72161149 on Build GRCh38
rs996322325 Chr.15: 72161291-72161291 on Build GRCh38
rs973521653 Chr.15: 72161005-72161005 on Build GRCh38
rs535770409 Chr.15: 72161095-72161095 on Build GRCh38
rs890678256 Chr.15: 72161323-72161323 on Build GRCh38
rs778788426 Chr.15: 72161324-72161324 on Build GRCh38
rs962485505 Chr.15: 72161229-72161229 on Build GRCh38
rs1030554301 Chr.15: 72161014-72161014 on Build GRCh38
rs988155965 Chr.15: 72161374-72161374 on Build GRCh38
rs753871537 Chr.15: 72161256-72161256 on Build GRCh38
rs927405906 Chr.15: 72161086-72161086 on Build GRCh38
rs868307734 Chr.15: 72161230-72161230 on Build GRCh38
rs917820276 Chr.15: 72161128-72161128 on Build GRCh38

View Full Product Details