Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 73885986-73886480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D21
Entrez Gene ID
161514
Gene Name
TBC1 domain family member 21
Gene Aliases
MgcRabGAP
UniGene
Hs.124512
Gene Chromosome Location
Chr.15: 73873556-73909783 on Build GRCh38
Transcripts
NM_001286434.1 NM_153356.2 XM_006720409.3 XM_006720410.3 XM_006720411.3 XM_011521281.2 XM_011521282.2 XM_011521283.2 XM_011521284.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7203039 c.601A>G p.I201V Chr.15: 73886099-73886099 on Build GRCh38
6142788 c.628C>A p.L210I Chr.15: 73886126-73886126 on Build GRCh38
78614 c.664G>C p.A222P Chr.15: 73886162-73886162 on Build GRCh38
3969253 c.606C>T p.G202G Chr.15: 73886104-73886104 on Build GRCh38
4390973 c.594C>T p.V198V Chr.15: 73886092-73886092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002649248 Chr.15: 73886432-73886432 on Build GRCh38
rs780679185 Chr.15: 73886057-73886057 on Build GRCh38
rs1051292232 Chr.15: 73886351-73886351 on Build GRCh38
rs923864589 Chr.15: 73886312-73886312 on Build GRCh38
rs562102779 Chr.15: 73886006-73886006 on Build GRCh38
rs201373488 Chr.15: 73886125-73886125 on Build GRCh38
rs547001500 Chr.15: 73886446-73886446 on Build GRCh38
rs537307415 Chr.15: 73886208-73886208 on Build GRCh38
rs780953340 Chr.15: 73886337-73886337 on Build GRCh38
rs909947904 Chr.15: 73886028-73886028 on Build GRCh38
rs947929195 Chr.15: 73886369-73886369 on Build GRCh38
rs563502705 Chr.15: 73886288-73886288 on Build GRCh38
rs755193530 Chr.15: 73886146-73886146 on Build GRCh38
rs374781495 Chr.15: 73886205-73886205 on Build GRCh38
rs765152969 Chr.15: 73886042-73886043 on Build GRCh38
rs147253961 Chr.15: 73886026-73886026 on Build GRCh38
rs760722127 Chr.15: 73886214-73886214 on Build GRCh38
rs377763535 Chr.15: 73886039-73886039 on Build GRCh38
rs140701676 Chr.15: 73886106-73886106 on Build GRCh38
rs528475469 Chr.15: 73886390-73886390 on Build GRCh38
rs766388584 Chr.15: 73886129-73886129 on Build GRCh38
rs374114488 Chr.15: 73886027-73886027 on Build GRCh38
rs748654518 Chr.15: 73886047-73886047 on Build GRCh38
rs898492324 Chr.15: 73886424-73886424 on Build GRCh38
rs558823981 Chr.15: 73886210-73886210 on Build GRCh38
rs749484319 Chr.15: 73886041-73886041 on Build GRCh38
rs36015799 Chr.15: 73886261-73886261 on Build GRCh38
rs374896626 Chr.15: 73886123-73886123 on Build GRCh38
rs550648717 Chr.15: 73886249-73886249 on Build GRCh38
rs774064809 Chr.15: 73886097-73886097 on Build GRCh38
rs150883737 Chr.15: 73886155-73886155 on Build GRCh38
rs75887909 Chr.15: 73886450-73886450 on Build GRCh38
rs762545276 Chr.15: 73886076-73886076 on Build GRCh38
rs371117471 Chr.15: 73886070-73886070 on Build GRCh38
rs943074053 Chr.15: 73886278-73886278 on Build GRCh38
rs913801910 Chr.15: 73886259-73886259 on Build GRCh38
rs185802293 Chr.15: 73886086-73886086 on Build GRCh38
rs757187433 Chr.15: 73886339-73886339 on Build GRCh38
rs747540767 Chr.15: 73886184-73886184 on Build GRCh38
rs769255168 Chr.15: 73886201-73886201 on Build GRCh38
rs935262955 Chr.15: 73886331-73886331 on Build GRCh38
rs960519442 Chr.15: 73886245-73886245 on Build GRCh38
rs758673857 Chr.15: 73886206-73886206 on Build GRCh38
rs772629260 Chr.15: 73886049-73886049 on Build GRCh38
rs943146948 Chr.15: 73886078-73886078 on Build GRCh38
rs1034217389 Chr.15: 73886237-73886237 on Build GRCh38
rs1029914194 Chr.15: 73886128-73886128 on Build GRCh38
rs140135602 Chr.15: 73886259-73886259 on Build GRCh38
rs147641890 Chr.15: 73886354-73886354 on Build GRCh38
rs148679811 Chr.15: 73886377-73886377 on Build GRCh38
rs778198874 Chr.15: 73886170-73886170 on Build GRCh38
rs1017307438 Chr.15: 73886393-73886393 on Build GRCh38
rs980880244 Chr.15: 73886236-73886236 on Build GRCh38
rs576718488 Chr.15: 73886257-73886257 on Build GRCh38
rs148646352 Chr.15: 73886124-73886124 on Build GRCh38
rs765443450 Chr.15: 73886104-73886104 on Build GRCh38
rs369305607 Chr.15: 73886343-73886343 on Build GRCh38
rs767627863 Chr.15: 73886105-73886105 on Build GRCh38
rs922607641 Chr.15: 73886079-73886079 on Build GRCh38
rs750320687 Chr.15: 73886207-73886208 on Build GRCh38
rs753911975 Chr.15: 73886221-73886221 on Build GRCh38
rs1004474462 Chr.15: 73886383-73886383 on Build GRCh38
rs2278863 Chr.15: 73886204-73886204 on Build GRCh38
rs755317259 Chr.15: 73886043-73886043 on Build GRCh38
rs766446949 Chr.15: 73886218-73886218 on Build GRCh38
rs552859465 Chr.15: 73886019-73886019 on Build GRCh38
rs2278864 Chr.15: 73886264-73886264 on Build GRCh38
rs142399940 Chr.15: 73886080-73886080 on Build GRCh38
rs779172698 Chr.15: 73886046-73886046 on Build GRCh38
rs754047502 Chr.15: 73886133-73886133 on Build GRCh38
rs1044783607 Chr.15: 73886382-73886382 on Build GRCh38
rs139385848 Chr.15: 73886156-73886156 on Build GRCh38
rs531015025 Chr.15: 73886299-73886299 on Build GRCh38
rs887326393 Chr.15: 73886368-73886368 on Build GRCh38
rs191109987 Chr.15: 73886260-73886260 on Build GRCh38
rs745320997 Chr.15: 73886069-73886069 on Build GRCh38
rs115208108 Chr.15: 73886149-73886149 on Build GRCh38
rs759787639 Chr.15: 73886226-73886226 on Build GRCh38
rs960801560 Chr.15: 73886454-73886454 on Build GRCh38

View Full Product Details