Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 76169733-76170226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM266
Entrez Gene ID
123591
Gene Name
transmembrane protein 266
Gene Aliases
C15orf27, HVRP1
UniGene
Hs.661411
Gene Chromosome Location
Chr.15: 76059929-76204963 on Build GRCh38
Transcripts
NM_152335.2 XM_005254160.2 XM_017021915.1 XM_017021916.1 XM_017021917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4757719 c.502A>G p.N168D Chr.15: 76169861-76169861 on Build GRCh38
1939735 c.467G>A p.R156Q Chr.15: 76169826-76169826 on Build GRCh38
1939737 c.490G>A p.D164N Chr.15: 76169849-76169849 on Build GRCh38
1939739 c.499G>A p.E167K Chr.15: 76169858-76169858 on Build GRCh38
5770084 c.474G>A p.V158V Chr.15: 76169833-76169833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936183535 Chr.15: 76170117-76170117 on Build GRCh38
rs746167663 Chr.15: 76170042-76170042 on Build GRCh38
rs893554672 Chr.15: 76170115-76170115 on Build GRCh38
rs368635240 Chr.15: 76169857-76169857 on Build GRCh38
rs993255161 Chr.15: 76169975-76169975 on Build GRCh38
rs747850201 Chr.15: 76169883-76169883 on Build GRCh38
rs1000357241 Chr.15: 76169803-76169803 on Build GRCh38
rs374989948 Chr.15: 76169911-76169911 on Build GRCh38
rs771236844 Chr.15: 76170196-76170196 on Build GRCh38
rs200133901 Chr.15: 76169828-76169828 on Build GRCh38
rs201844390 Chr.15: 76169811-76169811 on Build GRCh38
rs372692444 Chr.15: 76169882-76169882 on Build GRCh38
rs987463973 Chr.15: 76169894-76169894 on Build GRCh38
rs990660200 Chr.15: 76169973-76169973 on Build GRCh38
rs770764763 Chr.15: 76169899-76169899 on Build GRCh38
rs973464272 Chr.15: 76169931-76169931 on Build GRCh38
rs779450367 Chr.15: 76170012-76170012 on Build GRCh38
rs781729675 Chr.15: 76169859-76169859 on Build GRCh38
rs74024102 Chr.15: 76169922-76169922 on Build GRCh38
rs776219021 Chr.15: 76169791-76169791 on Build GRCh38
rs34621040 Chr.15: 76170142-76170142 on Build GRCh38
rs1044533514 Chr.15: 76170045-76170045 on Build GRCh38
rs763123500 Chr.15: 76169923-76169923 on Build GRCh38
rs904604982 Chr.15: 76170090-76170090 on Build GRCh38
rs766367286 Chr.15: 76169772-76169772 on Build GRCh38
rs963222959 Chr.15: 76169937-76169945 on Build GRCh38
rs890164090 Chr.15: 76170190-76170190 on Build GRCh38
rs12902022 Chr.15: 76169945-76169945 on Build GRCh38
rs781300503 Chr.15: 76169921-76169924 on Build GRCh38
rs550805757 Chr.15: 76169810-76169810 on Build GRCh38
rs1017302222 Chr.15: 76169930-76169930 on Build GRCh38
rs759224230 Chr.15: 76169905-76169905 on Build GRCh38
rs920649563 Chr.15: 76169944-76169944 on Build GRCh38
rs1053251336 Chr.15: 76170118-76170118 on Build GRCh38
rs148177324 Chr.15: 76170205-76170205 on Build GRCh38
rs998410345 Chr.15: 76170130-76170130 on Build GRCh38
rs751672162 Chr.15: 76169855-76169855 on Build GRCh38
rs777374205 Chr.15: 76169893-76169893 on Build GRCh38
rs140678420 Chr.15: 76169765-76169765 on Build GRCh38
rs772436838 Chr.15: 76170199-76170199 on Build GRCh38
rs758453308 Chr.15: 76169833-76169833 on Build GRCh38
rs893974802 Chr.15: 76169768-76169768 on Build GRCh38
rs756567807 Chr.15: 76169878-76169878 on Build GRCh38
rs750740043 Chr.15: 76169873-76169873 on Build GRCh38
rs776701382 Chr.15: 76169901-76169901 on Build GRCh38
rs112075349 Chr.15: 76170021-76170021 on Build GRCh38
rs929208738 Chr.15: 76169951-76169951 on Build GRCh38
rs751638430 Chr.15: 76169899-76169901 on Build GRCh38
rs755037091 Chr.15: 76169904-76169905 on Build GRCh38
rs764113105 Chr.15: 76169854-76169854 on Build GRCh38
rs996785288 Chr.15: 76170124-76170124 on Build GRCh38
rs1053950370 Chr.15: 76170067-76170067 on Build GRCh38
rs760638246 Chr.15: 76169766-76169766 on Build GRCh38
rs1029699715 Chr.15: 76169887-76169887 on Build GRCh38
rs934950565 Chr.15: 76170055-76170055 on Build GRCh38
rs1014861989 Chr.15: 76169908-76169908 on Build GRCh38
rs949265601 Chr.15: 76170015-76170015 on Build GRCh38
rs771869828 Chr.15: 76169890-76169890 on Build GRCh38

View Full Product Details