Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 77032772-77033263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSTPIP1
Entrez Gene ID
9051
Gene Name
proline-serine-threonine phosphatase interacting protein 1
Gene Aliases
CD2BP1, CD2BP1L, CD2BP1S, H-PIP, PAPAS, PSTPIP
UniGene
Hs.129758
Gene Chromosome Location
Chr.15: 76994680-77037475 on Build GRCh38
Transcripts
NM_001321135.1 NM_001321136.1 NM_001321137.1 NM_003978.4 NR_135552.1 XM_006720737.2 XM_011522163.1 XM_011522164.1 XM_011522165.2 XM_011522166.1 XM_011522167.1 XM_011522168.2 XM_011522169.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6758004 c.895C>T p.P299S Chr.15: 77032918-77032918 on Build GRCh38
1740208 c.916G>A p.G306S Chr.15: 77032939-77032939 on Build GRCh38
6376244 c.891C>A p.S297R Chr.15: 77032914-77032914 on Build GRCh38
6143079 c.912C>A p.S304S Chr.15: 77032935-77032935 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758699821 Chr.15: 77032932-77032932 on Build GRCh38
rs765764833 Chr.15: 77032988-77032988 on Build GRCh38
rs572080370 Chr.15: 77033001-77033001 on Build GRCh38
rs769222897 Chr.15: 77032865-77032865 on Build GRCh38
rs928551320 Chr.15: 77033011-77033011 on Build GRCh38
rs1029185400 Chr.15: 77033068-77033068 on Build GRCh38
rs1045583382 Chr.15: 77032794-77032794 on Build GRCh38
rs759640462 Chr.15: 77032848-77032849 on Build GRCh38
rs753081335 Chr.15: 77032963-77032963 on Build GRCh38
rs774891463 Chr.15: 77032870-77032870 on Build GRCh38
rs1031711354 Chr.15: 77032902-77032902 on Build GRCh38
rs569685600 Chr.15: 77033186-77033186 on Build GRCh38
rs752909805 Chr.15: 77033003-77033005 on Build GRCh38
rs1026417755 Chr.15: 77033233-77033233 on Build GRCh38
rs774212289 Chr.15: 77032962-77032962 on Build GRCh38
rs199891584 Chr.15: 77032965-77032965 on Build GRCh38
rs993192071 Chr.15: 77033198-77033198 on Build GRCh38
rs377437961 Chr.15: 77032879-77032879 on Build GRCh38
rs753925976 Chr.15: 77032986-77032986 on Build GRCh38
rs139351566 Chr.15: 77032862-77032862 on Build GRCh38
rs774164456 Chr.15: 77032888-77032888 on Build GRCh38
rs906551321 Chr.15: 77032801-77032801 on Build GRCh38
rs902121637 Chr.15: 77033040-77033040 on Build GRCh38
rs11858480 Chr.15: 77032938-77032938 on Build GRCh38
rs768632688 Chr.15: 77032957-77032957 on Build GRCh38
rs770543515 Chr.15: 77032845-77032845 on Build GRCh38
rs767855367 Chr.15: 77032972-77032973 on Build GRCh38
rs753889391 Chr.15: 77032913-77032913 on Build GRCh38
rs889298084 Chr.15: 77033075-77033075 on Build GRCh38
rs538133422 Chr.15: 77032817-77032817 on Build GRCh38
rs757324764 Chr.15: 77032925-77032925 on Build GRCh38
rs755491182 Chr.15: 77032920-77032920 on Build GRCh38
rs370601019 Chr.15: 77032943-77032943 on Build GRCh38
rs941187986 Chr.15: 77032795-77032795 on Build GRCh38
rs542610612 Chr.15: 77033006-77033006 on Build GRCh38
rs946810210 Chr.15: 77033046-77033046 on Build GRCh38
rs748788047 Chr.15: 77032947-77032947 on Build GRCh38
rs748514164 Chr.15: 77032841-77032841 on Build GRCh38
rs532174837 Chr.15: 77033213-77033213 on Build GRCh38
rs566589576 Chr.15: 77033237-77033237 on Build GRCh38
rs761580488 Chr.15: 77032892-77032892 on Build GRCh38
rs756288702 Chr.15: 77033036-77033036 on Build GRCh38
rs141814002 Chr.15: 77033153-77033153 on Build GRCh38
rs779332305 Chr.15: 77032924-77032924 on Build GRCh38
rs764043431 Chr.15: 77032831-77032831 on Build GRCh38
rs186070824 Chr.15: 77033111-77033111 on Build GRCh38
rs181432186 Chr.15: 77033060-77033060 on Build GRCh38
rs374204996 Chr.15: 77032939-77032939 on Build GRCh38
rs1052851341 Chr.15: 77033035-77033035 on Build GRCh38
rs939345232 Chr.15: 77032829-77032829 on Build GRCh38
rs962901045 Chr.15: 77032946-77032947 on Build GRCh38
rs780092404 Chr.15: 77033155-77033155 on Build GRCh38
rs749738192 Chr.15: 77032830-77032830 on Build GRCh38
rs756411737 Chr.15: 77032904-77032904 on Build GRCh38
rs939377597 Chr.15: 77033025-77033025 on Build GRCh38
rs763986763 Chr.15: 77032887-77032887 on Build GRCh38
rs200796501 Chr.15: 77032905-77032905 on Build GRCh38
rs1011101484 Chr.15: 77032867-77032867 on Build GRCh38
rs756250919 Chr.15: 77033136-77033136 on Build GRCh38
rs776337082 Chr.15: 77032846-77032846 on Build GRCh38
rs773172907 Chr.15: 77032970-77032970 on Build GRCh38
rs189773500 Chr.15: 77032931-77032931 on Build GRCh38
rs372335370 Chr.15: 77032969-77032969 on Build GRCh38
rs1019175249 Chr.15: 77032891-77032891 on Build GRCh38
rs766561019 Chr.15: 77032980-77032980 on Build GRCh38
rs774539506 Chr.15: 77032837-77032837 on Build GRCh38
rs750616270 Chr.15: 77032896-77032896 on Build GRCh38
rs976066810 Chr.15: 77032927-77032927 on Build GRCh38
rs760932713 Chr.15: 77032973-77032973 on Build GRCh38
rs916273615 Chr.15: 77032979-77032979 on Build GRCh38
rs767839065 Chr.15: 77032893-77032893 on Build GRCh38
rs1016762327 Chr.15: 77033118-77033118 on Build GRCh38
rs779290323 Chr.15: 77032832-77032832 on Build GRCh38
rs551466220 Chr.15: 77033217-77033217 on Build GRCh38

View Full Product Details