Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 77054086-77054589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN3
Entrez Gene ID
10099
Gene Name
tetraspanin 3
Gene Aliases
TM4-A, TM4SF8, TSPAN-3
UniGene
Hs.744863
Gene Chromosome Location
Chr.15: 77044017-77071228 on Build GRCh38
Transcripts
NM_001168412.1 NM_005724.5 NM_198902.2 XM_017021857.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5920907 c.358C>T p.Q120* Chr.15: 77054252-77054252 on Build GRCh38
1194579 c.425A>G p.Q142R Chr.15: 77054185-77054185 on Build GRCh38
6452531 c.349C>T p.R117C Chr.15: 77054261-77054261 on Build GRCh38
965168 c.415G>T p.D139Y Chr.15: 77054195-77054195 on Build GRCh38
5428586 c.407G>A p.R136Q Chr.15: 77054203-77054203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755840827 Chr.15: 77054268-77054268 on Build GRCh38
rs919248455 Chr.15: 77054184-77054184 on Build GRCh38
rs796244933 Chr.15: 77054475-77054475 on Build GRCh38
rs994581585 Chr.15: 77054466-77054466 on Build GRCh38
rs778879875 Chr.15: 77054190-77054190 on Build GRCh38
rs140675272 Chr.15: 77054202-77054202 on Build GRCh38
rs868633216 Chr.15: 77054452-77054452 on Build GRCh38
rs962962006 Chr.15: 77054458-77054458 on Build GRCh38
rs1051237558 Chr.15: 77054402-77054403 on Build GRCh38
rs1011208141 Chr.15: 77054457-77054457 on Build GRCh38
rs915957108 Chr.15: 77054215-77054215 on Build GRCh38
rs979939005 Chr.15: 77054526-77054526 on Build GRCh38
rs1017623126 Chr.15: 77054454-77054455 on Build GRCh38
rs1052722395 Chr.15: 77054560-77054560 on Build GRCh38
rs948906072 Chr.15: 77054238-77054238 on Build GRCh38
rs755026849 Chr.15: 77054181-77054181 on Build GRCh38
rs114741584 Chr.15: 77054229-77054229 on Build GRCh38
rs1026867842 Chr.15: 77054507-77054507 on Build GRCh38
rs748648249 Chr.15: 77054194-77054194 on Build GRCh38
rs74025050 Chr.15: 77054316-77054316 on Build GRCh38
rs367861337 Chr.15: 77054426-77054426 on Build GRCh38
rs972637025 Chr.15: 77054506-77054506 on Build GRCh38
rs370307435 Chr.15: 77054230-77054230 on Build GRCh38
rs758956494 Chr.15: 77054295-77054295 on Build GRCh38
rs766173493 Chr.15: 77054279-77054279 on Build GRCh38
rs1049619493 Chr.15: 77054214-77054214 on Build GRCh38
rs575255150 Chr.15: 77054440-77054440 on Build GRCh38
rs150707893 Chr.15: 77054491-77054491 on Build GRCh38
rs762537436 Chr.15: 77054231-77054231 on Build GRCh38
rs773776897 Chr.15: 77054244-77054244 on Build GRCh38
rs750216980 Chr.15: 77054261-77054261 on Build GRCh38
rs748944453 Chr.15: 77054322-77054322 on Build GRCh38
rs181755548 Chr.15: 77054498-77054498 on Build GRCh38
rs751467918 Chr.15: 77054135-77054135 on Build GRCh38
rs747433636 Chr.15: 77054212-77054212 on Build GRCh38
rs1052691426 Chr.15: 77054270-77054270 on Build GRCh38
rs375306231 Chr.15: 77054260-77054260 on Build GRCh38
rs779019458 Chr.15: 77054292-77054292 on Build GRCh38
rs370865112 Chr.15: 77054139-77054139 on Build GRCh38
rs748222133 Chr.15: 77054293-77054293 on Build GRCh38
rs185158565 Chr.15: 77054216-77054216 on Build GRCh38
rs773899866 Chr.15: 77054323-77054323 on Build GRCh38
rs190061534 Chr.15: 77054481-77054481 on Build GRCh38
rs141151737 Chr.15: 77054305-77054305 on Build GRCh38
rs138918071 Chr.15: 77054198-77054198 on Build GRCh38
rs1052830646 Chr.15: 77054366-77054366 on Build GRCh38
rs952142291 Chr.15: 77054521-77054521 on Build GRCh38
rs572864603 Chr.15: 77054495-77054495 on Build GRCh38
rs529794502 Chr.15: 77054522-77054522 on Build GRCh38
rs374104987 Chr.15: 77054142-77054142 on Build GRCh38
rs370129453 Chr.15: 77054138-77054138 on Build GRCh38
rs930778087 Chr.15: 77054213-77054213 on Build GRCh38
rs573850596 Chr.15: 77054559-77054559 on Build GRCh38
rs755187283 Chr.15: 77054290-77054290 on Build GRCh38
rs776003005 Chr.15: 77054141-77054141 on Build GRCh38
rs771652570 Chr.15: 77054538-77054538 on Build GRCh38
rs754041303 Chr.15: 77054286-77054286 on Build GRCh38
rs948790538 Chr.15: 77054547-77054547 on Build GRCh38
rs368050463 Chr.15: 77054153-77054153 on Build GRCh38
rs777963582 Chr.15: 77054548-77054548 on Build GRCh38
rs747519464 Chr.15: 77054315-77054315 on Build GRCh38
rs1028494298 Chr.15: 77054472-77054472 on Build GRCh38
rs953752912 Chr.15: 77054485-77054485 on Build GRCh38
rs375258110 Chr.15: 77054242-77054242 on Build GRCh38
rs1044693599 Chr.15: 77054233-77054233 on Build GRCh38
rs780323506 Chr.15: 77054143-77054143 on Build GRCh38
rs997852804 Chr.15: 77054383-77054383 on Build GRCh38
rs892787283 Chr.15: 77054441-77054441 on Build GRCh38
rs773686080 Chr.15: 77054209-77054209 on Build GRCh38
rs377046351 Chr.15: 77054136-77054136 on Build GRCh38

View Full Product Details