Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78763621-78764131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS7
Entrez Gene ID
11173
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 7
Gene Aliases
ADAM-TS 7, ADAM-TS7, ADAMTS-7
UniGene
Hs.16441
Gene Chromosome Location
Chr.15: 78759203-78811464 on Build GRCh38
Transcripts
NM_014272.4 XM_005254137.3 XM_011521166.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4837991 c.4688G>A p.R1563Q Chr.15: 78763751-78763751 on Build GRCh38
4891298 c.4577C>A p.T1526K Chr.15: 78763942-78763942 on Build GRCh38
5662168 c.4666G>A p.A1556T Chr.15: 78763773-78763773 on Build GRCh38
5148571 c.4533T>A p.P1511P Chr.15: 78763986-78763986 on Build GRCh38
5182621 c.4611C>T p.G1537G Chr.15: 78763828-78763828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773406331 Chr.15: 78763691-78763691 on Build GRCh38
rs188808047 Chr.15: 78763726-78763726 on Build GRCh38
rs769694748 Chr.15: 78763978-78763978 on Build GRCh38
rs771411348 Chr.15: 78764095-78764095 on Build GRCh38
rs369677594 Chr.15: 78763828-78763828 on Build GRCh38
rs753022997 Chr.15: 78763988-78763988 on Build GRCh38
rs751182871 Chr.15: 78764028-78764028 on Build GRCh38
rs1012873436 Chr.15: 78763684-78763684 on Build GRCh38
rs112320482 Chr.15: 78763780-78763780 on Build GRCh38
rs760566157 Chr.15: 78763762-78763762 on Build GRCh38
rs75987215 Chr.15: 78763984-78763984 on Build GRCh38
rs950768420 Chr.15: 78763998-78763998 on Build GRCh38
rs763145560 Chr.15: 78763874-78763874 on Build GRCh38
rs370675358 Chr.15: 78763674-78763674 on Build GRCh38
rs746471624 Chr.15: 78763947-78763947 on Build GRCh38
rs773235448 Chr.15: 78764092-78764092 on Build GRCh38
rs760383235 Chr.15: 78763699-78763699 on Build GRCh38
rs368960389 Chr.15: 78764051-78764051 on Build GRCh38
rs371092119 Chr.15: 78763983-78763983 on Build GRCh38
rs765441225 Chr.15: 78763719-78763719 on Build GRCh38
rs779679275 Chr.15: 78763662-78763662 on Build GRCh38
rs368609643 Chr.15: 78763754-78763754 on Build GRCh38
rs1017826266 Chr.15: 78763956-78763956 on Build GRCh38
rs376434066 Chr.15: 78764003-78764003 on Build GRCh38
rs749040886 Chr.15: 78764056-78764056 on Build GRCh38
rs781329890 Chr.15: 78764036-78764036 on Build GRCh38
rs770819174 Chr.15: 78763771-78763771 on Build GRCh38
rs772623491 Chr.15: 78763989-78763989 on Build GRCh38
rs761829287 Chr.15: 78763685-78763685 on Build GRCh38
rs776529306 Chr.15: 78763779-78763779 on Build GRCh38
rs756816783 Chr.15: 78764032-78764032 on Build GRCh38
rs374374714 Chr.15: 78763692-78763692 on Build GRCh38
rs372662933 Chr.15: 78763913-78763913 on Build GRCh38
rs776582671 Chr.15: 78763854-78763854 on Build GRCh38
rs948628888 Chr.15: 78763839-78763839 on Build GRCh38
rs746952523 Chr.15: 78763840-78763840 on Build GRCh38
rs536028988 Chr.15: 78763714-78763714 on Build GRCh38
rs1031361195 Chr.15: 78763720-78763720 on Build GRCh38
rs754626021 Chr.15: 78763811-78763811 on Build GRCh38
rs917218693 Chr.15: 78764020-78764020 on Build GRCh38
rs759731435 Chr.15: 78763716-78763716 on Build GRCh38
rs368245832 Chr.15: 78763703-78763703 on Build GRCh38
rs773685492 Chr.15: 78763991-78763991 on Build GRCh38
rs572718001 Chr.15: 78763850-78763850 on Build GRCh38
rs377101323 Chr.15: 78763824-78763824 on Build GRCh38
rs201523294 Chr.15: 78763733-78763733 on Build GRCh38
rs768246977 Chr.15: 78763751-78763751 on Build GRCh38
rs374253167 Chr.15: 78764024-78764024 on Build GRCh38
rs777170764 Chr.15: 78763918-78763918 on Build GRCh38
rs531863929 Chr.15: 78763970-78763970 on Build GRCh38
rs768825105 Chr.15: 78764061-78764061 on Build GRCh38
rs774297669 Chr.15: 78763987-78763987 on Build GRCh38
rs748967677 Chr.15: 78763748-78763748 on Build GRCh38
rs372565775 Chr.15: 78764065-78764065 on Build GRCh38
rs545403957 Chr.15: 78763797-78763797 on Build GRCh38
rs756082675 Chr.15: 78764042-78764042 on Build GRCh38
rs199657348 Chr.15: 78763901-78763901 on Build GRCh38
rs200080080 Chr.15: 78763758-78763758 on Build GRCh38
rs764052505 Chr.15: 78763892-78763892 on Build GRCh38
rs575738937 Chr.15: 78763752-78763752 on Build GRCh38
rs370476096 Chr.15: 78763992-78763992 on Build GRCh38
rs754089412 Chr.15: 78763711-78763711 on Build GRCh38
rs770211743 Chr.15: 78763862-78763862 on Build GRCh38
rs371317043 Chr.15: 78764004-78764004 on Build GRCh38
rs751990791 Chr.15: 78763795-78763795 on Build GRCh38
rs144850032 Chr.15: 78764082-78764082 on Build GRCh38
rs777806197 Chr.15: 78763835-78763835 on Build GRCh38
rs773063450 Chr.15: 78763697-78763697 on Build GRCh38
rs543253106 Chr.15: 78763895-78763895 on Build GRCh38
rs60815800 Chr.15: 78763654-78763654 on Build GRCh38
rs781273311 Chr.15: 78763971-78763971 on Build GRCh38
rs765499328 Chr.15: 78763785-78763785 on Build GRCh38
rs745593012 Chr.15: 78763855-78763855 on Build GRCh38
rs889552598 Chr.15: 78764094-78764094 on Build GRCh38
rs762317795 Chr.15: 78763899-78763899 on Build GRCh38
rs191049501 Chr.15: 78763787-78763787 on Build GRCh38
rs764721937 Chr.15: 78763793-78763793 on Build GRCh38
rs374879285 Chr.15: 78764081-78764081 on Build GRCh38
rs986433581 Chr.15: 78763756-78763756 on Build GRCh38
rs929378235 Chr.15: 78763806-78763806 on Build GRCh38
rs747281992 Chr.15: 78763655-78763655 on Build GRCh38
rs376800676 Chr.15: 78763727-78763727 on Build GRCh38
rs769495131 Chr.15: 78763659-78763659 on Build GRCh38
rs773808314 Chr.15: 78763677-78763677 on Build GRCh38
rs778532019 Chr.15: 78763812-78763812 on Build GRCh38
rs750853487 Chr.15: 78763903-78763903 on Build GRCh38
rs768120415 Chr.15: 78763675-78763675 on Build GRCh38
rs775564536 Chr.15: 78763870-78763870 on Build GRCh38
rs779734325 Chr.15: 78763741-78763741 on Build GRCh38
rs754334628 Chr.15: 78763772-78763772 on Build GRCh38
rs376569624 Chr.15: 78763773-78763773 on Build GRCh38
rs548005559 Chr.15: 78764085-78764085 on Build GRCh38
rs754182510 Chr.15: 78764022-78764022 on Build GRCh38
rs758791224 Chr.15: 78763653-78763653 on Build GRCh38
rs750358699 Chr.15: 78764041-78764041 on Build GRCh38
rs1044612503 Chr.15: 78763688-78763688 on Build GRCh38
rs757552302 Chr.15: 78763735-78763735 on Build GRCh38
rs113911388 Chr.15: 78764109-78764109 on Build GRCh38
rs753226757 Chr.15: 78763652-78763652 on Build GRCh38
rs917289114 Chr.15: 78763837-78763837 on Build GRCh38
rs757739119 Chr.15: 78763796-78763796 on Build GRCh38
rs369865693 Chr.15: 78763912-78763912 on Build GRCh38
rs775825723 Chr.15: 78763979-78763979 on Build GRCh38
rs112367046 Chr.15: 78764079-78764079 on Build GRCh38
rs747543647 Chr.15: 78763753-78763753 on Build GRCh38
rs935925745 Chr.15: 78764044-78764044 on Build GRCh38
rs745717820 Chr.15: 78763976-78763976 on Build GRCh38

View Full Product Details