Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 79924236-79924737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST20-AS1
Entrez Gene ID
283687
Gene Name
ST20 antisense RNA 1
Gene Aliases
C15orf37
UniGene
Hs.722219
Gene Chromosome Location
Chr.15: 79922771-79924854 on Build GRCh38
Transcripts
NR_028330.1

SNP Detail

SNP ID Chromosome Location
rs1023055049 Chr.15: 79924440-79924440 on Build GRCh38
rs947106987 Chr.15: 79924561-79924561 on Build GRCh38
rs139148010 Chr.15: 79924529-79924529 on Build GRCh38
rs150206275 Chr.15: 79924336-79924336 on Build GRCh38
rs902832256 Chr.15: 79924579-79924579 on Build GRCh38
rs952735103 Chr.15: 79924482-79924482 on Build GRCh38
rs748229416 Chr.15: 79924685-79924685 on Build GRCh38
rs113565724 Chr.15: 79924304-79924304 on Build GRCh38
rs113295976 Chr.15: 79924556-79924556 on Build GRCh38
rs1013589478 Chr.15: 79924628-79924628 on Build GRCh38
rs984214233 Chr.15: 79924486-79924486 on Build GRCh38
rs1004129301 Chr.15: 79924348-79924348 on Build GRCh38
rs972378937 Chr.15: 79924518-79924518 on Build GRCh38
rs1042667358 Chr.15: 79924346-79924346 on Build GRCh38
rs902867195 Chr.15: 79924381-79924381 on Build GRCh38
rs1023107238 Chr.15: 79924653-79924653 on Build GRCh38
rs894858286 Chr.15: 79924421-79924421 on Build GRCh38
rs1005615848 Chr.15: 79924675-79924675 on Build GRCh38
rs961419902 Chr.15: 79924513-79924513 on Build GRCh38
rs572218899 Chr.15: 79924490-79924490 on Build GRCh38
rs34172478 Chr.15: 79924589-79924589 on Build GRCh38
rs113648400 Chr.15: 79924701-79924701 on Build GRCh38
rs117655788 Chr.15: 79924406-79924406 on Build GRCh38
rs999487607 Chr.15: 79924389-79924389 on Build GRCh38
rs117397172 Chr.15: 79924622-79924622 on Build GRCh38
rs755846903 Chr.15: 79924380-79924380 on Build GRCh38
rs558349122 Chr.15: 79924438-79924438 on Build GRCh38
rs755530158 Chr.15: 79924573-79924573 on Build GRCh38
rs966891392 Chr.15: 79924435-79924435 on Build GRCh38
rs1003582429 Chr.15: 79924403-79924403 on Build GRCh38
rs961667936 Chr.15: 79924696-79924696 on Build GRCh38
rs933146494 Chr.15: 79924540-79924540 on Build GRCh38
rs896111580 Chr.15: 79924626-79924626 on Build GRCh38
rs1020045251 Chr.15: 79924525-79924525 on Build GRCh38
rs1013120468 Chr.15: 79924430-79924430 on Build GRCh38
rs552374948 Chr.15: 79924694-79924694 on Build GRCh38
rs372346606 Chr.15: 79924425-79924425 on Build GRCh38
rs532190398 Chr.15: 79924662-79924662 on Build GRCh38
rs758009946 Chr.15: 79924678-79924678 on Build GRCh38
rs749741125 Chr.15: 79924551-79924551 on Build GRCh38
rs192251726 Chr.15: 79924546-79924546 on Build GRCh38

View Full Product Details