Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67826334-67826842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSNAXIP1
Entrez Gene ID
55815
Gene Name
translin associated factor X interacting protein 1
Gene Aliases
TXI1
UniGene
Hs.632212
Gene Chromosome Location
Chr.16: 67806573-67828068 on Build GRCh38
Transcripts
NM_001288990.1 NM_001288991.1 NM_001288992.1 NM_001288993.1 NM_001288994.1 NM_018430.3 XM_005256049.4 XM_011523230.2 XM_011523231.2 XM_011523232.2 XM_011523233.2 XM_011523234.2 XM_011523235.1 XM_011523236.2 XM_011523237.2 XM_011523238.1 XM_011523239.1 XM_011523240.1 XM_011523241.1 XM_011523242.1 XM_011523243.1 XM_017023445.1 XM_017023446.1 XM_017023447.1 XM_017023448.1 XM_017023449.1 XM_017023450.1 XM_017023451.1 XM_017023452.1 XM_017023453.1 XM_017023454.1 XM_017023455.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3511109 c.1240-1G>A p.? Chr.16: 67826691-67826691 on Build GRCh38
5907686 c.1272C>A p.F424L Chr.16: 67826724-67826724 on Build GRCh38
4736955 c.1340T>C p.I447T Chr.16: 67826792-67826792 on Build GRCh38
6230788 c.1275G>A p.L425L Chr.16: 67826727-67826727 on Build GRCh38
6555806 c.1305G>T p.M435I Chr.16: 67826757-67826757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771040096 Chr.16: 67826718-67826718 on Build GRCh38
rs758788310 Chr.16: 67826467-67826467 on Build GRCh38
rs1045287163 Chr.16: 67826445-67826445 on Build GRCh38
rs140928585 Chr.16: 67826769-67826769 on Build GRCh38
rs752377842 Chr.16: 67826566-67826566 on Build GRCh38
rs757662184 Chr.16: 67826388-67826388 on Build GRCh38
rs201851680 Chr.16: 67826455-67826455 on Build GRCh38
rs781677880 Chr.16: 67826516-67826516 on Build GRCh38
rs142018352 Chr.16: 67826545-67826545 on Build GRCh38
rs768084129 Chr.16: 67826670-67826670 on Build GRCh38
rs900883415 Chr.16: 67826643-67826643 on Build GRCh38
rs755091308 Chr.16: 67826689-67826689 on Build GRCh38
rs762498229 Chr.16: 67826641-67826641 on Build GRCh38
rs767416251 Chr.16: 67826386-67826386 on Build GRCh38
rs531496148 Chr.16: 67826632-67826632 on Build GRCh38
rs755814947 Chr.16: 67826658-67826658 on Build GRCh38
rs747359241 Chr.16: 67826424-67826424 on Build GRCh38
rs954620476 Chr.16: 67826665-67826665 on Build GRCh38
rs758108829 Chr.16: 67826567-67826567 on Build GRCh38
rs562641068 Chr.16: 67826512-67826512 on Build GRCh38
rs774292022 Chr.16: 67826585-67826585 on Build GRCh38
rs751604606 Chr.16: 67826391-67826391 on Build GRCh38
rs755888692 Chr.16: 67826393-67826393 on Build GRCh38
rs775141002 Chr.16: 67826732-67826732 on Build GRCh38
rs774541709 Chr.16: 67826538-67826538 on Build GRCh38
rs143251450 Chr.16: 67826786-67826786 on Build GRCh38
rs748128084 Chr.16: 67826699-67826699 on Build GRCh38
rs775863743 Chr.16: 67826521-67826521 on Build GRCh38
rs759339633 Chr.16: 67826559-67826559 on Build GRCh38
rs752145096 Chr.16: 67826497-67826497 on Build GRCh38
rs770318196 Chr.16: 67826571-67826571 on Build GRCh38
rs527597953 Chr.16: 67826682-67826682 on Build GRCh38
rs369795844 Chr.16: 67826473-67826473 on Build GRCh38
rs145603202 Chr.16: 67826656-67826657 on Build GRCh38
rs773596926 Chr.16: 67826423-67826423 on Build GRCh38
rs751136721 Chr.16: 67826568-67826568 on Build GRCh38
rs199912731 Chr.16: 67826410-67826410 on Build GRCh38
rs746298999 Chr.16: 67826518-67826518 on Build GRCh38
rs1031433539 Chr.16: 67826475-67826475 on Build GRCh38
rs201633703 Chr.16: 67826401-67826401 on Build GRCh38
rs777185795 Chr.16: 67826435-67826435 on Build GRCh38
rs530130741 Chr.16: 67826362-67826362 on Build GRCh38
rs770797255 Chr.16: 67826598-67826598 on Build GRCh38
rs759803003 Chr.16: 67826439-67826439 on Build GRCh38
rs746173850 Chr.16: 67826570-67826570 on Build GRCh38
rs754944781 Chr.16: 67826491-67826493 on Build GRCh38
rs980141775 Chr.16: 67826815-67826815 on Build GRCh38
rs754622023 Chr.16: 67826482-67826482 on Build GRCh38
rs770070970 Chr.16: 67826519-67826519 on Build GRCh38
rs761958399 Chr.16: 67826591-67826591 on Build GRCh38
rs766371939 Chr.16: 67826551-67826551 on Build GRCh38
rs62620177 Chr.16: 67826734-67826734 on Build GRCh38
rs780610103 Chr.16: 67826572-67826572 on Build GRCh38
rs1011424509 Chr.16: 67826535-67826535 on Build GRCh38
rs373723111 Chr.16: 67826691-67826691 on Build GRCh38
rs548407646 Chr.16: 67826569-67826569 on Build GRCh38
rs750963806 Chr.16: 67826655-67826655 on Build GRCh38
rs375423798 Chr.16: 67826600-67826600 on Build GRCh38
rs761601692 Chr.16: 67826752-67826752 on Build GRCh38
rs748370404 Chr.16: 67826806-67826806 on Build GRCh38
rs201166742 Chr.16: 67826573-67826573 on Build GRCh38
rs778302098 Chr.16: 67826504-67826504 on Build GRCh38
rs763142409 Chr.16: 67826456-67826456 on Build GRCh38
rs772571979 Chr.16: 67826418-67826418 on Build GRCh38
rs954331930 Chr.16: 67826706-67826706 on Build GRCh38
rs371481914 Chr.16: 67826391-67826391 on Build GRCh38
rs773319828 Chr.16: 67826705-67826705 on Build GRCh38
rs774070646 Chr.16: 67826748-67826748 on Build GRCh38
rs978967217 Chr.16: 67826730-67826730 on Build GRCh38
rs767257043 Chr.16: 67826503-67826505 on Build GRCh38
rs200022723 Chr.16: 67826601-67826601 on Build GRCh38
rs376691444 Chr.16: 67826412-67826412 on Build GRCh38
rs764821678 Chr.16: 67826565-67826565 on Build GRCh38
rs917246666 Chr.16: 67826808-67826808 on Build GRCh38
rs750933068 Chr.16: 67826681-67826681 on Build GRCh38
rs763871387 Chr.16: 67826654-67826654 on Build GRCh38
rs372904895 Chr.16: 67826419-67826419 on Build GRCh38
rs753564518 Chr.16: 67826556-67826556 on Build GRCh38
rs185289128 Chr.16: 67826369-67826369 on Build GRCh38
rs771222905 Chr.16: 67826425-67826425 on Build GRCh38
rs542030907 Chr.16: 67826365-67826365 on Build GRCh38
rs201849676 Chr.16: 67826805-67826805 on Build GRCh38
rs779704797 Chr.16: 67826400-67826400 on Build GRCh38
rs182917915 Chr.16: 67826688-67826688 on Build GRCh38
rs745781596 Chr.16: 67826708-67826708 on Build GRCh38
rs765625149 Chr.16: 67826443-67826443 on Build GRCh38
rs572428966 Chr.16: 67826385-67826385 on Build GRCh38
rs767116882 Chr.16: 67826757-67826757 on Build GRCh38
rs190354931 Chr.16: 67826687-67826687 on Build GRCh38
rs778542295 Chr.16: 67826486-67826486 on Build GRCh38
rs763052010 Chr.16: 67826528-67826528 on Build GRCh38
rs775402727 Chr.16: 67826604-67826604 on Build GRCh38
rs768590269 Chr.16: 67826529-67826529 on Build GRCh38
rs747714006 Chr.16: 67826417-67826417 on Build GRCh38
rs34938417 Chr.16: 67826702-67826703 on Build GRCh38
rs756801674 Chr.16: 67826656-67826656 on Build GRCh38
rs763591980 Chr.16: 67826735-67826735 on Build GRCh38
rs200207521 Chr.16: 67826772-67826772 on Build GRCh38
rs950036461 Chr.16: 67826813-67826813 on Build GRCh38
rs746010457 Chr.16: 67826811-67826812 on Build GRCh38
rs764984494 Chr.16: 67826603-67826603 on Build GRCh38
rs748469209 Chr.16: 67826466-67826466 on Build GRCh38
rs778203413 Chr.16: 67826498-67826498 on Build GRCh38
rs779208418 Chr.16: 67826664-67826667 on Build GRCh38
rs772170509 Chr.16: 67826700-67826700 on Build GRCh38
rs373698457 Chr.16: 67826481-67826481 on Build GRCh38
rs768983845 Chr.16: 67826575-67826575 on Build GRCh38
rs200583228 Chr.16: 67826468-67826468 on Build GRCh38

View Full Product Details