Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67943384-67943878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCAT
Entrez Gene ID
3931
Gene Name
lecithin-cholesterol acyltransferase
Gene Aliases
-
UniGene
Hs.387239
Gene Chromosome Location
Chr.16: 67939884-67944112 on Build GRCh38
Transcripts
NM_000229.1 NM_001145961.1 NM_001145962.1 NM_001145963.1 NM_001145964.1 NM_005072.4

Gene Symbol
SLC12A4
Entrez Gene ID
6560
Gene Name
solute carrier family 12 member 4
Gene Aliases
CTC-479C5.17, KCC1, hKCC1
UniGene
Hs.10094
Gene Chromosome Location
Chr.16: 67943474-67968694 on Build GRCh38
Transcripts
NM_000229.1 NM_001145961.1 NM_001145962.1 NM_001145963.1 NM_001145964.1 NM_005072.4

SNP Detail

SNP ID Chromosome Location
rs1053495903 Chr.16: 67943840-67943840 on Build GRCh38
rs558320445 Chr.16: 67943408-67943408 on Build GRCh38
rs780385119 Chr.16: 67943729-67943729 on Build GRCh38
rs970528832 Chr.16: 67943527-67943527 on Build GRCh38
rs909020010 Chr.16: 67943760-67943760 on Build GRCh38
rs560548730 Chr.16: 67943655-67943655 on Build GRCh38
rs979405275 Chr.16: 67943788-67943788 on Build GRCh38
rs1036297235 Chr.16: 67943484-67943484 on Build GRCh38
rs956137165 Chr.16: 67943744-67943744 on Build GRCh38
rs773773206 Chr.16: 67943738-67943738 on Build GRCh38
rs926619618 Chr.16: 67943734-67943734 on Build GRCh38
rs890852242 Chr.16: 67943811-67943811 on Build GRCh38
rs988785013 Chr.16: 67943717-67943717 on Build GRCh38
rs527645619 Chr.16: 67943665-67943665 on Build GRCh38
rs535764529 Chr.16: 67943780-67943780 on Build GRCh38
rs745557321 Chr.16: 67943731-67943731 on Build GRCh38
rs11860115 Chr.16: 67943793-67943793 on Build GRCh38
rs946682218 Chr.16: 67943650-67943650 on Build GRCh38
rs933239904 Chr.16: 67943802-67943802 on Build GRCh38
rs932387287 Chr.16: 67943708-67943708 on Build GRCh38
rs987951185 Chr.16: 67943618-67943619 on Build GRCh38
rs939148520 Chr.16: 67943584-67943584 on Build GRCh38
rs926732687 Chr.16: 67943415-67943415 on Build GRCh38
rs562803017 Chr.16: 67943494-67943494 on Build GRCh38
rs1050799711 Chr.16: 67943715-67943715 on Build GRCh38
rs1109166 Chr.16: 67943479-67943479 on Build GRCh38
rs1023176964 Chr.16: 67943847-67943847 on Build GRCh38
rs887612430 Chr.16: 67943458-67943458 on Build GRCh38
rs17287341 Chr.16: 67943485-67943485 on Build GRCh38
rs958244155 Chr.16: 67943710-67943710 on Build GRCh38
rs568288823 Chr.16: 67943735-67943735 on Build GRCh38
rs955560735 Chr.16: 67943433-67943433 on Build GRCh38
rs939781889 Chr.16: 67943812-67943812 on Build GRCh38
rs576994918 Chr.16: 67943809-67943809 on Build GRCh38
rs558657897 Chr.16: 67943805-67943805 on Build GRCh38
rs190807031 Chr.16: 67943849-67943849 on Build GRCh38
rs984224013 Chr.16: 67943736-67943736 on Build GRCh38
rs770268396 Chr.16: 67943576-67943576 on Build GRCh38
rs898703267 Chr.16: 67943834-67943834 on Build GRCh38
rs1015784357 Chr.16: 67943732-67943732 on Build GRCh38
rs117817502 Chr.16: 67943825-67943825 on Build GRCh38
rs1007232844 Chr.16: 67943827-67943827 on Build GRCh38
rs890883465 Chr.16: 67943730-67943731 on Build GRCh38
rs1022995405 Chr.16: 67943512-67943512 on Build GRCh38
rs996087507 Chr.16: 67943846-67943846 on Build GRCh38
rs1055603610 Chr.16: 67943446-67943446 on Build GRCh38
rs144922262 Chr.16: 67943445-67943445 on Build GRCh38
rs913759218 Chr.16: 67943763-67943763 on Build GRCh38
rs531821701 Chr.16: 67943696-67943696 on Build GRCh38
rs529994211 Chr.16: 67943616-67943616 on Build GRCh38
rs554132257 Chr.16: 67943781-67943781 on Build GRCh38
rs115102600 Chr.16: 67943682-67943682 on Build GRCh38
rs566469303 Chr.16: 67943784-67943784 on Build GRCh38
rs11860141 Chr.16: 67943638-67943638 on Build GRCh38
rs570849714 Chr.16: 67943695-67943695 on Build GRCh38
rs34686744 Chr.16: 67943405-67943405 on Build GRCh38
rs746600912 Chr.16: 67943530-67943530 on Build GRCh38
rs549941664 Chr.16: 67943724-67943724 on Build GRCh38
rs1021230108 Chr.16: 67943718-67943718 on Build GRCh38

View Full Product Details