Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70253827-70254320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AARS
Entrez Gene ID
16
Gene Name
alanyl-tRNA synthetase
Gene Aliases
CMT2N, EIEE29
UniGene
Hs.315137
Gene Chromosome Location
Chr.16: 70252295-70289552 on Build GRCh38
Transcripts
NM_001605.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3672316 c.2447G>T p.R816L Chr.16: 70253992-70253992 on Build GRCh38
4644011 c.2505C>T p.A835A Chr.16: 70253934-70253934 on Build GRCh38
1749736 c.2440G>A p.E814K Chr.16: 70253999-70253999 on Build GRCh38
7089602 c.2425C>A p.Q809K Chr.16: 70254014-70254014 on Build GRCh38
347873 c.2401G>T p.A801S Chr.16: 70254038-70254038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051958602 Chr.16: 70254231-70254231 on Build GRCh38
rs963738939 Chr.16: 70254107-70254107 on Build GRCh38
rs759350421 Chr.16: 70253890-70253891 on Build GRCh38
rs371610807 Chr.16: 70254214-70254215 on Build GRCh38
rs954886001 Chr.16: 70254076-70254076 on Build GRCh38
rs780744801 Chr.16: 70254003-70254003 on Build GRCh38
rs1020488953 Chr.16: 70254281-70254281 on Build GRCh38
rs947154175 Chr.16: 70254203-70254203 on Build GRCh38
rs749139777 Chr.16: 70254015-70254015 on Build GRCh38
rs942002710 Chr.16: 70254259-70254259 on Build GRCh38
rs533029846 Chr.16: 70253999-70253999 on Build GRCh38
rs375886345 Chr.16: 70254083-70254083 on Build GRCh38
rs1002928424 Chr.16: 70254297-70254297 on Build GRCh38
rs375717670 Chr.16: 70254150-70254150 on Build GRCh38
rs750973700 Chr.16: 70253994-70253994 on Build GRCh38
rs891381242 Chr.16: 70253910-70253910 on Build GRCh38
rs763386228 Chr.16: 70254053-70254053 on Build GRCh38
rs754103479 Chr.16: 70253977-70253977 on Build GRCh38
rs897597773 Chr.16: 70253863-70253863 on Build GRCh38
rs903350828 Chr.16: 70254284-70254284 on Build GRCh38
rs745577157 Chr.16: 70253931-70253931 on Build GRCh38
rs892794489 Chr.16: 70254198-70254198 on Build GRCh38
rs377576408 Chr.16: 70254049-70254049 on Build GRCh38
rs754119012 Chr.16: 70253971-70253971 on Build GRCh38
rs530560296 Chr.16: 70254165-70254165 on Build GRCh38
rs371962301 Chr.16: 70253850-70253850 on Build GRCh38
rs746557071 Chr.16: 70253899-70253899 on Build GRCh38
rs1041532704 Chr.16: 70253895-70253895 on Build GRCh38
rs759906749 Chr.16: 70253979-70253979 on Build GRCh38
rs775304067 Chr.16: 70253936-70253936 on Build GRCh38
rs991338877 Chr.16: 70254181-70254181 on Build GRCh38
rs758988205 Chr.16: 70253852-70253852 on Build GRCh38
rs933124241 Chr.16: 70254224-70254224 on Build GRCh38
rs76446904 Chr.16: 70254028-70254028 on Build GRCh38
rs573301270 Chr.16: 70254190-70254190 on Build GRCh38
rs372552067 Chr.16: 70254037-70254037 on Build GRCh38
rs893420314 Chr.16: 70254275-70254275 on Build GRCh38
rs776276100 Chr.16: 70253920-70253920 on Build GRCh38
rs752099746 Chr.16: 70254066-70254066 on Build GRCh38
rs746662354 Chr.16: 70254278-70254278 on Build GRCh38
rs761717573 Chr.16: 70254027-70254027 on Build GRCh38
rs1007113248 Chr.16: 70253911-70253911 on Build GRCh38
rs745364673 Chr.16: 70254116-70254116 on Build GRCh38
rs1018549676 Chr.16: 70253917-70253917 on Build GRCh38
rs563232031 Chr.16: 70254101-70254101 on Build GRCh38
rs766300063 Chr.16: 70253846-70253847 on Build GRCh38
rs986362065 Chr.16: 70254086-70254086 on Build GRCh38
rs768412428 Chr.16: 70254018-70254018 on Build GRCh38
rs914285635 Chr.16: 70254135-70254135 on Build GRCh38
rs1038125515 Chr.16: 70254266-70254266 on Build GRCh38
rs777095182 Chr.16: 70253885-70253885 on Build GRCh38
rs757366070 Chr.16: 70253987-70253988 on Build GRCh38
rs147319762 Chr.16: 70253980-70253980 on Build GRCh38
rs751391874 Chr.16: 70253847-70253847 on Build GRCh38
rs1035654571 Chr.16: 70254108-70254108 on Build GRCh38
rs752933204 Chr.16: 70254052-70254052 on Build GRCh38
rs779777115 Chr.16: 70254014-70254014 on Build GRCh38
rs762777733 Chr.16: 70253947-70253947 on Build GRCh38
rs757800600 Chr.16: 70254070-70254070 on Build GRCh38
rs760816488 Chr.16: 70253954-70253954 on Build GRCh38
rs745489057 Chr.16: 70254006-70254006 on Build GRCh38
rs1039959129 Chr.16: 70254269-70254269 on Build GRCh38
rs1030519185 Chr.16: 70254057-70254057 on Build GRCh38
rs888754683 Chr.16: 70254248-70254248 on Build GRCh38
rs778478316 Chr.16: 70253993-70253993 on Build GRCh38
rs890624784 Chr.16: 70254282-70254282 on Build GRCh38
rs542357096 Chr.16: 70254258-70254258 on Build GRCh38
rs541254641 Chr.16: 70253856-70253856 on Build GRCh38
rs185732910 Chr.16: 70254184-70254184 on Build GRCh38
rs753158346 Chr.16: 70253986-70253986 on Build GRCh38
rs918994163 Chr.16: 70254218-70254218 on Build GRCh38
rs372138128 Chr.16: 70254054-70254054 on Build GRCh38
rs767164973 Chr.16: 70253957-70253958 on Build GRCh38
rs958709464 Chr.16: 70254111-70254111 on Build GRCh38
rs771133578 Chr.16: 70254036-70254036 on Build GRCh38
rs865779159 Chr.16: 70254025-70254025 on Build GRCh38
rs971951938 Chr.16: 70254217-70254217 on Build GRCh38
rs751129112 Chr.16: 70253982-70253983 on Build GRCh38
rs192447854 Chr.16: 70253851-70253851 on Build GRCh38
rs771519250 Chr.16: 70254159-70254159 on Build GRCh38
rs766651955 Chr.16: 70253963-70253963 on Build GRCh38
rs769571173 Chr.16: 70253934-70253934 on Build GRCh38
rs747782282 Chr.16: 70254011-70254011 on Build GRCh38
rs540218496 Chr.16: 70253865-70253865 on Build GRCh38
rs376619331 Chr.16: 70253900-70253900 on Build GRCh38
rs758787063 Chr.16: 70253991-70253991 on Build GRCh38
rs749957123 Chr.16: 70254088-70254088 on Build GRCh38

View Full Product Details