Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35934292-35934812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYZL6
Entrez Gene ID
57151
Gene Name
lysozyme like 6
Gene Aliases
HEL-S-6a, LYC1, LYZB, PRO1485, TKAL754, UNQ754
UniGene
Hs.97477
Gene Chromosome Location
Chr.17: 35934518-35943710 on Build GRCh38
Transcripts
NM_001199951.2 NM_020426.3

SNP Detail

SNP ID Chromosome Location
rs746788627 Chr.17: 35934578-35934578 on Build GRCh38
rs985576218 Chr.17: 35934526-35934526 on Build GRCh38
rs770851218 Chr.17: 35934765-35934765 on Build GRCh38
rs1040902609 Chr.17: 35934646-35934646 on Build GRCh38
rs146771787 Chr.17: 35934784-35934784 on Build GRCh38
rs773044107 Chr.17: 35934554-35934554 on Build GRCh38
rs903886084 Chr.17: 35934688-35934688 on Build GRCh38
rs931050342 Chr.17: 35934528-35934528 on Build GRCh38
rs745380551 Chr.17: 35934779-35934779 on Build GRCh38
rs918137307 Chr.17: 35934363-35934363 on Build GRCh38
rs2242596 Chr.17: 35934743-35934743 on Build GRCh38
rs36082813 Chr.17: 35934373-35934374 on Build GRCh38
rs778018131 Chr.17: 35934703-35934703 on Build GRCh38
rs140946550 Chr.17: 35934707-35934708 on Build GRCh38
rs770045882 Chr.17: 35934657-35934657 on Build GRCh38
rs976996285 Chr.17: 35934570-35934570 on Build GRCh38
rs182752125 Chr.17: 35934770-35934770 on Build GRCh38
rs544855705 Chr.17: 35934430-35934430 on Build GRCh38
rs368056415 Chr.17: 35934783-35934783 on Build GRCh38
rs758323863 Chr.17: 35934498-35934498 on Build GRCh38
rs572549745 Chr.17: 35934345-35934345 on Build GRCh38
rs758279829 Chr.17: 35934749-35934749 on Build GRCh38
rs544055007 Chr.17: 35934679-35934679 on Build GRCh38
rs767164812 Chr.17: 35934777-35934777 on Build GRCh38
rs779885847 Chr.17: 35934508-35934508 on Build GRCh38
rs116190619 Chr.17: 35934557-35934557 on Build GRCh38
rs529361389 Chr.17: 35934691-35934691 on Build GRCh38
rs919628058 Chr.17: 35934527-35934527 on Build GRCh38
rs961364136 Chr.17: 35934358-35934358 on Build GRCh38
rs752708655 Chr.17: 35934791-35934792 on Build GRCh38
rs1010188006 Chr.17: 35934672-35934672 on Build GRCh38
rs865882412 Chr.17: 35934648-35934648 on Build GRCh38
rs386796782 Chr.17: 35934784-35934786 on Build GRCh38
rs796158743 Chr.17: 35934699-35934699 on Build GRCh38
rs889612513 Chr.17: 35934544-35934544 on Build GRCh38
rs561172276 Chr.17: 35934686-35934686 on Build GRCh38
rs750305761 Chr.17: 35934366-35934366 on Build GRCh38
rs1026967199 Chr.17: 35934785-35934785 on Build GRCh38
rs879487209 Chr.17: 35934641-35934641 on Build GRCh38
rs994617600 Chr.17: 35934374-35934374 on Build GRCh38
rs778726776 Chr.17: 35934341-35934341 on Build GRCh38
rs753319101 Chr.17: 35934318-35934318 on Build GRCh38
rs769494439 Chr.17: 35934781-35934781 on Build GRCh38
rs530703843 Chr.17: 35934665-35934665 on Build GRCh38
rs566036022 Chr.17: 35934759-35934759 on Build GRCh38
rs868674195 Chr.17: 35934562-35934562 on Build GRCh38
rs759300555 Chr.17: 35934756-35934756 on Build GRCh38
rs777643606 Chr.17: 35934751-35934751 on Build GRCh38
rs140518522 Chr.17: 35934786-35934786 on Build GRCh38
rs992693900 Chr.17: 35934752-35934752 on Build GRCh38
rs1025146327 Chr.17: 35934780-35934780 on Build GRCh38
rs1027019391 Chr.17: 35934419-35934419 on Build GRCh38
rs945727385 Chr.17: 35934618-35934618 on Build GRCh38
rs972778989 Chr.17: 35934522-35934522 on Build GRCh38
rs1049530502 Chr.17: 35934540-35934540 on Build GRCh38
rs913476682 Chr.17: 35934613-35934613 on Build GRCh38
rs1020908291 Chr.17: 35934677-35934677 on Build GRCh38
rs1033694527 Chr.17: 35934719-35934719 on Build GRCh38

View Full Product Details