Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36064198-36064705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL18
Entrez Gene ID
6362
Gene Name
C-C motif chemokine ligand 18
Gene Aliases
AMAC-1, AMAC1, CKb7, DC-CK1, DCCK1, MIP-4, PARC, SCYA18
UniGene
Hs.143961
Gene Chromosome Location
Chr.17: 36064272-36071481 on Build GRCh38
Transcripts
NM_002988.3

SNP Detail

SNP ID Chromosome Location
rs772326996 Chr.17: 36064443-36064443 on Build GRCh38
rs374115816 Chr.17: 36064355-36064355 on Build GRCh38
rs966545588 Chr.17: 36064511-36064511 on Build GRCh38
rs760354932 Chr.17: 36064448-36064449 on Build GRCh38
rs878943325 Chr.17: 36064356-36064356 on Build GRCh38
rs748378437 Chr.17: 36064334-36064334 on Build GRCh38
rs572759641 Chr.17: 36064242-36064242 on Build GRCh38
rs1005922208 Chr.17: 36064407-36064407 on Build GRCh38
rs77975917 Chr.17: 36064660-36064660 on Build GRCh38
rs957941730 Chr.17: 36064607-36064607 on Build GRCh38
rs915555656 Chr.17: 36064267-36064267 on Build GRCh38
rs948452770 Chr.17: 36064272-36064272 on Build GRCh38
rs765174314 Chr.17: 36064351-36064351 on Build GRCh38
rs528490668 Chr.17: 36064493-36064493 on Build GRCh38
rs907348092 Chr.17: 36064282-36064282 on Build GRCh38
rs551626931 Chr.17: 36064509-36064509 on Build GRCh38
rs768658429 Chr.17: 36064518-36064518 on Build GRCh38
rs773250744 Chr.17: 36064446-36064446 on Build GRCh38
rs79020437 Chr.17: 36064630-36064630 on Build GRCh38
rs1032260335 Chr.17: 36064592-36064592 on Build GRCh38
rs762858470 Chr.17: 36064459-36064459 on Build GRCh38
rs749382552 Chr.17: 36064316-36064316 on Build GRCh38
rs2015086 Chr.17: 36064257-36064257 on Build GRCh38
rs376657736 Chr.17: 36064445-36064445 on Build GRCh38
rs955437489 Chr.17: 36064505-36064505 on Build GRCh38
rs759527592 Chr.17: 36064349-36064349 on Build GRCh38
rs558031922 Chr.17: 36064348-36064348 on Build GRCh38
rs558912524 Chr.17: 36064299-36064299 on Build GRCh38
rs970187640 Chr.17: 36064639-36064639 on Build GRCh38
rs749759747 Chr.17: 36064243-36064243 on Build GRCh38
rs770672987 Chr.17: 36064345-36064345 on Build GRCh38
rs775421030 Chr.17: 36064348-36064349 on Build GRCh38
rs369599857 Chr.17: 36064372-36064372 on Build GRCh38
rs559655581 Chr.17: 36064489-36064489 on Build GRCh38
rs73290772 Chr.17: 36064361-36064361 on Build GRCh38
rs768815599 Chr.17: 36064326-36064326 on Build GRCh38
rs745668927 Chr.17: 36064452-36064452 on Build GRCh38
rs750809251 Chr.17: 36064398-36064398 on Build GRCh38
rs762485548 Chr.17: 36064358-36064358 on Build GRCh38
rs199892163 Chr.17: 36064426-36064426 on Build GRCh38
rs769541673 Chr.17: 36064453-36064453 on Build GRCh38
rs780645213 Chr.17: 36064309-36064309 on Build GRCh38
rs957875264 Chr.17: 36064555-36064555 on Build GRCh38
rs566434732 Chr.17: 36064681-36064681 on Build GRCh38
rs749763771 Chr.17: 36064416-36064416 on Build GRCh38
rs906944226 Chr.17: 36064340-36064340 on Build GRCh38
rs774926948 Chr.17: 36064292-36064292 on Build GRCh38
rs375607603 Chr.17: 36064477-36064477 on Build GRCh38
rs767827152 Chr.17: 36064376-36064376 on Build GRCh38
rs377014840 Chr.17: 36064460-36064460 on Build GRCh38
rs369307586 Chr.17: 36064600-36064600 on Build GRCh38
rs996792947 Chr.17: 36064469-36064469 on Build GRCh38
rs1006555907 Chr.17: 36064662-36064662 on Build GRCh38
rs756926531 Chr.17: 36064375-36064375 on Build GRCh38
rs774457234 Chr.17: 36064333-36064333 on Build GRCh38
rs748387518 Chr.17: 36064441-36064441 on Build GRCh38
rs115039704 Chr.17: 36064673-36064673 on Build GRCh38
rs544452516 Chr.17: 36064405-36064405 on Build GRCh38
rs929482770 Chr.17: 36064666-36064666 on Build GRCh38
rs750940416 Chr.17: 36064659-36064659 on Build GRCh38
rs2015070 Chr.17: 36064423-36064423 on Build GRCh38
rs543650065 Chr.17: 36064566-36064566 on Build GRCh38
rs530988926 Chr.17: 36064618-36064618 on Build GRCh38
rs377682562 Chr.17: 36064362-36064362 on Build GRCh38
rs2015052 Chr.17: 36064653-36064653 on Build GRCh38
rs936253723 Chr.17: 36064227-36064227 on Build GRCh38
rs867915111 Chr.17: 36064454-36064454 on Build GRCh38
rs372574777 Chr.17: 36064458-36064458 on Build GRCh38

View Full Product Details