Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73231998-73232505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf80
Entrez Gene ID
55028
Gene Name
chromosome 17 open reading frame 80
Gene Aliases
HLC-8, MIG3, SPEP1
UniGene
Hs.12929
Gene Chromosome Location
Chr.17: 73232233-73248959 on Build GRCh38
Transcripts
NM_001288770.1 NM_001288771.1 XM_005257487.3 NM_001098832.1 NM_001289410.1 NM_001289411.1 NM_032837.2

Gene Symbol
FAM104A
Entrez Gene ID
84923
Gene Name
family with sequence similarity 104 member A
Gene Aliases
-
UniGene
Hs.741276
Gene Chromosome Location
Chr.17: 73207353-73232394 on Build GRCh38
Transcripts
NM_001288770.1 NM_001288771.1 XM_005257487.3 NM_001098832.1 NM_001289410.1 NM_001289411.1 NM_032837.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1385681 c.213C>G p.L71L Chr.17: 73232094-73232094 on Build GRCh38
6317509 c.167C>A p.A56E Chr.17: 73232140-73232140 on Build GRCh38
1385683 c.208C>T p.R70C Chr.17: 73232099-73232099 on Build GRCh38
296573 c.9_10insG p.R4fs*8 Chr.17: 73232297-73232298 on Build GRCh38
6370944 c.47C>T p.P16L Chr.17: 73232260-73232260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959697233 Chr.17: 73232423-73232423 on Build GRCh38
rs749399643 Chr.17: 73232072-73232072 on Build GRCh38
rs544913626 Chr.17: 73232359-73232359 on Build GRCh38
rs941287240 Chr.17: 73232291-73232291 on Build GRCh38
rs988568802 Chr.17: 73232306-73232306 on Build GRCh38
rs767339351 Chr.17: 73232178-73232178 on Build GRCh38
rs377066963 Chr.17: 73232363-73232363 on Build GRCh38
rs377592119 Chr.17: 73232304-73232304 on Build GRCh38
rs755185778 Chr.17: 73232302-73232302 on Build GRCh38
rs377722890 Chr.17: 73232245-73232245 on Build GRCh38
rs748324805 Chr.17: 73232269-73232269 on Build GRCh38
rs773744672 Chr.17: 73232265-73232265 on Build GRCh38
rs775189353 Chr.17: 73232319-73232319 on Build GRCh38
rs774414396 Chr.17: 73232078-73232078 on Build GRCh38
rs928402948 Chr.17: 73232146-73232146 on Build GRCh38
rs983614465 Chr.17: 73232080-73232080 on Build GRCh38
rs560347523 Chr.17: 73232361-73232361 on Build GRCh38
rs375753271 Chr.17: 73232237-73232237 on Build GRCh38
rs746061460 Chr.17: 73232156-73232156 on Build GRCh38
rs370554488 Chr.17: 73232309-73232309 on Build GRCh38
rs758485651 Chr.17: 73232256-73232256 on Build GRCh38
rs772316149 Chr.17: 73232088-73232088 on Build GRCh38
rs995050294 Chr.17: 73232369-73232369 on Build GRCh38
rs150645939 Chr.17: 73232100-73232100 on Build GRCh38
rs367575990 Chr.17: 73232327-73232327 on Build GRCh38
rs754844655 Chr.17: 73232142-73232142 on Build GRCh38
rs777259359 Chr.17: 73232164-73232164 on Build GRCh38
rs539692494 Chr.17: 73232159-73232159 on Build GRCh38
rs761203220 Chr.17: 73232090-73232090 on Build GRCh38
rs200728518 Chr.17: 73232322-73232322 on Build GRCh38
rs760809940 Chr.17: 73232444-73232445 on Build GRCh38
rs753786628 Chr.17: 73232293-73232293 on Build GRCh38
rs780412367 Chr.17: 73232141-73232141 on Build GRCh38
rs146728336 Chr.17: 73232163-73232163 on Build GRCh38
rs745962111 Chr.17: 73232201-73232201 on Build GRCh38
rs760537927 Chr.17: 73232286-73232286 on Build GRCh38
rs375022322 Chr.17: 73232171-73232171 on Build GRCh38
rs765855105 Chr.17: 73232242-73232242 on Build GRCh38
rs770348285 Chr.17: 73232232-73232232 on Build GRCh38
rs767157414 Chr.17: 73232282-73232282 on Build GRCh38
rs375573521 Chr.17: 73232093-73232093 on Build GRCh38
rs201327693 Chr.17: 73232298-73232298 on Build GRCh38
rs750007517 Chr.17: 73232165-73232165 on Build GRCh38
rs141873101 Chr.17: 73232089-73232089 on Build GRCh38
rs750016710 Chr.17: 73232119-73232119 on Build GRCh38
rs199818438 Chr.17: 73232261-73232261 on Build GRCh38
rs749798858 Chr.17: 73232266-73232266 on Build GRCh38
rs770885253 Chr.17: 73232154-73232154 on Build GRCh38
rs777774613 Chr.17: 73232051-73232051 on Build GRCh38
rs763111693 Chr.17: 73232322-73232323 on Build GRCh38
rs1051606230 Chr.17: 73232355-73232355 on Build GRCh38
rs755934111 Chr.17: 73232248-73232248 on Build GRCh38
rs201703597 Chr.17: 73232331-73232331 on Build GRCh38
rs756130514 Chr.17: 73232188-73232188 on Build GRCh38
rs759126875 Chr.17: 73232279-73232279 on Build GRCh38
rs996228609 Chr.17: 73232228-73232228 on Build GRCh38
rs752498500 Chr.17: 73232247-73232247 on Build GRCh38
rs747449032 Chr.17: 73232194-73232194 on Build GRCh38
rs369388162 Chr.17: 73232116-73232116 on Build GRCh38
rs773493866 Chr.17: 73232298-73232298 on Build GRCh38
rs762457464 Chr.17: 73232166-73232166 on Build GRCh38
rs188013285 Chr.17: 73232075-73232075 on Build GRCh38
rs375693359 Chr.17: 73232337-73232337 on Build GRCh38
rs781642632 Chr.17: 73232263-73232263 on Build GRCh38
rs202088358 Chr.17: 73232083-73232083 on Build GRCh38
rs911372207 Chr.17: 73232077-73232077 on Build GRCh38
rs1047043142 Chr.17: 73232134-73232134 on Build GRCh38
rs770953647 Chr.17: 73232332-73232333 on Build GRCh38
rs770000277 Chr.17: 73232297-73232298 on Build GRCh38
rs761753641 Chr.17: 73232329-73232329 on Build GRCh38
rs924225684 Chr.17: 73232035-73232035 on Build GRCh38
rs777206174 Chr.17: 73232224-73232224 on Build GRCh38
rs755310458 Chr.17: 73232260-73232260 on Build GRCh38
rs1030278223 Chr.17: 73232233-73232233 on Build GRCh38
rs1015027122 Chr.17: 73232082-73232082 on Build GRCh38
rs750423203 Chr.17: 73232251-73232251 on Build GRCh38
rs896411499 Chr.17: 73232193-73232193 on Build GRCh38
rs775222018 Chr.17: 73232284-73232284 on Build GRCh38
rs1041073947 Chr.17: 73232086-73232086 on Build GRCh38
rs770422960 Chr.17: 73232274-73232274 on Build GRCh38
rs113787330 Chr.17: 73232221-73232221 on Build GRCh38
rs1035237612 Chr.17: 73232419-73232419 on Build GRCh38
rs745410064 Chr.17: 73232312-73232312 on Build GRCh38
rs1003811928 Chr.17: 73232407-73232407 on Build GRCh38
rs76917873 Chr.17: 73232479-73232479 on Build GRCh38
rs949929969 Chr.17: 73232447-73232447 on Build GRCh38
rs751651372 Chr.17: 73232336-73232336 on Build GRCh38
rs371082623 Chr.17: 73232249-73232249 on Build GRCh38
rs768870080 Chr.17: 73232205-73232206 on Build GRCh38
rs569512877 Chr.17: 73232152-73232152 on Build GRCh38
rs548215808 Chr.17: 73232300-73232300 on Build GRCh38
rs773959760 Chr.17: 73232244-73232244 on Build GRCh38
rs899242254 Chr.17: 73232366-73232366 on Build GRCh38
rs780448847 Chr.17: 73232217-73232217 on Build GRCh38
rs1042981933 Chr.17: 73232214-73232214 on Build GRCh38
rs751702545 Chr.17: 73232299-73232299 on Build GRCh38
rs764479072 Chr.17: 73232464-73232464 on Build GRCh38
rs766061910 Chr.17: 73232168-73232168 on Build GRCh38
rs769155305 Chr.17: 73232257-73232257 on Build GRCh38
rs969689298 Chr.17: 73232404-73232404 on Build GRCh38
rs761434261 Chr.17: 73232111-73232111 on Build GRCh38
rs779701151 Chr.17: 73232133-73232133 on Build GRCh38
rs373736498 Chr.17: 73232313-73232313 on Build GRCh38
rs909878911 Chr.17: 73232416-73232416 on Build GRCh38
rs754744268 Chr.17: 73232176-73232176 on Build GRCh38
rs3764363 Chr.17: 73232039-73232039 on Build GRCh38
rs748131479 Chr.17: 73232153-73232153 on Build GRCh38
rs941411988 Chr.17: 73232430-73232430 on Build GRCh38
rs761727566 Chr.17: 73232296-73232296 on Build GRCh38
rs180917408 Chr.17: 73232240-73232240 on Build GRCh38
rs779115552 Chr.17: 73232155-73232155 on Build GRCh38
rs544578624 Chr.17: 73232303-73232303 on Build GRCh38
rs970035327 Chr.17: 73232294-73232294 on Build GRCh38
rs369159410 Chr.17: 73232220-73232220 on Build GRCh38
rs747286426 Chr.17: 73232218-73232218 on Build GRCh38
rs549066550 Chr.17: 73232396-73232396 on Build GRCh38
rs758967246 Chr.17: 73232131-73232132 on Build GRCh38
rs753994928 Chr.17: 73232480-73232480 on Build GRCh38
rs573011340 Chr.17: 73232185-73232185 on Build GRCh38
rs112550532 Chr.17: 73232020-73232020 on Build GRCh38
rs781506110 Chr.17: 73232262-73232270 on Build GRCh38
rs748653682 Chr.17: 73232225-73232225 on Build GRCh38
rs758505668 Chr.17: 73232208-73232208 on Build GRCh38
rs527407008 Chr.17: 73232368-73232368 on Build GRCh38
rs888388577 Chr.17: 73232138-73232138 on Build GRCh38
rs778796342 Chr.17: 73232194-73232194 on Build GRCh38
rs988474005 Chr.17: 73232445-73232445 on Build GRCh38

View Full Product Details