Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75109903-75110391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC7
Entrez Gene ID
79637
Gene Name
armadillo repeat containing 7
Gene Aliases
-
UniGene
Hs.413047
Gene Chromosome Location
Chr.17: 75109952-75130265 on Build GRCh38
Transcripts
NM_001304271.1 NM_001304272.1 NM_001304273.1 NM_024585.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7181090 c.15G>A p.P5P Chr.17: 75110303-75110303 on Build GRCh38
1385924 c.50T>C p.L17P Chr.17: 75110338-75110338 on Build GRCh38
7277181 c.91G>A p.D31N Chr.17: 75110379-75110379 on Build GRCh38
6317511 c.39G>T p.R13R Chr.17: 75110327-75110327 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs144858776 Chr.17: 75110306-75110306 on Build GRCh38
rs758453579 Chr.17: 75110311-75110311 on Build GRCh38
rs967482997 Chr.17: 75109955-75109955 on Build GRCh38
rs888494107 Chr.17: 75110308-75110308 on Build GRCh38
rs898894803 Chr.17: 75110025-75110025 on Build GRCh38
rs553233536 Chr.17: 75109924-75109924 on Build GRCh38
rs922866191 Chr.17: 75110301-75110301 on Build GRCh38
rs371523218 Chr.17: 75110341-75110341 on Build GRCh38
rs576953229 Chr.17: 75110160-75110160 on Build GRCh38
rs757269151 Chr.17: 75110111-75110111 on Build GRCh38
rs746044018 Chr.17: 75110176-75110176 on Build GRCh38
rs774547674 Chr.17: 75109951-75109951 on Build GRCh38
rs200063152 Chr.17: 75110323-75110323 on Build GRCh38
rs555343981 Chr.17: 75110099-75110099 on Build GRCh38
rs760590004 Chr.17: 75110294-75110294 on Build GRCh38
rs748856081 Chr.17: 75110338-75110338 on Build GRCh38
rs566864546 Chr.17: 75109979-75109979 on Build GRCh38
rs755123704 Chr.17: 75110314-75110314 on Build GRCh38
rs774170159 Chr.17: 75110260-75110260 on Build GRCh38
rs901163750 Chr.17: 75110006-75110006 on Build GRCh38
rs942966602 Chr.17: 75109993-75109993 on Build GRCh38
rs575608536 Chr.17: 75110131-75110131 on Build GRCh38
rs1047123938 Chr.17: 75110086-75110086 on Build GRCh38
rs1037289444 Chr.17: 75110251-75110251 on Build GRCh38
rs1014165065 Chr.17: 75110135-75110135 on Build GRCh38
rs772654381 Chr.17: 75110256-75110256 on Build GRCh38
rs1053970583 Chr.17: 75109974-75109974 on Build GRCh38
rs748604383 Chr.17: 75110246-75110246 on Build GRCh38
rs922088892 Chr.17: 75110221-75110221 on Build GRCh38
rs748531608 Chr.17: 75110321-75110321 on Build GRCh38
rs540300515 Chr.17: 75110137-75110137 on Build GRCh38
rs888036441 Chr.17: 75110015-75110015 on Build GRCh38
rs74697881 Chr.17: 75110337-75110337 on Build GRCh38
rs989607943 Chr.17: 75110224-75110224 on Build GRCh38
rs900758320 Chr.17: 75109985-75109985 on Build GRCh38
rs936809273 Chr.17: 75109966-75109966 on Build GRCh38
rs1052431955 Chr.17: 75110232-75110232 on Build GRCh38
rs1005320790 Chr.17: 75110109-75110109 on Build GRCh38
rs947415506 Chr.17: 75110189-75110189 on Build GRCh38
rs1039199388 Chr.17: 75109981-75109981 on Build GRCh38
rs988876998 Chr.17: 75109968-75109968 on Build GRCh38
rs1041408396 Chr.17: 75109994-75109994 on Build GRCh38
rs981473359 Chr.17: 75109938-75109938 on Build GRCh38
rs1004616792 Chr.17: 75110001-75110001 on Build GRCh38
rs533465135 Chr.17: 75109922-75109923 on Build GRCh38
rs546060870 Chr.17: 75110192-75110192 on Build GRCh38
rs557998906 Chr.17: 75110153-75110153 on Build GRCh38
rs772645095 Chr.17: 75110280-75110285 on Build GRCh38
rs767427266 Chr.17: 75110266-75110266 on Build GRCh38
rs933608973 Chr.17: 75110222-75110222 on Build GRCh38
rs958669641 Chr.17: 75109937-75109937 on Build GRCh38
rs199703778 Chr.17: 75110275-75110275 on Build GRCh38
rs200494633 Chr.17: 75110262-75110262 on Build GRCh38
rs911400129 Chr.17: 75110230-75110230 on Build GRCh38
rs1015979566 Chr.17: 75110022-75110022 on Build GRCh38
rs781421043 Chr.17: 75110316-75110316 on Build GRCh38
rs940378855 Chr.17: 75110249-75110249 on Build GRCh38
rs1023390403 Chr.17: 75110029-75110029 on Build GRCh38
rs781575668 Chr.17: 75110240-75110240 on Build GRCh38
rs759276703 Chr.17: 75110351-75110351 on Build GRCh38
rs1034203133 Chr.17: 75109925-75109925 on Build GRCh38
rs992867086 Chr.17: 75109949-75109949 on Build GRCh38
rs974917103 Chr.17: 75110191-75110191 on Build GRCh38
rs764090357 Chr.17: 75110300-75110300 on Build GRCh38
rs770276810 Chr.17: 75110250-75110250 on Build GRCh38
rs957127077 Chr.17: 75110195-75110195 on Build GRCh38
rs977167434 Chr.17: 75109959-75109959 on Build GRCh38
rs780055517 Chr.17: 75110312-75110312 on Build GRCh38
rs771840843 Chr.17: 75110362-75110362 on Build GRCh38
rs79652007 Chr.17: 75110027-75110027 on Build GRCh38
rs770596381 Chr.17: 75110340-75110340 on Build GRCh38
rs925715505 Chr.17: 75109962-75109962 on Build GRCh38
rs775218554 Chr.17: 75110366-75110366 on Build GRCh38
rs888545928 Chr.17: 75110100-75110100 on Build GRCh38
rs1047069682 Chr.17: 75110307-75110307 on Build GRCh38
rs970704183 Chr.17: 75110076-75110076 on Build GRCh38
rs925662794 Chr.17: 75110178-75110178 on Build GRCh38
rs751807136 Chr.17: 75110313-75110313 on Build GRCh38
rs1024682859 Chr.17: 75110152-75110152 on Build GRCh38
rs1034150981 Chr.17: 75110120-75110120 on Build GRCh38

View Full Product Details