Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 47839740-47840242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAD2
Entrez Gene ID
4087
Gene Name
SMAD family member 2
Gene Aliases
JV18, JV18-1, MADH2, MADR2, hMAD-2, hSMAD2
UniGene
Hs.12253
Gene Chromosome Location
Chr.18: 47833095-47931193 on Build GRCh38
Transcripts
NM_001003652.3 NM_001135937.2 NM_005901.5 XM_005258259.3 XM_006722451.3 XM_011525984.2 XM_011525985.2 XM_017025745.1 XM_017025746.1 XM_017025747.1 XM_017025748.1 XM_017025750.1

SNP Detail

SNP ID Chromosome Location
rs917293892 Chr.18: 47840019-47840019 on Build GRCh38
rs996761568 Chr.18: 47839907-47839907 on Build GRCh38
rs948725318 Chr.18: 47840013-47840013 on Build GRCh38
rs956103529 Chr.18: 47840191-47840191 on Build GRCh38
rs987002152 Chr.18: 47839928-47839928 on Build GRCh38
rs776208819 Chr.18: 47839795-47839795 on Build GRCh38
rs567548228 Chr.18: 47840119-47840119 on Build GRCh38
rs996686382 Chr.18: 47840134-47840134 on Build GRCh38
rs974595193 Chr.18: 47839972-47839972 on Build GRCh38
rs1009396203 Chr.18: 47840093-47840093 on Build GRCh38
rs1044340522 Chr.18: 47840034-47840034 on Build GRCh38
rs892323430 Chr.18: 47840074-47840074 on Build GRCh38
rs886850106 Chr.18: 47840094-47840094 on Build GRCh38
rs752940516 Chr.18: 47839972-47839972 on Build GRCh38
rs372958015 Chr.18: 47839780-47839780 on Build GRCh38
rs955628973 Chr.18: 47840149-47840149 on Build GRCh38
rs771567730 Chr.18: 47839981-47839982 on Build GRCh38
rs908726027 Chr.18: 47839944-47839944 on Build GRCh38
rs961551473 Chr.18: 47839959-47839959 on Build GRCh38
rs956134643 Chr.18: 47839996-47839996 on Build GRCh38
rs75567201 Chr.18: 47840038-47840038 on Build GRCh38
rs571111674 Chr.18: 47840028-47840028 on Build GRCh38
rs536675735 Chr.18: 47840148-47840148 on Build GRCh38
rs12954768 Chr.18: 47840051-47840051 on Build GRCh38
rs968925929 Chr.18: 47840156-47840156 on Build GRCh38
rs1028568117 Chr.18: 47839910-47839910 on Build GRCh38
rs192498194 Chr.18: 47839813-47839813 on Build GRCh38
rs996524000 Chr.18: 47840145-47840145 on Build GRCh38
rs758146248 Chr.18: 47840080-47840080 on Build GRCh38
rs974228576 Chr.18: 47840209-47840209 on Build GRCh38
rs751488332 Chr.18: 47840183-47840183 on Build GRCh38
rs995628729 Chr.18: 47839777-47839777 on Build GRCh38
rs903567459 Chr.18: 47840127-47840127 on Build GRCh38
rs1003942275 Chr.18: 47840107-47840107 on Build GRCh38
rs948941895 Chr.18: 47840026-47840026 on Build GRCh38
rs965247466 Chr.18: 47839870-47839870 on Build GRCh38
rs1056365738 Chr.18: 47840065-47840065 on Build GRCh38
rs374947832 Chr.18: 47839929-47839929 on Build GRCh38
rs900883503 Chr.18: 47840133-47840133 on Build GRCh38
rs1022033322 Chr.18: 47839787-47839787 on Build GRCh38
rs764054985 Chr.18: 47840168-47840168 on Build GRCh38
rs556561829 Chr.18: 47840167-47840167 on Build GRCh38
rs920006422 Chr.18: 47840002-47840002 on Build GRCh38
rs72661151 Chr.18: 47839769-47839769 on Build GRCh38
rs1043906749 Chr.18: 47840110-47840110 on Build GRCh38
rs72661150 Chr.18: 47839839-47839839 on Build GRCh38
rs756206334 Chr.18: 47840187-47840187 on Build GRCh38
rs187564036 Chr.18: 47839793-47839793 on Build GRCh38
rs1008921721 Chr.18: 47840160-47840160 on Build GRCh38
rs765895454 Chr.18: 47839779-47839779 on Build GRCh38
rs764805650 Chr.18: 47839867-47839867 on Build GRCh38

View Full Product Details