Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 49583470-49583975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIPG
Entrez Gene ID
9388
Gene Name
lipase G, endothelial type
Gene Aliases
EDL, EL, PRO719
UniGene
Hs.465102
Gene Chromosome Location
Chr.18: 49560699-49592908 on Build GRCh38
Transcripts
NM_001308006.1 NM_006033.3 XM_005258390.1 XM_011526265.2 XM_011526267.1 XM_017026095.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7170422 c.1261C>G p.Q421E Chr.18: 49583659-49583659 on Build GRCh38
6386480 c.1324C>T p.R442W Chr.18: 49583722-49583722 on Build GRCh38
6383861 c.1361G>A p.G454E Chr.18: 49583759-49583759 on Build GRCh38
1662591 c.1171G>A p.E391K Chr.18: 49583569-49583569 on Build GRCh38
7375113 c.1187A>G p.N396S Chr.18: 49583585-49583585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370475831 Chr.18: 49583566-49583566 on Build GRCh38
rs776284980 Chr.18: 49583568-49583568 on Build GRCh38
rs36095398 Chr.18: 49583894-49583894 on Build GRCh38
rs773201036 Chr.18: 49583805-49583805 on Build GRCh38
rs569210245 Chr.18: 49583875-49583875 on Build GRCh38
rs35406007 Chr.18: 49583842-49583843 on Build GRCh38
rs998877022 Chr.18: 49583853-49583853 on Build GRCh38
rs77960347 Chr.18: 49583585-49583585 on Build GRCh38
rs140183486 Chr.18: 49583711-49583711 on Build GRCh38
rs769393759 Chr.18: 49583683-49583683 on Build GRCh38
rs867833592 Chr.18: 49583882-49583882 on Build GRCh38
rs1017805073 Chr.18: 49583934-49583934 on Build GRCh38
rs372872369 Chr.18: 49583723-49583723 on Build GRCh38
rs758746305 Chr.18: 49583610-49583610 on Build GRCh38
rs941163860 Chr.18: 49583746-49583746 on Build GRCh38
rs538703118 Chr.18: 49583792-49583794 on Build GRCh38
rs747420198 Chr.18: 49583741-49583741 on Build GRCh38
rs780127285 Chr.18: 49583781-49583782 on Build GRCh38
rs745918589 Chr.18: 49583658-49583659 on Build GRCh38
rs755091056 Chr.18: 49583719-49583719 on Build GRCh38
rs767465938 Chr.18: 49583712-49583712 on Build GRCh38
rs749538688 Chr.18: 49583784-49583796 on Build GRCh38
rs201429079 Chr.18: 49583592-49583592 on Build GRCh38
rs965317058 Chr.18: 49583952-49583952 on Build GRCh38
rs769490055 Chr.18: 49583770-49583770 on Build GRCh38
rs778212352 Chr.18: 49583626-49583626 on Build GRCh38
rs757211939 Chr.18: 49583528-49583529 on Build GRCh38
rs368406414 Chr.18: 49583792-49583792 on Build GRCh38
rs757537964 Chr.18: 49583747-49583747 on Build GRCh38
rs755003256 Chr.18: 49583604-49583604 on Build GRCh38
rs957601708 Chr.18: 49583642-49583642 on Build GRCh38
rs975476102 Chr.18: 49583685-49583685 on Build GRCh38
rs753952241 Chr.18: 49583597-49583597 on Build GRCh38
rs891689752 Chr.18: 49583605-49583605 on Build GRCh38
rs138438163 Chr.18: 49583569-49583569 on Build GRCh38
rs762656984 Chr.18: 49583691-49583691 on Build GRCh38
rs1024350402 Chr.18: 49583551-49583551 on Build GRCh38
rs763600095 Chr.18: 49583709-49583709 on Build GRCh38
rs768125009 Chr.18: 49583633-49583633 on Build GRCh38
rs746368914 Chr.18: 49583756-49583756 on Build GRCh38
rs367553125 Chr.18: 49583722-49583722 on Build GRCh38
rs777226346 Chr.18: 49583653-49583653 on Build GRCh38
rs750339464 Chr.18: 49583714-49583714 on Build GRCh38
rs1024716304 Chr.18: 49583840-49583840 on Build GRCh38
rs142768331 Chr.18: 49583507-49583508 on Build GRCh38
rs144717284 Chr.18: 49583563-49583563 on Build GRCh38
rs748682092 Chr.18: 49583771-49583771 on Build GRCh38
rs146165263 Chr.18: 49583589-49583589 on Build GRCh38
rs771823302 Chr.18: 49583804-49583804 on Build GRCh38
rs748339975 Chr.18: 49583608-49583608 on Build GRCh38
rs766237114 Chr.18: 49583818-49583818 on Build GRCh38
rs374219453 Chr.18: 49583552-49583552 on Build GRCh38
rs777816384 Chr.18: 49583740-49583740 on Build GRCh38
rs189018769 Chr.18: 49583909-49583909 on Build GRCh38
rs756283819 Chr.18: 49583683-49583684 on Build GRCh38
rs1029669146 Chr.18: 49583884-49583884 on Build GRCh38
rs756218912 Chr.18: 49583509-49583509 on Build GRCh38
rs907228622 Chr.18: 49583845-49583845 on Build GRCh38
rs747269340 Chr.18: 49583631-49583631 on Build GRCh38
rs536799767 Chr.18: 49583689-49583689 on Build GRCh38
rs745417719 Chr.18: 49583657-49583657 on Build GRCh38
rs774203324 Chr.18: 49583710-49583710 on Build GRCh38
rs752851746 Chr.18: 49583823-49583823 on Build GRCh38
rs184281707 Chr.18: 49583900-49583900 on Build GRCh38
rs768873191 Chr.18: 49583532-49583532 on Build GRCh38
rs201924713 Chr.18: 49583559-49583559 on Build GRCh38
rs758649469 Chr.18: 49583728-49583728 on Build GRCh38
rs748536123 Chr.18: 49583546-49583546 on Build GRCh38
rs751513507 Chr.18: 49583586-49583586 on Build GRCh38
rs181279169 Chr.18: 49583564-49583564 on Build GRCh38
rs201383506 Chr.18: 49583579-49583579 on Build GRCh38
rs768389316 Chr.18: 49583780-49583780 on Build GRCh38
rs889788828 Chr.18: 49583886-49583886 on Build GRCh38
rs552138141 Chr.18: 49583514-49583514 on Build GRCh38
rs753125646 Chr.18: 49583763-49583763 on Build GRCh38
rs79592405 Chr.18: 49583807-49583807 on Build GRCh38
rs922299691 Chr.18: 49583707-49583707 on Build GRCh38
rs749311974 Chr.18: 49583524-49583524 on Build GRCh38
rs771499100 Chr.18: 49583637-49583637 on Build GRCh38
rs781773640 Chr.18: 49583749-49583749 on Build GRCh38
rs773827861 Chr.18: 49583788-49583788 on Build GRCh38
rs773494142 Chr.18: 49583682-49583682 on Build GRCh38
rs781157720 Chr.18: 49583652-49583655 on Build GRCh38

View Full Product Details