Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 50920374-50920876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ME2
Entrez Gene ID
4200
Gene Name
malic enzyme 2
Gene Aliases
ODS1
UniGene
Hs.233119
Gene Chromosome Location
Chr.18: 50878604-50949795 on Build GRCh38
Transcripts
NM_001168335.1 NM_002396.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5645659 c.828C>G p.F276L Chr.18: 50920549-50920549 on Build GRCh38
6083637 c.878C>T p.A293V Chr.18: 50920694-50920694 on Build GRCh38
6399265 c.878C>A p.A293E Chr.18: 50920694-50920694 on Build GRCh38
988892 c.762C>T p.F254F Chr.18: 50920483-50920483 on Build GRCh38
4839758 c.763G>A p.E255K Chr.18: 50920484-50920484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141872254 Chr.18: 50920720-50920720 on Build GRCh38
rs753027874 Chr.18: 50920422-50920422 on Build GRCh38
rs531338985 Chr.18: 50920803-50920803 on Build GRCh38
rs770434462 Chr.18: 50920452-50920452 on Build GRCh38
rs778949812 Chr.18: 50920699-50920699 on Build GRCh38
rs147495819 Chr.18: 50920464-50920464 on Build GRCh38
rs767066614 Chr.18: 50920625-50920625 on Build GRCh38
rs915760264 Chr.18: 50920698-50920698 on Build GRCh38
rs201924704 Chr.18: 50920530-50920530 on Build GRCh38
rs769826563 Chr.18: 50920474-50920474 on Build GRCh38
rs768258864 Chr.18: 50920725-50920725 on Build GRCh38
rs777427784 Chr.18: 50920429-50920429 on Build GRCh38
rs749238872 Chr.18: 50920571-50920571 on Build GRCh38
rs768582400 Chr.18: 50920483-50920483 on Build GRCh38
rs766432811 Chr.18: 50920484-50920484 on Build GRCh38
rs753838703 Chr.18: 50920502-50920502 on Build GRCh38
rs200088300 Chr.18: 50920726-50920726 on Build GRCh38
rs771958910 Chr.18: 50920710-50920710 on Build GRCh38
rs674210 Chr.18: 50920583-50920583 on Build GRCh38
rs757710305 Chr.18: 50920624-50920624 on Build GRCh38
rs762826369 Chr.18: 50920480-50920480 on Build GRCh38
rs749183956 Chr.18: 50920659-50920659 on Build GRCh38
rs977355174 Chr.18: 50920603-50920603 on Build GRCh38
rs188927164 Chr.18: 50920569-50920569 on Build GRCh38
rs779443827 Chr.18: 50920418-50920418 on Build GRCh38
rs1007153532 Chr.18: 50920832-50920832 on Build GRCh38
rs372850521 Chr.18: 50920689-50920689 on Build GRCh38
rs759556181 Chr.18: 50920587-50920587 on Build GRCh38
rs748122922 Chr.18: 50920804-50920804 on Build GRCh38
rs571098710 Chr.18: 50920795-50920795 on Build GRCh38
rs148114734 Chr.18: 50920529-50920529 on Build GRCh38
rs767927111 Chr.18: 50920751-50920751 on Build GRCh38
rs747890306 Chr.18: 50920700-50920700 on Build GRCh38
rs780078846 Chr.18: 50920553-50920553 on Build GRCh38
rs368690374 Chr.18: 50920594-50920594 on Build GRCh38
rs564269423 Chr.18: 50920471-50920471 on Build GRCh38
rs775638732 Chr.18: 50920479-50920479 on Build GRCh38
rs780594255 Chr.18: 50920461-50920461 on Build GRCh38
rs929332333 Chr.18: 50920771-50920771 on Build GRCh38
rs766182822 Chr.18: 50920773-50920773 on Build GRCh38
rs776769224 Chr.18: 50920715-50920715 on Build GRCh38
rs369204116 Chr.18: 50920737-50920737 on Build GRCh38
rs754849203 Chr.18: 50920697-50920697 on Build GRCh38
rs774983052 Chr.18: 50920730-50920730 on Build GRCh38
rs34223640 Chr.18: 50920700-50920700 on Build GRCh38
rs779738940 Chr.18: 50920642-50920642 on Build GRCh38
rs765726932 Chr.18: 50920507-50920507 on Build GRCh38
rs778031621 Chr.18: 50920528-50920528 on Build GRCh38
rs776977350 Chr.18: 50920586-50920586 on Build GRCh38
rs929522394 Chr.18: 50920433-50920433 on Build GRCh38
rs760140218 Chr.18: 50920760-50920760 on Build GRCh38
rs759651947 Chr.18: 50920506-50920506 on Build GRCh38
rs375262017 Chr.18: 50920562-50920562 on Build GRCh38
rs377204472 Chr.18: 50920410-50920410 on Build GRCh38
rs1018066785 Chr.18: 50920851-50920851 on Build GRCh38
rs562932846 Chr.18: 50920546-50920546 on Build GRCh38
rs775526709 Chr.18: 50920724-50920724 on Build GRCh38
rs763473128 Chr.18: 50920597-50920597 on Build GRCh38
rs548293496 Chr.18: 50920622-50920622 on Build GRCh38
rs1018453407 Chr.18: 50920533-50920533 on Build GRCh38
rs190877059 Chr.18: 50920794-50920794 on Build GRCh38
rs769483854 Chr.18: 50920551-50920551 on Build GRCh38
rs376084308 Chr.18: 50920717-50920717 on Build GRCh38
rs775937507 Chr.18: 50920596-50920596 on Build GRCh38
rs147910635 Chr.18: 50920573-50920573 on Build GRCh38
rs750111248 Chr.18: 50920639-50920639 on Build GRCh38
rs764715481 Chr.18: 50920619-50920619 on Build GRCh38
rs371999190 Chr.18: 50920772-50920772 on Build GRCh38
rs774037320 Chr.18: 50920575-50920575 on Build GRCh38
rs755614871 Chr.18: 50920641-50920641 on Build GRCh38
rs533138526 Chr.18: 50920545-50920545 on Build GRCh38
rs753218190 Chr.18: 50920523-50920523 on Build GRCh38
rs965131015 Chr.18: 50920537-50920537 on Build GRCh38
rs758697436 Chr.18: 50920525-50920525 on Build GRCh38
rs899554244 Chr.18: 50920540-50920540 on Build GRCh38
rs748022985 Chr.18: 50920580-50920580 on Build GRCh38
rs758822402 Chr.18: 50920428-50920428 on Build GRCh38
rs937521096 Chr.18: 50920604-50920604 on Build GRCh38
rs910082392 Chr.18: 50920601-50920601 on Build GRCh38
rs754329785 Chr.18: 50920405-50920405 on Build GRCh38
rs370224149 Chr.18: 50920442-50920442 on Build GRCh38
rs772753626 Chr.18: 50920645-50920648 on Build GRCh38

View Full Product Details