Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36393627-36394142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP82
Entrez Gene ID
284406
Gene Name
ZFP82 zinc finger protein
Gene Aliases
ZNF545
UniGene
Hs.558734
Gene Chromosome Location
Chr.19: 36383102-36418674 on Build GRCh38
Transcripts
NM_001321917.1 NM_001321918.1 NM_001321919.1 NM_133466.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4812266 c.664G>A p.G222S Chr.19: 36393676-36393676 on Build GRCh38
347091 c.475_476CC>TA p.P159>? Chr.19: 36393864-36393865 on Build GRCh38
3532926 c.407G>A p.G136E Chr.19: 36393933-36393933 on Build GRCh38
5524233 c.675C>T p.L225L Chr.19: 36393665-36393665 on Build GRCh38
418305 c.664G>A p.G222S Chr.19: 36393676-36393676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144032300 Chr.19: 36393979-36393979 on Build GRCh38
rs539169356 Chr.19: 36393841-36393841 on Build GRCh38
rs368377208 Chr.19: 36393879-36393879 on Build GRCh38
rs373111886 Chr.19: 36393795-36393795 on Build GRCh38
rs756799775 Chr.19: 36393986-36393986 on Build GRCh38
rs370541972 Chr.19: 36394052-36394052 on Build GRCh38
rs773420403 Chr.19: 36394050-36394050 on Build GRCh38
rs748645640 Chr.19: 36394011-36394011 on Build GRCh38
rs114627398 Chr.19: 36393749-36393749 on Build GRCh38
rs200441887 Chr.19: 36393736-36393736 on Build GRCh38
rs750725834 Chr.19: 36393960-36393961 on Build GRCh38
rs761926690 Chr.19: 36393759-36393759 on Build GRCh38
rs746631245 Chr.19: 36393829-36393829 on Build GRCh38
rs752586717 Chr.19: 36393931-36393931 on Build GRCh38
rs749551905 Chr.19: 36393850-36393850 on Build GRCh38
rs1028168703 Chr.19: 36394059-36394059 on Build GRCh38
rs779462448 Chr.19: 36394009-36394009 on Build GRCh38
rs776665195 Chr.19: 36394095-36394095 on Build GRCh38
rs1016802799 Chr.19: 36393922-36393922 on Build GRCh38
rs950270480 Chr.19: 36393959-36393959 on Build GRCh38
rs138067157 Chr.19: 36393806-36393806 on Build GRCh38
rs745557666 Chr.19: 36394007-36394007 on Build GRCh38
rs1055932667 Chr.19: 36393845-36393845 on Build GRCh38
rs200935785 Chr.19: 36393750-36393750 on Build GRCh38
rs759830950 Chr.19: 36394097-36394097 on Build GRCh38
rs573960775 Chr.19: 36394015-36394015 on Build GRCh38
rs775310362 Chr.19: 36394115-36394115 on Build GRCh38
rs777293057 Chr.19: 36393826-36393826 on Build GRCh38
rs201807470 Chr.19: 36393735-36393735 on Build GRCh38
rs561996621 Chr.19: 36393960-36393960 on Build GRCh38
rs983728234 Chr.19: 36393871-36393871 on Build GRCh38
rs376078593 Chr.19: 36393858-36393858 on Build GRCh38
rs757850059 Chr.19: 36393812-36393812 on Build GRCh38
rs779465110 Chr.19: 36393768-36393768 on Build GRCh38
rs867214075 Chr.19: 36393666-36393666 on Build GRCh38
rs148540683 Chr.19: 36393982-36393982 on Build GRCh38
rs764982529 Chr.19: 36394099-36394099 on Build GRCh38
rs776916108 Chr.19: 36393892-36393892 on Build GRCh38
rs963573736 Chr.19: 36393935-36393935 on Build GRCh38
rs945254327 Chr.19: 36394084-36394084 on Build GRCh38
rs560042774 Chr.19: 36393880-36393880 on Build GRCh38
rs867505795 Chr.19: 36393843-36393843 on Build GRCh38
rs766173004 Chr.19: 36393782-36393782 on Build GRCh38
rs748530647 Chr.19: 36393770-36393770 on Build GRCh38
rs371577902 Chr.19: 36393861-36393861 on Build GRCh38
rs987832155 Chr.19: 36393671-36393671 on Build GRCh38
rs573122449 Chr.19: 36393862-36393862 on Build GRCh38
rs926359846 Chr.19: 36393994-36393994 on Build GRCh38
rs763506381 Chr.19: 36393743-36393743 on Build GRCh38
rs149521606 Chr.19: 36393842-36393842 on Build GRCh38
rs913551821 Chr.19: 36393690-36393690 on Build GRCh38
rs370323805 Chr.19: 36393658-36393658 on Build GRCh38
rs780655678 Chr.19: 36394005-36394005 on Build GRCh38
rs374534753 Chr.19: 36393867-36393867 on Build GRCh38
rs867761074 Chr.19: 36393778-36393778 on Build GRCh38
rs767727975 Chr.19: 36393764-36393764 on Build GRCh38
rs753632446 Chr.19: 36393794-36393794 on Build GRCh38
rs770153980 Chr.19: 36393709-36393709 on Build GRCh38
rs765098015 Chr.19: 36393896-36393896 on Build GRCh38
rs778648836 Chr.19: 36393796-36393796 on Build GRCh38
rs780307893 Chr.19: 36393660-36393660 on Build GRCh38
rs375155109 Chr.19: 36394063-36394063 on Build GRCh38
rs35344339 Chr.19: 36393903-36393904 on Build GRCh38
rs759194234 Chr.19: 36393895-36393895 on Build GRCh38
rs766575200 Chr.19: 36393887-36393887 on Build GRCh38

View Full Product Details