Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37769823-37770337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF573
Entrez Gene ID
126231
Gene Name
zinc finger protein 573
Gene Aliases
-
UniGene
Hs.531262
Gene Chromosome Location
Chr.19: 37738274-37779626 on Build GRCh38
Transcripts
NM_001172689.1 NM_001172690.1 NM_001172691.1 NM_001172692.1 NM_152360.3 XM_011526446.2 XM_017026275.1 XM_017026276.1 XM_017026277.1 XM_017026278.1 XM_017026279.1 XM_017026280.1 XM_017026281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5781034 c.212C>G p.S71C Chr.19: 37770088-37770088 on Build GRCh38
7399339 c.206G>T p.G69V Chr.19: 37770094-37770094 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs957755627 Chr.19: 37769917-37769917 on Build GRCh38
rs856304 Chr.19: 37770270-37770270 on Build GRCh38
rs1020383432 Chr.19: 37769962-37769962 on Build GRCh38
rs372466448 Chr.19: 37769943-37769943 on Build GRCh38
rs773195380 Chr.19: 37769899-37769899 on Build GRCh38
rs748663709 Chr.19: 37770138-37770138 on Build GRCh38
rs538473339 Chr.19: 37770271-37770271 on Build GRCh38
rs905510967 Chr.19: 37769878-37769878 on Build GRCh38
rs752291976 Chr.19: 37769959-37769959 on Build GRCh38
rs754393559 Chr.19: 37769889-37769889 on Build GRCh38
rs556871418 Chr.19: 37770312-37770312 on Build GRCh38
rs927015881 Chr.19: 37770193-37770193 on Build GRCh38
rs1013680045 Chr.19: 37770184-37770184 on Build GRCh38
rs1032415814 Chr.19: 37769873-37769873 on Build GRCh38
rs974385815 Chr.19: 37770027-37770027 on Build GRCh38
rs955418620 Chr.19: 37769974-37769974 on Build GRCh38
rs778180572 Chr.19: 37770052-37770052 on Build GRCh38
rs974443776 Chr.19: 37769927-37769927 on Build GRCh38
rs750040203 Chr.19: 37770025-37770025 on Build GRCh38
rs554953077 Chr.19: 37770122-37770122 on Build GRCh38
rs907646105 Chr.19: 37770119-37770119 on Build GRCh38
rs762888360 Chr.19: 37769963-37769963 on Build GRCh38
rs758035860 Chr.19: 37769991-37769991 on Build GRCh38
rs566437757 Chr.19: 37769995-37769995 on Build GRCh38
rs936993911 Chr.19: 37770202-37770202 on Build GRCh38
rs781503104 Chr.19: 37770301-37770301 on Build GRCh38
rs533898906 Chr.19: 37770151-37770151 on Build GRCh38
rs933082181 Chr.19: 37769967-37769967 on Build GRCh38
rs1032618353 Chr.19: 37770236-37770236 on Build GRCh38
rs776148768 Chr.19: 37770272-37770272 on Build GRCh38
rs780796010 Chr.19: 37770006-37770006 on Build GRCh38
rs907741798 Chr.19: 37769984-37769984 on Build GRCh38
rs986870341 Chr.19: 37769975-37769975 on Build GRCh38
rs907689682 Chr.19: 37770287-37770288 on Build GRCh38
rs779268011 Chr.19: 37770075-37770075 on Build GRCh38
rs187088490 Chr.19: 37770269-37770269 on Build GRCh38
rs778564911 Chr.19: 37769934-37769934 on Build GRCh38
rs1000173103 Chr.19: 37770219-37770219 on Build GRCh38
rs1045436061 Chr.19: 37770170-37770170 on Build GRCh38
rs982892786 Chr.19: 37770091-37770091 on Build GRCh38
rs1001480352 Chr.19: 37769892-37769892 on Build GRCh38
rs890945972 Chr.19: 37770286-37770286 on Build GRCh38
rs951583091 Chr.19: 37770080-37770080 on Build GRCh38
rs949829860 Chr.19: 37770153-37770153 on Build GRCh38
rs551405434 Chr.19: 37769933-37769933 on Build GRCh38
rs1033338874 Chr.19: 37769915-37769915 on Build GRCh38
rs763650255 Chr.19: 37769985-37769985 on Build GRCh38
rs1025560644 Chr.19: 37770185-37770185 on Build GRCh38
rs140912721 Chr.19: 37770282-37770284 on Build GRCh38
rs987229825 Chr.19: 37769981-37769981 on Build GRCh38
rs920139743 Chr.19: 37770039-37770039 on Build GRCh38
rs762380162 Chr.19: 37770013-37770014 on Build GRCh38

View Full Product Details