Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52445673-52446172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCCHC11
Entrez Gene ID
23318
Gene Name
zinc finger CCHC-type containing 11
Gene Aliases
PAPD3, TUT4
UniGene
Hs.655407
Gene Chromosome Location
Chr.1: 52423275-52553463 on Build GRCh38
Transcripts
NM_001009881.2 NM_015269.2 XM_005270676.2 XM_005270678.2 XM_005270679.2 XM_005270680.2 XM_005270681.2 XM_005270682.2 XM_005270683.2 XM_006710498.2 XM_006710499.3 XM_006710500.1 XM_011541097.1 XM_011541098.1 XM_011541099.1 XM_011541100.2 XM_017000803.1 XM_017000804.1 XM_017000805.1 XM_017000806.1 XM_017000807.1 XM_017000808.1 XM_017000809.1 XM_017000810.1 XM_017000811.1 XM_017000812.1 XM_017000813.1 XM_017000814.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4747255 c.3738_3739delAA p.K1246fs*29 Chr.1: 52445957-52445958 on Build GRCh38
5984668 c.3741G>A p.M1247I Chr.1: 52445868-52445868 on Build GRCh38
1474070 c.3807C>T p.L1269L Chr.1: 52445802-52445802 on Build GRCh38
1343385 c.3817G>T p.E1273* Chr.1: 52445792-52445792 on Build GRCh38
1343386 c.3771T>C p.N1257N Chr.1: 52445838-52445838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201195865 Chr.1: 52445875-52445875 on Build GRCh38
rs374119914 Chr.1: 52445970-52445970 on Build GRCh38
rs906537328 Chr.1: 52445705-52445705 on Build GRCh38
rs994237592 Chr.1: 52445721-52445721 on Build GRCh38
rs939118747 Chr.1: 52446141-52446141 on Build GRCh38
rs142509964 Chr.1: 52446040-52446040 on Build GRCh38
rs150564690 Chr.1: 52446075-52446075 on Build GRCh38
rs998065682 Chr.1: 52446050-52446050 on Build GRCh38
rs948234431 Chr.1: 52446095-52446095 on Build GRCh38
rs773149209 Chr.1: 52445760-52445760 on Build GRCh38
rs752747844 Chr.1: 52445886-52445886 on Build GRCh38
rs778091417 Chr.1: 52445901-52445901 on Build GRCh38
rs746446170 Chr.1: 52446018-52446018 on Build GRCh38
rs899676675 Chr.1: 52445750-52445750 on Build GRCh38
rs192115020 Chr.1: 52445745-52445745 on Build GRCh38
rs781682430 Chr.1: 52446002-52446002 on Build GRCh38
rs138145860 Chr.1: 52445800-52445800 on Build GRCh38
rs759462090 Chr.1: 52445874-52445874 on Build GRCh38
rs183236746 Chr.1: 52446090-52446090 on Build GRCh38
rs1018139797 Chr.1: 52445892-52445892 on Build GRCh38
rs1046802985 Chr.1: 52445737-52445737 on Build GRCh38
rs776293467 Chr.1: 52446111-52446111 on Build GRCh38
rs777858778 Chr.1: 52446105-52446105 on Build GRCh38
rs551755421 Chr.1: 52445719-52445719 on Build GRCh38
rs758357576 Chr.1: 52445897-52445897 on Build GRCh38
rs148422801 Chr.1: 52445805-52445805 on Build GRCh38
rs760229907 Chr.1: 52446043-52446043 on Build GRCh38
rs766103372 Chr.1: 52445838-52445838 on Build GRCh38
rs1031006944 Chr.1: 52445863-52445863 on Build GRCh38
rs760722000 Chr.1: 52446021-52446021 on Build GRCh38
rs760523193 Chr.1: 52445829-52445829 on Build GRCh38
rs527978226 Chr.1: 52445889-52445889 on Build GRCh38
rs1005371340 Chr.1: 52445746-52445746 on Build GRCh38
rs770498274 Chr.1: 52446052-52446052 on Build GRCh38
rs774752717 Chr.1: 52446092-52446092 on Build GRCh38
rs771416715 Chr.1: 52446037-52446037 on Build GRCh38
rs773561655 Chr.1: 52445772-52445774 on Build GRCh38
rs371567599 Chr.1: 52445807-52445807 on Build GRCh38
rs763467366 Chr.1: 52445888-52445890 on Build GRCh38
rs199880908 Chr.1: 52445776-52445776 on Build GRCh38
rs751669412 Chr.1: 52445964-52445964 on Build GRCh38
rs369000420 Chr.1: 52445770-52445770 on Build GRCh38
rs765338116 Chr.1: 52445881-52445881 on Build GRCh38
rs1029300649 Chr.1: 52446074-52446074 on Build GRCh38
rs2274146 Chr.1: 52445734-52445734 on Build GRCh38
rs925720938 Chr.1: 52446125-52446125 on Build GRCh38
rs770395824 Chr.1: 52446019-52446019 on Build GRCh38
rs187351370 Chr.1: 52446093-52446093 on Build GRCh38
rs768341925 Chr.1: 52445802-52445802 on Build GRCh38
rs145521302 Chr.1: 52445743-52445743 on Build GRCh38
rs202129609 Chr.1: 52445844-52445844 on Build GRCh38
rs376906157 Chr.1: 52445828-52445828 on Build GRCh38
rs958182102 Chr.1: 52446138-52446138 on Build GRCh38
rs771163607 Chr.1: 52445774-52445774 on Build GRCh38
rs747717491 Chr.1: 52446025-52446025 on Build GRCh38
rs758612882 Chr.1: 52445764-52445764 on Build GRCh38
rs954830057 Chr.1: 52445814-52445814 on Build GRCh38
rs202000815 Chr.1: 52445865-52445865 on Build GRCh38
rs957139079 Chr.1: 52445924-52445925 on Build GRCh38

View Full Product Details