Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 121367481-121367975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLASP1
Entrez Gene ID
23332
Gene Name
cytoplasmic linker associated protein 1
Gene Aliases
MAST1
UniGene
Hs.469840
Gene Chromosome Location
Chr.2: 121337776-121649476 on Build GRCh38
Transcripts
NM_001142273.1 NM_001142274.1 NM_001207051.1 NM_015282.2 XM_006712377.1 XM_006712381.1 XM_006712382.1 XM_006712383.1 XM_006712385.1 XM_011510848.1 XM_011510849.1 XM_017003657.1 XM_017003658.1 XM_017003659.1 XM_017003660.1 XM_017003661.1 XM_017003662.1 XM_017003663.1 XM_017003664.1 XM_017003665.1 XM_017003666.1 XM_017003667.1 XM_017003668.1 XM_017003669.1 XM_017003670.1 XM_017003671.1 XM_017003672.1 XM_017003673.1 XM_017003674.1 XM_017003675.1 XM_017003676.1 XM_017003677.1 XM_017003678.1 XM_017003679.1 XM_017003680.1 XM_017003681.1 XM_017003682.1 XM_017003683.1 XM_017003684.1 XM_017003685.1 XM_017003686.1 XM_017003687.1 XM_017003688.1 XM_017003689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5564320 c.2912G>A p.R971H Chr.2: 121367713-121367713 on Build GRCh38
1006017 c.3755C>A p.P1252H Chr.2: 121367719-121367719 on Build GRCh38
6301182 c.3565A>G p.N1189D Chr.2: 121367729-121367729 on Build GRCh38
1717290 c.3814G>T p.A1272S Chr.2: 121367660-121367660 on Build GRCh38
4302743 c.2989G>A p.A997T Chr.2: 121367636-121367636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144957722 Chr.2: 121367797-121367797 on Build GRCh38
rs769422360 Chr.2: 121367547-121367547 on Build GRCh38
rs765342978 Chr.2: 121367745-121367745 on Build GRCh38
rs376007044 Chr.2: 121367677-121367677 on Build GRCh38
rs778364630 Chr.2: 121367784-121367784 on Build GRCh38
rs773791218 Chr.2: 121367706-121367706 on Build GRCh38
rs749719822 Chr.2: 121367782-121367782 on Build GRCh38
rs764411908 Chr.2: 121367690-121367690 on Build GRCh38
rs779224200 Chr.2: 121367695-121367695 on Build GRCh38
rs184405260 Chr.2: 121367691-121367691 on Build GRCh38
rs555397997 Chr.2: 121367841-121367841 on Build GRCh38
rs750842299 Chr.2: 121367574-121367574 on Build GRCh38
rs759537654 Chr.2: 121367667-121367667 on Build GRCh38
rs753994205 Chr.2: 121367615-121367615 on Build GRCh38
rs770007213 Chr.2: 121367825-121367825 on Build GRCh38
rs767022894 Chr.2: 121367756-121367756 on Build GRCh38
rs772833206 Chr.2: 121367548-121367548 on Build GRCh38
rs777809365 Chr.2: 121367610-121367610 on Build GRCh38
rs751272268 Chr.2: 121367571-121367571 on Build GRCh38
rs777501878 Chr.2: 121367700-121367700 on Build GRCh38
rs756909067 Chr.2: 121367783-121367783 on Build GRCh38
rs374210311 Chr.2: 121367565-121367565 on Build GRCh38
rs202127504 Chr.2: 121367842-121367842 on Build GRCh38
rs367574144 Chr.2: 121367711-121367711 on Build GRCh38
rs573678385 Chr.2: 121367526-121367526 on Build GRCh38
rs748671391 Chr.2: 121367570-121367571 on Build GRCh38
rs560712580 Chr.2: 121367533-121367533 on Build GRCh38
rs368425278 Chr.2: 121367696-121367696 on Build GRCh38
rs150310586 Chr.2: 121367716-121367716 on Build GRCh38
rs541102905 Chr.2: 121367912-121367912 on Build GRCh38
rs994974732 Chr.2: 121367529-121367529 on Build GRCh38
rs757376442 Chr.2: 121367692-121367692 on Build GRCh38
rs746947848 Chr.2: 121367823-121367823 on Build GRCh38
rs779499735 Chr.2: 121367622-121367622 on Build GRCh38
rs767902675 Chr.2: 121367805-121367805 on Build GRCh38
rs753045343 Chr.2: 121367807-121367807 on Build GRCh38
rs750076025 Chr.2: 121367879-121367879 on Build GRCh38
rs377065133 Chr.2: 121367786-121367786 on Build GRCh38
rs192188649 Chr.2: 121367646-121367646 on Build GRCh38
rs756386666 Chr.2: 121367609-121367609 on Build GRCh38
rs369126224 Chr.2: 121367755-121367755 on Build GRCh38
rs772564602 Chr.2: 121367645-121367645 on Build GRCh38
rs747654599 Chr.2: 121367871-121367871 on Build GRCh38
rs767942296 Chr.2: 121367846-121367846 on Build GRCh38
rs755786834 Chr.2: 121367698-121367698 on Build GRCh38
rs762773952 Chr.2: 121367551-121367551 on Build GRCh38
rs758961971 Chr.2: 121367636-121367636 on Build GRCh38
rs373127015 Chr.2: 121367873-121367873 on Build GRCh38
rs917438660 Chr.2: 121367909-121367909 on Build GRCh38
rs1024332216 Chr.2: 121367744-121367744 on Build GRCh38
rs773704349 Chr.2: 121367781-121367782 on Build GRCh38
rs370655146 Chr.2: 121367684-121367684 on Build GRCh38
rs764169257 Chr.2: 121367719-121367719 on Build GRCh38
rs201624831 Chr.2: 121367785-121367785 on Build GRCh38
rs757668989 Chr.2: 121367616-121367616 on Build GRCh38
rs539277662 Chr.2: 121367767-121367767 on Build GRCh38
rs2304663 Chr.2: 121367528-121367528 on Build GRCh38
rs749879429 Chr.2: 121367815-121367815 on Build GRCh38
rs188768398 Chr.2: 121367697-121367697 on Build GRCh38
rs755382948 Chr.2: 121367762-121367762 on Build GRCh38
rs751922581 Chr.2: 121367757-121367757 on Build GRCh38
rs771179725 Chr.2: 121367815-121367817 on Build GRCh38
rs553380975 Chr.2: 121367676-121367676 on Build GRCh38
rs200536508 Chr.2: 121367704-121367704 on Build GRCh38
rs755502714 Chr.2: 121367802-121367802 on Build GRCh38
rs776920291 Chr.2: 121367721-121367721 on Build GRCh38
rs927353757 Chr.2: 121367632-121367632 on Build GRCh38
rs778562546 Chr.2: 121367813-121367813 on Build GRCh38
rs774358218 Chr.2: 121367657-121367657 on Build GRCh38
rs116176232 Chr.2: 121367637-121367637 on Build GRCh38
rs866540425 Chr.2: 121367790-121367790 on Build GRCh38
rs770222718 Chr.2: 121367768-121367770 on Build GRCh38
rs374069974 Chr.2: 121367589-121367589 on Build GRCh38
rs984647654 Chr.2: 121367689-121367689 on Build GRCh38
rs1007355864 Chr.2: 121367604-121367604 on Build GRCh38
rs769616989 Chr.2: 121367641-121367641 on Build GRCh38
rs371844669 Chr.2: 121367712-121367712 on Build GRCh38
rs369870304 Chr.2: 121367572-121367572 on Build GRCh38
rs769690058 Chr.2: 121367640-121367640 on Build GRCh38
rs555457273 Chr.2: 121367503-121367503 on Build GRCh38
rs1026229769 Chr.2: 121367820-121367820 on Build GRCh38
rs780634314 Chr.2: 121367553-121367554 on Build GRCh38
rs773579904 Chr.2: 121367826-121367826 on Build GRCh38
rs773383542 Chr.2: 121367789-121367789 on Build GRCh38
rs373849955 Chr.2: 121367654-121367654 on Build GRCh38
rs368892378 Chr.2: 121367822-121367822 on Build GRCh38
rs200313982 Chr.2: 121367886-121367886 on Build GRCh38
rs771129609 Chr.2: 121367840-121367840 on Build GRCh38
rs202106861 Chr.2: 121367611-121367611 on Build GRCh38
rs762003925 Chr.2: 121367741-121367741 on Build GRCh38
rs958786885 Chr.2: 121367854-121367854 on Build GRCh38
rs1049285706 Chr.2: 121367830-121367830 on Build GRCh38
rs369750810 Chr.2: 121367788-121367788 on Build GRCh38
rs377671445 Chr.2: 121367569-121367569 on Build GRCh38
rs756895698 Chr.2: 121367812-121367812 on Build GRCh38
rs1046414924 Chr.2: 121367864-121367864 on Build GRCh38
rs763058711 Chr.2: 121367836-121367836 on Build GRCh38
rs376745867 Chr.2: 121367814-121367814 on Build GRCh38
rs752913631 Chr.2: 121367593-121367593 on Build GRCh38
rs750577439 Chr.2: 121367754-121367754 on Build GRCh38
rs775228149 Chr.2: 121367713-121367713 on Build GRCh38
rs753058200 Chr.2: 121367863-121367863 on Build GRCh38
rs748688170 Chr.2: 121367703-121367703 on Build GRCh38
rs764805681 Chr.2: 121367877-121367877 on Build GRCh38
rs1041333700 Chr.2: 121367529-121367529 on Build GRCh38
rs886587287 Chr.2: 121367839-121367839 on Build GRCh38
rs748958721 Chr.2: 121367779-121367779 on Build GRCh38
rs747786775 Chr.2: 121367638-121367638 on Build GRCh38
rs571030723 Chr.2: 121367773-121367773 on Build GRCh38
rs757912999 Chr.2: 121367816-121367816 on Build GRCh38
rs76724451 Chr.2: 121367824-121367824 on Build GRCh38
rs761337856 Chr.2: 121367544-121367544 on Build GRCh38
rs774676325 Chr.2: 121367849-121367850 on Build GRCh38

View Full Product Details