Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50503988-50504476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMF2
Entrez Gene ID
91289
Gene Name
lipase maturation factor 2
Gene Aliases
TMEM112B, TMEM153
UniGene
Hs.150540
Gene Chromosome Location
Chr.22: 50502947-50507840 on Build GRCh38
Transcripts
NM_033200.2 XM_005261955.3 XM_006724426.3 XM_006724427.3 XM_017029077.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6239519 c.1718+8T>C p.? Chr.22: 50504332-50504332 on Build GRCh38
4105200 c.1587C>A p.P529P Chr.22: 50504396-50504396 on Build GRCh38
4783197 c.1709G>A p.G570E Chr.22: 50504349-50504349 on Build GRCh38
4907306 c.1662C>A p.P554P Chr.22: 50504396-50504396 on Build GRCh38
4874974 c.1633G>A p.A545T Chr.22: 50504425-50504425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192045599 Chr.22: 50504201-50504201 on Build GRCh38
rs745822259 Chr.22: 50504364-50504364 on Build GRCh38
rs759894936 Chr.22: 50504423-50504423 on Build GRCh38
rs367900609 Chr.22: 50504274-50504275 on Build GRCh38
rs997688047 Chr.22: 50504295-50504295 on Build GRCh38
rs146779057 Chr.22: 50504432-50504432 on Build GRCh38
rs769895409 Chr.22: 50504360-50504360 on Build GRCh38
rs775288847 Chr.22: 50504375-50504375 on Build GRCh38
rs530782702 Chr.22: 50504014-50504014 on Build GRCh38
rs370633418 Chr.22: 50504317-50504317 on Build GRCh38
rs377562246 Chr.22: 50504193-50504193 on Build GRCh38
rs768083548 Chr.22: 50504457-50504457 on Build GRCh38
rs878862441 Chr.22: 50504230-50504230 on Build GRCh38
rs762357142 Chr.22: 50504386-50504386 on Build GRCh38
rs765015884 Chr.22: 50504308-50504351 on Build GRCh38
rs202179841 Chr.22: 50504369-50504369 on Build GRCh38
rs58497169 Chr.22: 50504149-50504149 on Build GRCh38
rs752923800 Chr.22: 50504396-50504396 on Build GRCh38
rs61372881 Chr.22: 50504131-50504131 on Build GRCh38
rs564792178 Chr.22: 50504104-50504104 on Build GRCh38
rs762663080 Chr.22: 50504428-50504433 on Build GRCh38
rs776714453 Chr.22: 50504320-50504320 on Build GRCh38
rs867265336 Chr.22: 50504280-50504280 on Build GRCh38
rs548014548 Chr.22: 50504287-50504287 on Build GRCh38
rs774296217 Chr.22: 50504324-50504324 on Build GRCh38
rs183650051 Chr.22: 50504329-50504329 on Build GRCh38
rs749455077 Chr.22: 50504401-50504401 on Build GRCh38
rs753506205 Chr.22: 50504338-50504338 on Build GRCh38
rs1004742898 Chr.22: 50504339-50504339 on Build GRCh38
rs61419617 Chr.22: 50504280-50504281 on Build GRCh38
rs2334101 Chr.22: 50504068-50504068 on Build GRCh38
rs142850563 Chr.22: 50504438-50504438 on Build GRCh38
rs759076289 Chr.22: 50504451-50504451 on Build GRCh38
rs748323549 Chr.22: 50504358-50504358 on Build GRCh38
rs770881323 Chr.22: 50504408-50504408 on Build GRCh38
rs756873828 Chr.22: 50504340-50504340 on Build GRCh38
rs747462945 Chr.22: 50504349-50504349 on Build GRCh38
rs144069068 Chr.22: 50504389-50504389 on Build GRCh38
rs746352399 Chr.22: 50504305-50504305 on Build GRCh38
rs61150400 Chr.22: 50504237-50504238 on Build GRCh38
rs143227882 Chr.22: 50504433-50504433 on Build GRCh38
rs373917352 Chr.22: 50504385-50504385 on Build GRCh38
rs868064002 Chr.22: 50504220-50504220 on Build GRCh38
rs867101890 Chr.22: 50504347-50504347 on Build GRCh38
rs368135165 Chr.22: 50504414-50504414 on Build GRCh38
rs773213058 Chr.22: 50504363-50504363 on Build GRCh38
rs1004544745 Chr.22: 50504445-50504445 on Build GRCh38
rs868113446 Chr.22: 50504150-50504150 on Build GRCh38
rs111814702 Chr.22: 50504239-50504239 on Build GRCh38
rs868814669 Chr.22: 50504387-50504387 on Build GRCh38
rs781260726 Chr.22: 50504443-50504443 on Build GRCh38
rs767223957 Chr.22: 50504328-50504328 on Build GRCh38
rs753121892 Chr.22: 50504428-50504428 on Build GRCh38
rs775063528 Chr.22: 50504314-50504315 on Build GRCh38
rs529772302 Chr.22: 50504276-50504276 on Build GRCh38
rs778814819 Chr.22: 50504435-50504435 on Build GRCh38
rs60021445 Chr.22: 50504332-50504332 on Build GRCh38
rs553512868 Chr.22: 50504379-50504379 on Build GRCh38
rs755267470 Chr.22: 50504352-50504352 on Build GRCh38
rs188985791 Chr.22: 50504194-50504194 on Build GRCh38
rs755394795 Chr.22: 50504393-50504393 on Build GRCh38
rs372168527 Chr.22: 50504458-50504458 on Build GRCh38
rs773678812 Chr.22: 50504449-50504449 on Build GRCh38
rs751988553 Chr.22: 50504390-50504390 on Build GRCh38
rs376470321 Chr.22: 50504307-50504307 on Build GRCh38
rs149509833 Chr.22: 50504376-50504376 on Build GRCh38
rs773021426 Chr.22: 50504447-50504447 on Build GRCh38
rs763869613 Chr.22: 50504333-50504333 on Build GRCh38
rs768524142 Chr.22: 50504411-50504411 on Build GRCh38
rs759433180 Chr.22: 50504325-50504325 on Build GRCh38
rs151061077 Chr.22: 50504446-50504446 on Build GRCh38
rs868727077 Chr.22: 50504060-50504060 on Build GRCh38
rs139124819 Chr.22: 50504345-50504345 on Build GRCh38
rs545815428 Chr.22: 50504015-50504015 on Build GRCh38
rs563666374 Chr.22: 50504230-50504231 on Build GRCh38
rs748268355 Chr.22: 50504321-50504321 on Build GRCh38
rs866789637 Chr.22: 50504043-50504043 on Build GRCh38
rs900215378 Chr.22: 50504040-50504040 on Build GRCh38
rs574969929 Chr.22: 50504381-50504381 on Build GRCh38
rs370076519 Chr.22: 50504415-50504415 on Build GRCh38
rs867280987 Chr.22: 50504274-50504274 on Build GRCh38
rs780084108 Chr.22: 50504410-50504410 on Build GRCh38
rs756274227 Chr.22: 50504399-50504399 on Build GRCh38
rs759281476 Chr.22: 50504303-50504346 on Build GRCh38
rs764787807 Chr.22: 50504341-50504341 on Build GRCh38
rs767885328 Chr.22: 50504425-50504425 on Build GRCh38
rs765758854 Chr.22: 50504420-50504420 on Build GRCh38
rs769844514 Chr.22: 50504323-50504323 on Build GRCh38
rs781498229 Chr.22: 50504395-50504395 on Build GRCh38
rs757106437 Chr.22: 50504096-50504097 on Build GRCh38
rs61000892 Chr.22: 50504104-50504104 on Build GRCh38
rs911356061 Chr.22: 50504252-50504252 on Build GRCh38
rs752140596 Chr.22: 50504422-50504422 on Build GRCh38
rs779599525 Chr.22: 50504296-50504296 on Build GRCh38
rs139964831 Chr.22: 50504400-50504400 on Build GRCh38
rs535045211 Chr.22: 50504346-50504346 on Build GRCh38
rs373352800 Chr.22: 50504306-50504306 on Build GRCh38
rs60335508 Chr.22: 50504105-50504105 on Build GRCh38
rs756470526 Chr.22: 50504431-50504431 on Build GRCh38
rs770481325 Chr.22: 50504448-50504448 on Build GRCh38
rs868557673 Chr.22: 50504238-50504238 on Build GRCh38
rs139320797 Chr.22: 50504057-50504057 on Build GRCh38
rs748022209 Chr.22: 50504444-50504444 on Build GRCh38
rs572188318 Chr.22: 50504126-50504126 on Build GRCh38
rs922780956 Chr.22: 50504417-50504417 on Build GRCh38
rs781526513 Chr.22: 50504357-50504357 on Build GRCh38
rs772994637 Chr.22: 50504418-50504418 on Build GRCh38
rs931130532 Chr.22: 50504421-50504421 on Build GRCh38
rs143503153 Chr.22: 50504412-50504412 on Build GRCh38
rs780295812 Chr.22: 50504348-50504348 on Build GRCh38
rs368599145 Chr.22: 50504437-50504437 on Build GRCh38

View Full Product Details