Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 8879819-8880348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD18
Entrez Gene ID
56852
Gene Name
RAD18, E3 ubiquitin protein ligase
Gene Aliases
RNF73
UniGene
Hs.375684
Gene Chromosome Location
Chr.3: 8877196-8969249 on Build GRCh38
Transcripts
NM_020165.3 XM_017006873.1

SNP Detail

SNP ID Chromosome Location
rs986620872 Chr.3: 8880217-8880217 on Build GRCh38
rs981102325 Chr.3: 8880064-8880064 on Build GRCh38
rs986029486 Chr.3: 8879841-8879841 on Build GRCh38
rs973993058 Chr.3: 8880135-8880135 on Build GRCh38
rs1002798541 Chr.3: 8880038-8880038 on Build GRCh38
rs999063233 Chr.3: 8879950-8879950 on Build GRCh38
rs548704765 Chr.3: 8879869-8879869 on Build GRCh38
rs571899959 Chr.3: 8880047-8880047 on Build GRCh38
rs892214980 Chr.3: 8879972-8879972 on Build GRCh38
rs879243495 Chr.3: 8880199-8880199 on Build GRCh38
rs936324167 Chr.3: 8879900-8879900 on Build GRCh38
rs538277586 Chr.3: 8880073-8880073 on Build GRCh38
rs945149068 Chr.3: 8880000-8880000 on Build GRCh38
rs1005495849 Chr.3: 8880105-8880105 on Build GRCh38
rs759878424 Chr.3: 8880059-8880059 on Build GRCh38
rs148700041 Chr.3: 8879901-8879901 on Build GRCh38
rs998184119 Chr.3: 8880266-8880266 on Build GRCh38
rs766751805 Chr.3: 8879842-8879842 on Build GRCh38
rs888719925 Chr.3: 8880080-8880080 on Build GRCh38
rs146737733 Chr.3: 8880079-8880079 on Build GRCh38
rs181039968 Chr.3: 8880123-8880123 on Build GRCh38
rs866632631 Chr.3: 8880211-8880211 on Build GRCh38
rs35527284 Chr.3: 8880062-8880063 on Build GRCh38
rs934928209 Chr.3: 8880276-8880276 on Build GRCh38
rs536600339 Chr.3: 8880256-8880256 on Build GRCh38
rs906669606 Chr.3: 8880035-8880035 on Build GRCh38
rs893283447 Chr.3: 8879981-8879981 on Build GRCh38
rs185547222 Chr.3: 8880270-8880270 on Build GRCh38
rs1040642738 Chr.3: 8879879-8879879 on Build GRCh38
rs919766708 Chr.3: 8880139-8880139 on Build GRCh38
rs961438674 Chr.3: 8880146-8880146 on Build GRCh38
rs903436515 Chr.3: 8879891-8879891 on Build GRCh38
rs547213901 Chr.3: 8880019-8880019 on Build GRCh38
rs944741156 Chr.3: 8880255-8880255 on Build GRCh38
rs1013095670 Chr.3: 8879990-8879990 on Build GRCh38
rs535383116 Chr.3: 8879964-8879964 on Build GRCh38
rs557395301 Chr.3: 8880088-8880088 on Build GRCh38
rs34992215 Chr.3: 8880136-8880137 on Build GRCh38
rs910631957 Chr.3: 8880240-8880240 on Build GRCh38
rs932465942 Chr.3: 8880213-8880213 on Build GRCh38

View Full Product Details