Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 8985045-8985570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRGAP3
Entrez Gene ID
9901
Gene Name
SLIT-ROBO Rho GTPase activating protein 3
Gene Aliases
ARHGAP14, MEGAP, SRGAP2, WRP
UniGene
Hs.654743
Gene Chromosome Location
Chr.3: 8980591-9362979 on Build GRCh38
Transcripts
NM_001033117.2 NM_014850.3 XM_011534295.1 XM_011534296.1 XM_011534301.2 XM_011534303.1 XM_017007574.1 XM_017007575.1 XM_017007576.1 XM_017007577.1 XM_017007578.1 XM_017007579.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4992066 c.3263T>A p.F1088Y Chr.3: 8985556-8985556 on Build GRCh38
4813248 c.3185A>G p.K1062R Chr.3: 8985562-8985562 on Build GRCh38
4327775 c.3221C>T p.T1074I Chr.3: 8985526-8985526 on Build GRCh38
6510870 c.3204A>T p.S1068S Chr.3: 8985543-8985543 on Build GRCh38
4992067 c.3191T>A p.F1064Y Chr.3: 8985556-8985556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950132445 Chr.3: 8985093-8985094 on Build GRCh38
rs940701365 Chr.3: 8985256-8985256 on Build GRCh38
rs368794309 Chr.3: 8985495-8985495 on Build GRCh38
rs760849011 Chr.3: 8985498-8985498 on Build GRCh38
rs35501713 Chr.3: 8985252-8985253 on Build GRCh38
rs755646327 Chr.3: 8985525-8985525 on Build GRCh38
rs780611555 Chr.3: 8985514-8985514 on Build GRCh38
rs996743244 Chr.3: 8985352-8985352 on Build GRCh38
rs115133120 Chr.3: 8985358-8985358 on Build GRCh38
rs758892207 Chr.3: 8985508-8985508 on Build GRCh38
rs898399877 Chr.3: 8985118-8985119 on Build GRCh38
rs1009837639 Chr.3: 8985177-8985177 on Build GRCh38
rs780724549 Chr.3: 8985102-8985102 on Build GRCh38
rs1027963215 Chr.3: 8985131-8985131 on Build GRCh38
rs906953263 Chr.3: 8985445-8985445 on Build GRCh38
rs1020656065 Chr.3: 8985372-8985372 on Build GRCh38
rs759441251 Chr.3: 8985492-8985492 on Build GRCh38
rs747830121 Chr.3: 8985468-8985468 on Build GRCh38
rs777396931 Chr.3: 8985180-8985180 on Build GRCh38
rs1028247613 Chr.3: 8985357-8985357 on Build GRCh38
rs1051369777 Chr.3: 8985097-8985097 on Build GRCh38
rs1016562220 Chr.3: 8985512-8985512 on Build GRCh38
rs746476396 Chr.3: 8985205-8985205 on Build GRCh38
rs950095378 Chr.3: 8985261-8985261 on Build GRCh38
rs996797353 Chr.3: 8985120-8985120 on Build GRCh38
rs909446529 Chr.3: 8985240-8985240 on Build GRCh38
rs143155288 Chr.3: 8985228-8985228 on Build GRCh38
rs61740045 Chr.3: 8985541-8985541 on Build GRCh38
rs754144480 Chr.3: 8985501-8985501 on Build GRCh38
rs898344554 Chr.3: 8985321-8985321 on Build GRCh38
rs919720233 Chr.3: 8985529-8985529 on Build GRCh38
rs942650159 Chr.3: 8985371-8985371 on Build GRCh38
rs562275122 Chr.3: 8985212-8985212 on Build GRCh38
rs759347546 Chr.3: 8985291-8985291 on Build GRCh38
rs971714405 Chr.3: 8985463-8985463 on Build GRCh38
rs780734116 Chr.3: 8985198-8985198 on Build GRCh38
rs566209855 Chr.3: 8985347-8985347 on Build GRCh38
rs985019564 Chr.3: 8985237-8985237 on Build GRCh38
rs957551103 Chr.3: 8985448-8985448 on Build GRCh38
rs1038200199 Chr.3: 8985319-8985319 on Build GRCh38
rs749225938 Chr.3: 8985105-8985105 on Build GRCh38
rs879685288 Chr.3: 8985406-8985406 on Build GRCh38
rs770690937 Chr.3: 8985547-8985547 on Build GRCh38
rs531838303 Chr.3: 8985155-8985155 on Build GRCh38
rs186889 Chr.3: 8985134-8985134 on Build GRCh38
rs1016818299 Chr.3: 8985107-8985108 on Build GRCh38
rs916155543 Chr.3: 8985461-8985461 on Build GRCh38
rs73024065 Chr.3: 8985171-8985171 on Build GRCh38
rs749036920 Chr.3: 8985478-8985478 on Build GRCh38
rs915888466 Chr.3: 8985250-8985250 on Build GRCh38
rs148236663 Chr.3: 8985307-8985307 on Build GRCh38
rs886905837 Chr.3: 8985099-8985099 on Build GRCh38
rs991660901 Chr.3: 8985455-8985455 on Build GRCh38
rs539832416 Chr.3: 8985351-8985351 on Build GRCh38
rs139699619 Chr.3: 8985094-8985095 on Build GRCh38
rs73810778 Chr.3: 8985380-8985380 on Build GRCh38
rs772868465 Chr.3: 8985473-8985473 on Build GRCh38
rs750925547 Chr.3: 8985503-8985503 on Build GRCh38
rs78955855 Chr.3: 8985107-8985107 on Build GRCh38
rs774317420 Chr.3: 8985491-8985491 on Build GRCh38
rs752093030 Chr.3: 8985518-8985518 on Build GRCh38
rs1033109494 Chr.3: 8985420-8985420 on Build GRCh38
rs769367571 Chr.3: 8985469-8985469 on Build GRCh38
rs375961023 Chr.3: 8985488-8985488 on Build GRCh38
rs199538709 Chr.3: 8985500-8985500 on Build GRCh38
rs770654909 Chr.3: 8985236-8985236 on Build GRCh38
rs765566401 Chr.3: 8985502-8985502 on Build GRCh38
rs756291568 Chr.3: 8985090-8985093 on Build GRCh38
rs893039996 Chr.3: 8985397-8985397 on Build GRCh38
rs776224749 Chr.3: 8985243-8985243 on Build GRCh38
rs986528458 Chr.3: 8985152-8985152 on Build GRCh38
rs777368562 Chr.3: 8985540-8985540 on Build GRCh38
rs1038482508 Chr.3: 8985499-8985499 on Build GRCh38
rs1009892038 Chr.3: 8985404-8985404 on Build GRCh38
rs972053848 Chr.3: 8985263-8985263 on Build GRCh38
rs887189867 Chr.3: 8985279-8985279 on Build GRCh38
rs979178873 Chr.3: 8985412-8985412 on Build GRCh38

View Full Product Details