Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196584797-196585308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO45
Entrez Gene ID
200933
Gene Name
F-box protein 45
Gene Aliases
Fbx45
UniGene
Hs.169815
Gene Chromosome Location
Chr.3: 196568854-196589059 on Build GRCh38
Transcripts
NM_001105573.1

SNP Detail

SNP ID Chromosome Location
rs577473305 Chr.3: 196585175-196585175 on Build GRCh38
rs1037977345 Chr.3: 196585247-196585247 on Build GRCh38
rs1013933018 Chr.3: 196584891-196584894 on Build GRCh38
rs959615798 Chr.3: 196585229-196585229 on Build GRCh38
rs772104525 Chr.3: 196585069-196585069 on Build GRCh38
rs75324718 Chr.3: 196584984-196584984 on Build GRCh38
rs2014220 Chr.3: 196585253-196585253 on Build GRCh38
rs1020729375 Chr.3: 196585173-196585173 on Build GRCh38
rs920557445 Chr.3: 196584868-196584868 on Build GRCh38
rs954669816 Chr.3: 196585077-196585077 on Build GRCh38
rs76759144 Chr.3: 196585014-196585014 on Build GRCh38
rs533291492 Chr.3: 196585112-196585112 on Build GRCh38
rs543156681 Chr.3: 196584978-196584978 on Build GRCh38
rs774846019 Chr.3: 196584894-196584897 on Build GRCh38
rs762069072 Chr.3: 196584912-196584916 on Build GRCh38
rs913994425 Chr.3: 196585058-196585058 on Build GRCh38
rs563538810 Chr.3: 196585256-196585256 on Build GRCh38
rs973411411 Chr.3: 196584846-196584846 on Build GRCh38
rs895451055 Chr.3: 196584888-196584889 on Build GRCh38
rs139332492 Chr.3: 196585052-196585052 on Build GRCh38
rs11298661 Chr.3: 196584875-196584875 on Build GRCh38
rs1041274219 Chr.3: 196585041-196585041 on Build GRCh38
rs920609403 Chr.3: 196585249-196585249 on Build GRCh38
rs184685440 Chr.3: 196585053-196585053 on Build GRCh38
rs893447626 Chr.3: 196585089-196585089 on Build GRCh38
rs397877731 Chr.3: 196584886-196584886 on Build GRCh38
rs1026084403 Chr.3: 196584886-196584886 on Build GRCh38
rs760710268 Chr.3: 196585258-196585258 on Build GRCh38
rs929396013 Chr.3: 196584873-196584873 on Build GRCh38
rs748915848 Chr.3: 196584838-196584838 on Build GRCh38
rs943602135 Chr.3: 196585209-196585209 on Build GRCh38
rs973629156 Chr.3: 196585246-196585246 on Build GRCh38
rs1003751699 Chr.3: 196585182-196585182 on Build GRCh38
rs1034232033 Chr.3: 196585219-196585219 on Build GRCh38
rs200261609 Chr.3: 196584887-196584887 on Build GRCh38
rs373955685 Chr.3: 196585115-196585116 on Build GRCh38
rs987428040 Chr.3: 196585106-196585106 on Build GRCh38
rs1047779563 Chr.3: 196584924-196584924 on Build GRCh38
rs770495297 Chr.3: 196584957-196584957 on Build GRCh38

View Full Product Details