Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196585118-196585618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO45
Entrez Gene ID
200933
Gene Name
F-box protein 45
Gene Aliases
Fbx45
UniGene
Hs.169815
Gene Chromosome Location
Chr.3: 196568854-196589059 on Build GRCh38
Transcripts
NM_001105573.1

SNP Detail

SNP ID Chromosome Location
rs577473305 Chr.3: 196585175-196585175 on Build GRCh38
rs1037977345 Chr.3: 196585247-196585247 on Build GRCh38
rs747562708 Chr.3: 196585519-196585521 on Build GRCh38
rs917452014 Chr.3: 196585579-196585579 on Build GRCh38
rs893709244 Chr.3: 196585399-196585399 on Build GRCh38
rs920859223 Chr.3: 196585533-196585533 on Build GRCh38
rs974031416 Chr.3: 196585530-196585530 on Build GRCh38
rs959615798 Chr.3: 196585229-196585229 on Build GRCh38
rs910522711 Chr.3: 196585523-196585523 on Build GRCh38
rs906293127 Chr.3: 196585461-196585461 on Build GRCh38
rs2014220 Chr.3: 196585253-196585253 on Build GRCh38
rs949763242 Chr.3: 196585581-196585581 on Build GRCh38
rs1029487517 Chr.3: 196585460-196585460 on Build GRCh38
rs569170307 Chr.3: 196585526-196585526 on Build GRCh38
rs538989139 Chr.3: 196585447-196585447 on Build GRCh38
rs1020729375 Chr.3: 196585173-196585173 on Build GRCh38
rs1047918400 Chr.3: 196585563-196585563 on Build GRCh38
rs550557634 Chr.3: 196585469-196585470 on Build GRCh38
rs921055933 Chr.3: 196585386-196585386 on Build GRCh38
rs557132058 Chr.3: 196585468-196585468 on Build GRCh38
rs994992156 Chr.3: 196585548-196585548 on Build GRCh38
rs945083913 Chr.3: 196585425-196585425 on Build GRCh38
rs929585677 Chr.3: 196585552-196585552 on Build GRCh38
rs575750641 Chr.3: 196585474-196585474 on Build GRCh38
rs530517654 Chr.3: 196585350-196585350 on Build GRCh38
rs571594799 Chr.3: 196585444-196585444 on Build GRCh38
rs1053600546 Chr.3: 196585394-196585394 on Build GRCh38
rs563538810 Chr.3: 196585256-196585256 on Build GRCh38
rs546209199 Chr.3: 196585402-196585402 on Build GRCh38
rs768722307 Chr.3: 196585321-196585321 on Build GRCh38
rs1047474880 Chr.3: 196585333-196585333 on Build GRCh38
rs920609403 Chr.3: 196585249-196585249 on Build GRCh38
rs1044440659 Chr.3: 196585370-196585370 on Build GRCh38
rs895566109 Chr.3: 196585361-196585362 on Build GRCh38
rs1033868034 Chr.3: 196585498-196585498 on Build GRCh38
rs534283960 Chr.3: 196585389-196585389 on Build GRCh38
rs1032941346 Chr.3: 196585411-196585411 on Build GRCh38
rs555014744 Chr.3: 196585578-196585578 on Build GRCh38
rs760710268 Chr.3: 196585258-196585258 on Build GRCh38
rs943602135 Chr.3: 196585209-196585209 on Build GRCh38
rs973629156 Chr.3: 196585246-196585246 on Build GRCh38
rs567256566 Chr.3: 196585375-196585375 on Build GRCh38
rs1003751699 Chr.3: 196585182-196585182 on Build GRCh38
rs115443470 Chr.3: 196585362-196585362 on Build GRCh38
rs1034232033 Chr.3: 196585219-196585219 on Build GRCh38
rs954783463 Chr.3: 196585469-196585469 on Build GRCh38
rs976620193 Chr.3: 196585385-196585385 on Build GRCh38
rs1010026006 Chr.3: 196585452-196585452 on Build GRCh38

View Full Product Details